Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33145368-33145866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN2
Entrez Gene ID
113451
Gene Name
antizyme inhibitor 2
Gene Aliases
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp
UniGene
Hs.101807
Gene Chromosome Location
Chr.1: 33081110-33162288 on Build GRCh38
Transcripts
XM_017000167.1 XM_017000174.1 NM_018207.2 XM_011541705.2 XM_017001629.1 XM_017001630.1

Gene Symbol
TRIM62
Entrez Gene ID
55223
Gene Name
tripartite motif containing 62
Gene Aliases
DEAR1
UniGene
Hs.656006
Gene Chromosome Location
Chr.1: 33144603-33184774 on Build GRCh38
Transcripts
XM_017000167.1 XM_017000174.1 NM_018207.2 XM_011541705.2 XM_017001629.1 XM_017001630.1

SNP Detail

SNP ID Chromosome Location
rs529089966 Chr.1: 33145525-33145525 on Build GRCh38
rs901751034 Chr.1: 33145725-33145725 on Build GRCh38
rs904905848 Chr.1: 33145839-33145839 on Build GRCh38
rs953414128 Chr.1: 33145677-33145677 on Build GRCh38
rs572488342 Chr.1: 33145460-33145460 on Build GRCh38
rs567908067 Chr.1: 33145785-33145785 on Build GRCh38
rs536955221 Chr.1: 33145679-33145679 on Build GRCh38
rs919408096 Chr.1: 33145787-33145787 on Build GRCh38
rs566901710 Chr.1: 33145615-33145615 on Build GRCh38
rs561984789 Chr.1: 33145499-33145499 on Build GRCh38
rs913083899 Chr.1: 33145640-33145640 on Build GRCh38
rs72884397 Chr.1: 33145583-33145583 on Build GRCh38
rs942252293 Chr.1: 33145757-33145757 on Build GRCh38
rs761229926 Chr.1: 33145799-33145799 on Build GRCh38
rs1009393503 Chr.1: 33145556-33145556 on Build GRCh38
rs1029116274 Chr.1: 33145441-33145441 on Build GRCh38
rs1054228546 Chr.1: 33145638-33145638 on Build GRCh38
rs904878421 Chr.1: 33145431-33145431 on Build GRCh38
rs1006397756 Chr.1: 33145471-33145471 on Build GRCh38
rs75069742 Chr.1: 33145432-33145432 on Build GRCh38
rs1032226049 Chr.1: 33145459-33145459 on Build GRCh38
rs901864303 Chr.1: 33145406-33145406 on Build GRCh38
rs6682860 Chr.1: 33145749-33145749 on Build GRCh38
rs533505959 Chr.1: 33145598-33145598 on Build GRCh38
rs1040732565 Chr.1: 33145724-33145724 on Build GRCh38
rs945880433 Chr.1: 33145671-33145671 on Build GRCh38
rs551890810 Chr.1: 33145600-33145600 on Build GRCh38
rs150149479 Chr.1: 33145597-33145597 on Build GRCh38
rs996186948 Chr.1: 33145733-33145733 on Build GRCh38
rs540181610 Chr.1: 33145477-33145477 on Build GRCh38
rs779567991 Chr.1: 33145509-33145509 on Build GRCh38
rs915735209 Chr.1: 33145433-33145433 on Build GRCh38
rs758181490 Chr.1: 33145453-33145453 on Build GRCh38
rs764522902 Chr.1: 33145822-33145822 on Build GRCh38
rs971512120 Chr.1: 33145557-33145557 on Build GRCh38
rs544349516 Chr.1: 33145543-33145543 on Build GRCh38
rs957907544 Chr.1: 33145527-33145527 on Build GRCh38
rs527656853 Chr.1: 33145678-33145678 on Build GRCh38
rs185530104 Chr.1: 33145632-33145632 on Build GRCh38
rs745315010 Chr.1: 33145409-33145409 on Build GRCh38

View Full Product Details