Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35557704-35558209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCDN
Entrez Gene ID
23154
Gene Name
neurochondrin
Gene Aliases
-
UniGene
Hs.121870
Gene Chromosome Location
Chr.1: 35557792-35566779 on Build GRCh38
Transcripts
NM_001014839.1 NM_001014841.1 NM_014284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007362 c.13G>A p.D5N Chr.1: 35558203-35558203 on Build GRCh38
6318782 c.3G>T p.M1I Chr.1: 35558193-35558193 on Build GRCh38
5938723 c.8G>T p.C3F Chr.1: 35558198-35558198 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1021461997 Chr.1: 35557815-35557815 on Build GRCh38
rs764758035 Chr.1: 35558058-35558059 on Build GRCh38
rs763276981 Chr.1: 35557879-35557879 on Build GRCh38
rs931594469 Chr.1: 35557934-35557934 on Build GRCh38
rs773118200 Chr.1: 35557878-35557878 on Build GRCh38
rs905191826 Chr.1: 35557918-35557918 on Build GRCh38
rs968664338 Chr.1: 35557817-35557817 on Build GRCh38
rs1013131610 Chr.1: 35557812-35557812 on Build GRCh38
rs149137884 Chr.1: 35557990-35557990 on Build GRCh38
rs751202913 Chr.1: 35557920-35557920 on Build GRCh38
rs887094241 Chr.1: 35557790-35557790 on Build GRCh38
rs756045292 Chr.1: 35558144-35558144 on Build GRCh38
rs754464292 Chr.1: 35557927-35557927 on Build GRCh38
rs987258425 Chr.1: 35557842-35557842 on Build GRCh38
rs770658764 Chr.1: 35558171-35558171 on Build GRCh38
rs978451461 Chr.1: 35557827-35557827 on Build GRCh38
rs114784860 Chr.1: 35557823-35557823 on Build GRCh38
rs895708534 Chr.1: 35558006-35558006 on Build GRCh38
rs768786946 Chr.1: 35558186-35558186 on Build GRCh38
rs777750148 Chr.1: 35558151-35558151 on Build GRCh38
rs765249153 Chr.1: 35557840-35557841 on Build GRCh38
rs898783766 Chr.1: 35557727-35557727 on Build GRCh38
rs910255408 Chr.1: 35557859-35557859 on Build GRCh38
rs998785531 Chr.1: 35558086-35558086 on Build GRCh38
rs763108609 Chr.1: 35558185-35558187 on Build GRCh38
rs554686888 Chr.1: 35557789-35557789 on Build GRCh38
rs943124359 Chr.1: 35557865-35557865 on Build GRCh38
rs60735382 Chr.1: 35558070-35558071 on Build GRCh38
rs745978262 Chr.1: 35557811-35557811 on Build GRCh38
rs1010089528 Chr.1: 35558015-35558015 on Build GRCh38
rs780270207 Chr.1: 35558179-35558179 on Build GRCh38
rs74969398 Chr.1: 35558058-35558058 on Build GRCh38
rs953142736 Chr.1: 35558131-35558131 on Build GRCh38
rs1045433062 Chr.1: 35557767-35557768 on Build GRCh38
rs1002969501 Chr.1: 35557793-35557793 on Build GRCh38
rs930687911 Chr.1: 35557906-35557906 on Build GRCh38
rs112310803 Chr.1: 35558059-35558059 on Build GRCh38
rs1021494575 Chr.1: 35558056-35558056 on Build GRCh38
rs879117333 Chr.1: 35558070-35558070 on Build GRCh38
rs954406863 Chr.1: 35558140-35558140 on Build GRCh38
rs912084644 Chr.1: 35557849-35557849 on Build GRCh38
rs954497596 Chr.1: 35557834-35557834 on Build GRCh38
rs1020089266 Chr.1: 35558043-35558043 on Build GRCh38
rs543685892 Chr.1: 35557986-35557986 on Build GRCh38
rs187297307 Chr.1: 35557788-35557788 on Build GRCh38
rs887131396 Chr.1: 35557966-35557966 on Build GRCh38
rs1029040596 Chr.1: 35557835-35557835 on Build GRCh38
rs760765600 Chr.1: 35557762-35557762 on Build GRCh38
rs749075480 Chr.1: 35558155-35558155 on Build GRCh38
rs924302522 Chr.1: 35557832-35557832 on Build GRCh38
rs780695783 Chr.1: 35557962-35557962 on Build GRCh38
rs1025623175 Chr.1: 35557768-35557768 on Build GRCh38
rs765910923 Chr.1: 35557836-35557836 on Build GRCh38
rs894587597 Chr.1: 35558026-35558026 on Build GRCh38
rs572111894 Chr.1: 35557751-35557751 on Build GRCh38
rs372072924 Chr.1: 35558104-35558104 on Build GRCh38
rs1013005579 Chr.1: 35558038-35558038 on Build GRCh38
rs1035564226 Chr.1: 35557786-35557786 on Build GRCh38
rs780810516 Chr.1: 35557845-35557845 on Build GRCh38
rs747493625 Chr.1: 35557798-35557798 on Build GRCh38
rs958566139 Chr.1: 35557809-35557809 on Build GRCh38
rs920693781 Chr.1: 35557902-35557902 on Build GRCh38
rs11545629 Chr.1: 35558187-35558187 on Build GRCh38
rs937300077 Chr.1: 35557970-35557970 on Build GRCh38
rs75022183 Chr.1: 35558059-35558059 on Build GRCh38
rs761695031 Chr.1: 35557761-35557762 on Build GRCh38
rs1057223771 Chr.1: 35557995-35557995 on Build GRCh38
rs995712469 Chr.1: 35557742-35557742 on Build GRCh38
rs968527184 Chr.1: 35558060-35558060 on Build GRCh38
rs766025124 Chr.1: 35557914-35557914 on Build GRCh38
rs904278031 Chr.1: 35558054-35558054 on Build GRCh38
rs752566333 Chr.1: 35557967-35557967 on Build GRCh38
rs750786783 Chr.1: 35557843-35557843 on Build GRCh38
rs1000343223 Chr.1: 35558064-35558065 on Build GRCh38
rs996555747 Chr.1: 35557985-35558005 on Build GRCh38
rs987575886 Chr.1: 35558183-35558183 on Build GRCh38
rs953688468 Chr.1: 35557837-35557837 on Build GRCh38
rs374178077 Chr.1: 35558134-35558134 on Build GRCh38
rs577437486 Chr.1: 35558023-35558023 on Build GRCh38
rs899200035 Chr.1: 35557732-35557732 on Build GRCh38
rs940848217 Chr.1: 35557851-35557851 on Build GRCh38
rs987680489 Chr.1: 35557840-35557840 on Build GRCh38
rs920075020 Chr.1: 35557909-35557909 on Build GRCh38

View Full Product Details