Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35563689-35564194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCDN
Entrez Gene ID
23154
Gene Name
neurochondrin
Gene Aliases
-
UniGene
Hs.121870
Gene Chromosome Location
Chr.1: 35557792-35566779 on Build GRCh38
Transcripts
NM_001014839.1 NM_001014841.1 NM_014284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667296 c.1682G>T p.G561V Chr.1: 35563838-35563838 on Build GRCh38
908719 c.1561C>T p.R521C Chr.1: 35563768-35563768 on Build GRCh38
6063260 c.1734G>A p.R578R Chr.1: 35563890-35563890 on Build GRCh38
3667297 c.1631G>T p.G544V Chr.1: 35563838-35563838 on Build GRCh38
6063262 c.515G>A p.R172Q Chr.1: 35563890-35563890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911036691 Chr.1: 35563769-35563769 on Build GRCh38
rs906988059 Chr.1: 35563972-35563972 on Build GRCh38
rs756736463 Chr.1: 35563738-35563738 on Build GRCh38
rs918278780 Chr.1: 35563869-35563869 on Build GRCh38
rs141517113 Chr.1: 35563818-35563818 on Build GRCh38
rs761056103 Chr.1: 35563834-35563834 on Build GRCh38
rs1055327892 Chr.1: 35564047-35564047 on Build GRCh38
rs937054050 Chr.1: 35563974-35563974 on Build GRCh38
rs761372029 Chr.1: 35563890-35563890 on Build GRCh38
rs955448691 Chr.1: 35563753-35563753 on Build GRCh38
rs561451909 Chr.1: 35564161-35564161 on Build GRCh38
rs753439848 Chr.1: 35563725-35563725 on Build GRCh38
rs1012071630 Chr.1: 35564121-35564121 on Build GRCh38
rs749484417 Chr.1: 35563807-35563807 on Build GRCh38
rs779835266 Chr.1: 35563843-35563844 on Build GRCh38
rs745305905 Chr.1: 35563749-35563749 on Build GRCh38
rs573888247 Chr.1: 35564151-35564151 on Build GRCh38
rs751500260 Chr.1: 35563933-35563933 on Build GRCh38
rs778277282 Chr.1: 35563739-35563739 on Build GRCh38
rs746018023 Chr.1: 35563931-35563931 on Build GRCh38
rs777983350 Chr.1: 35563774-35563774 on Build GRCh38
rs758044776 Chr.1: 35563754-35563757 on Build GRCh38
rs369119475 Chr.1: 35563945-35563945 on Build GRCh38
rs147373266 Chr.1: 35563773-35563773 on Build GRCh38
rs891912603 Chr.1: 35564107-35564109 on Build GRCh38
rs766265626 Chr.1: 35563932-35563932 on Build GRCh38
rs978202606 Chr.1: 35563722-35563722 on Build GRCh38
rs1011175207 Chr.1: 35564064-35564064 on Build GRCh38
rs772452951 Chr.1: 35564070-35564070 on Build GRCh38
rs368544792 Chr.1: 35563758-35563758 on Build GRCh38
rs780016756 Chr.1: 35563754-35563754 on Build GRCh38
rs745981353 Chr.1: 35563815-35563815 on Build GRCh38
rs764383759 Chr.1: 35563840-35563840 on Build GRCh38
rs774285827 Chr.1: 35563812-35563812 on Build GRCh38
rs758301640 Chr.1: 35563746-35563746 on Build GRCh38
rs773834451 Chr.1: 35563720-35563720 on Build GRCh38
rs771411177 Chr.1: 35564066-35564066 on Build GRCh38
rs902519609 Chr.1: 35564154-35564154 on Build GRCh38
rs760497699 Chr.1: 35563904-35563904 on Build GRCh38
rs895286244 Chr.1: 35564062-35564062 on Build GRCh38
rs1043061449 Chr.1: 35564124-35564124 on Build GRCh38
rs746708014 Chr.1: 35563847-35563847 on Build GRCh38
rs12094935 Chr.1: 35563742-35563742 on Build GRCh38
rs900495510 Chr.1: 35564132-35564132 on Build GRCh38
rs1053662133 Chr.1: 35564073-35564074 on Build GRCh38
rs200493007 Chr.1: 35563825-35563825 on Build GRCh38
rs777114789 Chr.1: 35563852-35563852 on Build GRCh38
rs771241844 Chr.1: 35563809-35563809 on Build GRCh38
rs1044657827 Chr.1: 35563971-35563971 on Build GRCh38

View Full Product Details