Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59109789-59110323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKA2
Entrez Gene ID
348235
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
UniGene
Hs.463607
Gene Chromosome Location
Chr.17: 59109947-59155439 on Build GRCh38
Transcripts
NM_001100595.1 NM_182620.3 XM_011524745.2

SNP Detail

SNP ID Chromosome Location
rs530606723 Chr.17: 59109856-59109856 on Build GRCh38
rs566055710 Chr.17: 59110227-59110227 on Build GRCh38
rs950708792 Chr.17: 59110071-59110071 on Build GRCh38
rs371116921 Chr.17: 59109985-59109986 on Build GRCh38
rs952082227 Chr.17: 59110149-59110149 on Build GRCh38
rs933486677 Chr.17: 59110216-59110216 on Build GRCh38
rs1035379979 Chr.17: 59110105-59110105 on Build GRCh38
rs1053383164 Chr.17: 59109863-59109863 on Build GRCh38
rs947699742 Chr.17: 59110235-59110236 on Build GRCh38
rs1025990931 Chr.17: 59110034-59110034 on Build GRCh38
rs1003426715 Chr.17: 59110102-59110102 on Build GRCh38
rs923409207 Chr.17: 59110211-59110211 on Build GRCh38
rs910682492 Chr.17: 59110207-59110207 on Build GRCh38
rs748331025 Chr.17: 59110205-59110213 on Build GRCh38
rs547640963 Chr.17: 59110191-59110191 on Build GRCh38
rs893585044 Chr.17: 59109989-59109989 on Build GRCh38
rs377334994 Chr.17: 59110122-59110122 on Build GRCh38
rs1023400294 Chr.17: 59110030-59110030 on Build GRCh38
rs944954352 Chr.17: 59109891-59109891 on Build GRCh38
rs879355403 Chr.17: 59110264-59110264 on Build GRCh38
rs1052115449 Chr.17: 59109968-59109968 on Build GRCh38
rs758651370 Chr.17: 59110000-59110000 on Build GRCh38
rs1035471495 Chr.17: 59109984-59109985 on Build GRCh38
rs986233336 Chr.17: 59110186-59110186 on Build GRCh38
rs913474911 Chr.17: 59110224-59110224 on Build GRCh38
rs1041030288 Chr.17: 59109914-59109914 on Build GRCh38
rs909249491 Chr.17: 59110166-59110166 on Build GRCh38
rs994018406 Chr.17: 59110011-59110011 on Build GRCh38
rs944877307 Chr.17: 59110246-59110246 on Build GRCh38
rs772595846 Chr.17: 59110206-59110206 on Build GRCh38
rs548851465 Chr.17: 59110120-59110120 on Build GRCh38
rs934984401 Chr.17: 59110290-59110290 on Build GRCh38
rs182874831 Chr.17: 59110121-59110121 on Build GRCh38
rs1040622642 Chr.17: 59110261-59110261 on Build GRCh38
rs750410893 Chr.17: 59109880-59109880 on Build GRCh38
rs972085038 Chr.17: 59110124-59110124 on Build GRCh38
rs1050613522 Chr.17: 59109849-59109849 on Build GRCh38
rs1052020589 Chr.17: 59110294-59110294 on Build GRCh38
rs999633245 Chr.17: 59109954-59109954 on Build GRCh38
rs149114699 Chr.17: 59110147-59110147 on Build GRCh38
rs985194580 Chr.17: 59110137-59110137 on Build GRCh38
rs889297873 Chr.17: 59109853-59109853 on Build GRCh38
rs1021333089 Chr.17: 59109909-59109911 on Build GRCh38
rs900802616 Chr.17: 59109937-59109937 on Build GRCh38
rs933540482 Chr.17: 59109842-59109843 on Build GRCh38
rs969627933 Chr.17: 59110069-59110069 on Build GRCh38
rs1001270177 Chr.17: 59109981-59109981 on Build GRCh38
rs1053328597 Chr.17: 59110217-59110217 on Build GRCh38

View Full Product Details