Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59111074-59111574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKA2
Entrez Gene ID
348235
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
UniGene
Hs.463607
Gene Chromosome Location
Chr.17: 59109947-59155439 on Build GRCh38
Transcripts
NM_001100595.1 NM_182620.3 XM_011524745.2

SNP Detail

SNP ID Chromosome Location
rs1031748873 Chr.17: 59111212-59111212 on Build GRCh38
rs956203903 Chr.17: 59111228-59111228 on Build GRCh38
rs929708885 Chr.17: 59111203-59111203 on Build GRCh38
rs919049635 Chr.17: 59111338-59111338 on Build GRCh38
rs939039779 Chr.17: 59111291-59111291 on Build GRCh38
rs1048896572 Chr.17: 59111343-59111343 on Build GRCh38
rs929107564 Chr.17: 59111340-59111340 on Build GRCh38
rs1017998739 Chr.17: 59111104-59111104 on Build GRCh38
rs749021797 Chr.17: 59111207-59111207 on Build GRCh38
rs1034514009 Chr.17: 59111495-59111495 on Build GRCh38
rs909064102 Chr.17: 59111346-59111346 on Build GRCh38
rs992310526 Chr.17: 59111294-59111294 on Build GRCh38
rs542668501 Chr.17: 59111187-59111187 on Build GRCh38
rs941210956 Chr.17: 59111265-59111265 on Build GRCh38
rs978950888 Chr.17: 59111437-59111437 on Build GRCh38
rs970257583 Chr.17: 59111243-59111243 on Build GRCh38
rs992447455 Chr.17: 59111183-59111183 on Build GRCh38
rs997974474 Chr.17: 59111195-59111195 on Build GRCh38
rs187708854 Chr.17: 59111369-59111369 on Build GRCh38
rs71372875 Chr.17: 59111273-59111273 on Build GRCh38
rs771553238 Chr.17: 59111129-59111129 on Build GRCh38
rs764705066 Chr.17: 59111111-59111111 on Build GRCh38
rs958935663 Chr.17: 59111509-59111509 on Build GRCh38
rs1038958587 Chr.17: 59111381-59111381 on Build GRCh38
rs916889019 Chr.17: 59111517-59111517 on Build GRCh38
rs899745641 Chr.17: 59111296-59111296 on Build GRCh38
rs901831485 Chr.17: 59111447-59111447 on Build GRCh38
rs997563364 Chr.17: 59111494-59111494 on Build GRCh38
rs969254731 Chr.17: 59111408-59111408 on Build GRCh38
rs1020377860 Chr.17: 59111398-59111401 on Build GRCh38
rs1021817041 Chr.17: 59111239-59111239 on Build GRCh38
rs916943915 Chr.17: 59111192-59111192 on Build GRCh38
rs998135090 Chr.17: 59111304-59111304 on Build GRCh38
rs1030322788 Chr.17: 59111312-59111312 on Build GRCh38
rs192642304 Chr.17: 59111477-59111477 on Build GRCh38
rs929670091 Chr.17: 59111530-59111530 on Build GRCh38
rs990299335 Chr.17: 59111237-59111237 on Build GRCh38
rs760287602 Chr.17: 59111510-59111510 on Build GRCh38
rs151201885 Chr.17: 59111170-59111170 on Build GRCh38
rs550354455 Chr.17: 59111245-59111245 on Build GRCh38

View Full Product Details