Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62666364-62666871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRC2
Entrez Gene ID
9902
Gene Name
mannose receptor C type 2
Gene Aliases
CD280, CLEC13E, ENDO180, UPARAP
UniGene
Hs.7835
Gene Chromosome Location
Chr.17: 62627401-62693601 on Build GRCh38
Transcripts
NM_006039.4 XM_011525543.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2794386 c.911A>C p.D304A Chr.17: 62666808-62666808 on Build GRCh38
6025960 c.936G>A p.S312S Chr.17: 62666833-62666833 on Build GRCh38
6342455 c.873G>T p.G291G Chr.17: 62666770-62666770 on Build GRCh38
3773000 c.840C>T p.H280H Chr.17: 62666600-62666600 on Build GRCh38
4513361 c.935C>T p.S312L Chr.17: 62666832-62666832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759349430 Chr.17: 62666811-62666811 on Build GRCh38
rs780746693 Chr.17: 62666618-62666618 on Build GRCh38
rs751342786 Chr.17: 62666731-62666732 on Build GRCh38
rs772278283 Chr.17: 62666655-62666655 on Build GRCh38
rs765054987 Chr.17: 62666574-62666574 on Build GRCh38
rs375083798 Chr.17: 62666732-62666732 on Build GRCh38
rs770189073 Chr.17: 62666533-62666533 on Build GRCh38
rs749578061 Chr.17: 62666642-62666642 on Build GRCh38
rs778725977 Chr.17: 62666842-62666842 on Build GRCh38
rs762676746 Chr.17: 62666737-62666737 on Build GRCh38
rs374699186 Chr.17: 62666474-62666474 on Build GRCh38
rs983759116 Chr.17: 62666525-62666525 on Build GRCh38
rs771517970 Chr.17: 62666551-62666551 on Build GRCh38
rs775031075 Chr.17: 62666569-62666569 on Build GRCh38
rs780639886 Chr.17: 62666801-62666801 on Build GRCh38
rs759712103 Chr.17: 62666408-62666408 on Build GRCh38
rs763721017 Chr.17: 62666832-62666832 on Build GRCh38
rs767721644 Chr.17: 62666571-62666571 on Build GRCh38
rs143442592 Chr.17: 62666812-62666812 on Build GRCh38
rs117366246 Chr.17: 62666462-62666462 on Build GRCh38
rs891394408 Chr.17: 62666445-62666447 on Build GRCh38
rs373012647 Chr.17: 62666808-62666808 on Build GRCh38
rs201908329 Chr.17: 62666797-62666797 on Build GRCh38
rs367605863 Chr.17: 62666459-62666459 on Build GRCh38
rs945638657 Chr.17: 62666749-62666749 on Build GRCh38
rs374671479 Chr.17: 62666661-62666661 on Build GRCh38
rs752975300 Chr.17: 62666435-62666435 on Build GRCh38
rs112681150 Chr.17: 62666708-62666708 on Build GRCh38
rs767811631 Chr.17: 62666423-62666423 on Build GRCh38
rs759625924 Chr.17: 62666570-62666570 on Build GRCh38
rs1034122186 Chr.17: 62666407-62666407 on Build GRCh38
rs776702256 Chr.17: 62666806-62666806 on Build GRCh38
rs1024471740 Chr.17: 62666672-62666672 on Build GRCh38
rs1033438543 Chr.17: 62666484-62666484 on Build GRCh38
rs758312714 Chr.17: 62666466-62666466 on Build GRCh38
rs747812874 Chr.17: 62666493-62666493 on Build GRCh38
rs1042066603 Chr.17: 62666753-62666753 on Build GRCh38
rs1013974319 Chr.17: 62666666-62666666 on Build GRCh38
rs1000046747 Chr.17: 62666659-62666659 on Build GRCh38
rs1015195991 Chr.17: 62666738-62666738 on Build GRCh38
rs746991535 Chr.17: 62666802-62666802 on Build GRCh38
rs543936237 Chr.17: 62666389-62666389 on Build GRCh38
rs758241670 Chr.17: 62666591-62666591 on Build GRCh38
rs754471916 Chr.17: 62666617-62666617 on Build GRCh38
rs753544798 Chr.17: 62666798-62666798 on Build GRCh38
rs764179617 Chr.17: 62666722-62666722 on Build GRCh38
rs73333680 Chr.17: 62666445-62666445 on Build GRCh38
rs751510203 Chr.17: 62666607-62666607 on Build GRCh38
rs76093441 Chr.17: 62666705-62666705 on Build GRCh38
rs766371745 Chr.17: 62666490-62666493 on Build GRCh38
rs765003324 Chr.17: 62666460-62666460 on Build GRCh38
rs201556805 Chr.17: 62666647-62666647 on Build GRCh38
rs904418180 Chr.17: 62666658-62666658 on Build GRCh38
rs749780987 Chr.17: 62666550-62666550 on Build GRCh38
rs746440094 Chr.17: 62666648-62666648 on Build GRCh38
rs772653243 Chr.17: 62666800-62666800 on Build GRCh38
rs527564493 Chr.17: 62666682-62666682 on Build GRCh38
rs1051313621 Chr.17: 62666436-62666436 on Build GRCh38
rs757693226 Chr.17: 62666392-62666392 on Build GRCh38
rs376839199 Chr.17: 62666724-62666724 on Build GRCh38
rs370584741 Chr.17: 62666833-62666833 on Build GRCh38
rs766236786 Chr.17: 62666600-62666600 on Build GRCh38
rs777506014 Chr.17: 62666630-62666630 on Build GRCh38
rs562083245 Chr.17: 62666644-62666644 on Build GRCh38
rs751494028 Chr.17: 62666757-62666757 on Build GRCh38
rs768793719 Chr.17: 62666803-62666803 on Build GRCh38
rs750134590 Chr.17: 62666590-62666590 on Build GRCh38
rs752100861 Chr.17: 62666780-62666780 on Build GRCh38
rs779491205 Chr.17: 62666799-62666799 on Build GRCh38
rs755771987 Chr.17: 62666627-62666627 on Build GRCh38
rs139029815 Chr.17: 62666815-62666815 on Build GRCh38
rs774181832 Chr.17: 62666809-62666809 on Build GRCh38
rs746479929 Chr.17: 62666469-62666469 on Build GRCh38
rs149617729 Chr.17: 62666764-62666764 on Build GRCh38
rs368751421 Chr.17: 62666543-62666543 on Build GRCh38
rs747805837 Chr.17: 62666625-62666625 on Build GRCh38
rs757014647 Chr.17: 62666841-62666841 on Build GRCh38
rs547029864 Chr.17: 62666704-62666704 on Build GRCh38
rs987552256 Chr.17: 62666816-62666816 on Build GRCh38
rs766218305 Chr.17: 62666740-62666740 on Build GRCh38
rs866735634 Chr.17: 62666793-62666793 on Build GRCh38
rs372126711 Chr.17: 62666573-62666573 on Build GRCh38
rs267604980 Chr.17: 62666782-62666782 on Build GRCh38
rs548780279 Chr.17: 62666387-62666387 on Build GRCh38
rs766993462 Chr.17: 62666767-62666767 on Build GRCh38
rs903479830 Chr.17: 62666810-62666810 on Build GRCh38
rs34759830 Chr.17: 62666662-62666662 on Build GRCh38

View Full Product Details