Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45437263-45437755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM1
Entrez Gene ID
9842
Gene Name
pleckstrin homology and RUN domain containing M1
Gene Aliases
AP162, B2, OPTB6
UniGene
Hs.514242
Gene Chromosome Location
Chr.17: 45434262-45490780 on Build GRCh38
Transcripts
NM_014798.2 NR_027774.1 NR_027782.1 XM_006722201.3 XM_011525523.2 XM_011525524.1 XM_011525525.1 XM_011525526.1 XM_011525528.2 XM_017025451.1 XM_017025452.1 XM_017025453.1 XM_017025454.1

SNP Detail

SNP ID Chromosome Location
rs749893379 Chr.17: 45437286-45437286 on Build GRCh38
rs71373561 Chr.17: 45437598-45437598 on Build GRCh38
rs747339681 Chr.17: 45437685-45437685 on Build GRCh38
rs976266049 Chr.17: 45437382-45437382 on Build GRCh38
rs566243650 Chr.17: 45437300-45437300 on Build GRCh38
rs1009336470 Chr.17: 45437565-45437565 on Build GRCh38
rs966203466 Chr.17: 45437632-45437632 on Build GRCh38
rs948763107 Chr.17: 45437504-45437504 on Build GRCh38
rs115554660 Chr.17: 45437336-45437336 on Build GRCh38
rs956641190 Chr.17: 45437330-45437330 on Build GRCh38
rs986203409 Chr.17: 45437599-45437599 on Build GRCh38
rs918992757 Chr.17: 45437656-45437656 on Build GRCh38
rs1003792180 Chr.17: 45437326-45437326 on Build GRCh38
rs777026258 Chr.17: 45437453-45437453 on Build GRCh38
rs929502324 Chr.17: 45437658-45437658 on Build GRCh38
rs775629714 Chr.17: 45437726-45437726 on Build GRCh38
rs762942189 Chr.17: 45437733-45437733 on Build GRCh38
rs755669380 Chr.17: 45437297-45437297 on Build GRCh38
rs541599792 Chr.17: 45437664-45437664 on Build GRCh38
rs769981567 Chr.17: 45437724-45437724 on Build GRCh38
rs771446470 Chr.17: 45437702-45437702 on Build GRCh38
rs778185567 Chr.17: 45437352-45437352 on Build GRCh38
rs772731866 Chr.17: 45437431-45437431 on Build GRCh38
rs771211470 Chr.17: 45437413-45437413 on Build GRCh38
rs1022154030 Chr.17: 45437337-45437337 on Build GRCh38
rs1012902866 Chr.17: 45437293-45437293 on Build GRCh38
rs781725317 Chr.17: 45437714-45437714 on Build GRCh38
rs189306775 Chr.17: 45437555-45437555 on Build GRCh38
rs559310874 Chr.17: 45437640-45437640 on Build GRCh38
rs775474818 Chr.17: 45437569-45437569 on Build GRCh38
rs888034431 Chr.17: 45437496-45437496 on Build GRCh38
rs929448628 Chr.17: 45437433-45437433 on Build GRCh38
rs907281935 Chr.17: 45437560-45437561 on Build GRCh38
rs985807729 Chr.17: 45437359-45437359 on Build GRCh38
rs541240729 Chr.17: 45437634-45437634 on Build GRCh38
rs769898598 Chr.17: 45437564-45437564 on Build GRCh38
rs1017247449 Chr.17: 45437601-45437601 on Build GRCh38
rs751285366 Chr.17: 45437606-45437606 on Build GRCh38
rs1034773818 Chr.17: 45437329-45437335 on Build GRCh38
rs555039577 Chr.17: 45437363-45437363 on Build GRCh38
rs529881326 Chr.17: 45437661-45437661 on Build GRCh38
rs761624763 Chr.17: 45437625-45437625 on Build GRCh38
rs1049366203 Chr.17: 45437437-45437437 on Build GRCh38
rs759640600 Chr.17: 45437469-45437469 on Build GRCh38
rs1044806665 Chr.17: 45437731-45437731 on Build GRCh38
rs767105875 Chr.17: 45437629-45437629 on Build GRCh38
rs746074368 Chr.17: 45437719-45437719 on Build GRCh38
rs867849280 Chr.17: 45437562-45437562 on Build GRCh38
rs62064655 Chr.17: 45437588-45437588 on Build GRCh38
rs970625636 Chr.17: 45437565-45437587 on Build GRCh38
rs576553782 Chr.17: 45437372-45437372 on Build GRCh38
rs758882817 Chr.17: 45437684-45437684 on Build GRCh38
rs748656215 Chr.17: 45437301-45437301 on Build GRCh38
rs948956239 Chr.17: 45437729-45437729 on Build GRCh38

View Full Product Details