Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58419981-58420503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF5
Entrez Gene ID
124535
Gene Name
heat shock transcription factor 5
Gene Aliases
HSF 5, HSTF 5
UniGene
Hs.380061
Gene Chromosome Location
Chr.17: 58420167-58488401 on Build GRCh38
Transcripts
NM_001080439.2 XM_011524283.1

SNP Detail

SNP ID Chromosome Location
rs1031897997 Chr.17: 58420136-58420136 on Build GRCh38
rs367621557 Chr.17: 58420214-58420214 on Build GRCh38
rs982591755 Chr.17: 58420269-58420269 on Build GRCh38
rs932266538 Chr.17: 58420431-58420431 on Build GRCh38
rs559269173 Chr.17: 58420315-58420315 on Build GRCh38
rs1022082738 Chr.17: 58420242-58420242 on Build GRCh38
rs967216582 Chr.17: 58420098-58420098 on Build GRCh38
rs1011672134 Chr.17: 58420045-58420045 on Build GRCh38
rs958631484 Chr.17: 58420184-58420184 on Build GRCh38
rs980120935 Chr.17: 58420324-58420324 on Build GRCh38
rs182994363 Chr.17: 58420377-58420377 on Build GRCh38
rs1002647041 Chr.17: 58420145-58420145 on Build GRCh38
rs990146574 Chr.17: 58420239-58420239 on Build GRCh38
rs929239135 Chr.17: 58420388-58420388 on Build GRCh38
rs369614566 Chr.17: 58420116-58420116 on Build GRCh38
rs759949763 Chr.17: 58420022-58420022 on Build GRCh38
rs188691728 Chr.17: 58420379-58420379 on Build GRCh38
rs781050944 Chr.17: 58420097-58420097 on Build GRCh38
rs556352521 Chr.17: 58420038-58420039 on Build GRCh38
rs577812094 Chr.17: 58420358-58420358 on Build GRCh38
rs866575837 Chr.17: 58420222-58420222 on Build GRCh38
rs1020750360 Chr.17: 58420067-58420067 on Build GRCh38
rs573064990 Chr.17: 58420314-58420314 on Build GRCh38
rs952442845 Chr.17: 58420226-58420226 on Build GRCh38
rs1035592735 Chr.17: 58420149-58420149 on Build GRCh38
rs148719983 Chr.17: 58420025-58420025 on Build GRCh38
rs956517371 Chr.17: 58420167-58420167 on Build GRCh38
rs991967829 Chr.17: 58420369-58420369 on Build GRCh38
rs541545249 Chr.17: 58420146-58420146 on Build GRCh38
rs1048719762 Chr.17: 58420462-58420462 on Build GRCh38
rs940868239 Chr.17: 58420333-58420333 on Build GRCh38
rs991785754 Chr.17: 58420213-58420213 on Build GRCh38

View Full Product Details