Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76009454-76009961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
UniGene
Hs.500635
Gene Chromosome Location
Chr.17: 76006845-76027452 on Build GRCh38
Transcripts
NM_001320747.1 NM_001988.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656957 c.3429G>A p.E1143E Chr.17: 76009776-76009776 on Build GRCh38
3820605 c.3743T>G p.V1248G Chr.17: 76009462-76009462 on Build GRCh38
4397522 c.3405C>T p.I1135I Chr.17: 76009800-76009800 on Build GRCh38
7289184 c.3522C>T p.S1174S Chr.17: 76009683-76009683 on Build GRCh38
2742716 c.3313C>T p.R1105W Chr.17: 76009892-76009892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750661217 Chr.17: 76009698-76009698 on Build GRCh38
rs867816728 Chr.17: 76009775-76009775 on Build GRCh38
rs758317834 Chr.17: 76009664-76009664 on Build GRCh38
rs752000423 Chr.17: 76009573-76009573 on Build GRCh38
rs755721021 Chr.17: 76009581-76009581 on Build GRCh38
rs766924736 Chr.17: 76009571-76009571 on Build GRCh38
rs115192818 Chr.17: 76009891-76009891 on Build GRCh38
rs142112425 Chr.17: 76009682-76009682 on Build GRCh38
rs200935750 Chr.17: 76009893-76009893 on Build GRCh38
rs144952445 Chr.17: 76009622-76009622 on Build GRCh38
rs148100325 Chr.17: 76009480-76009480 on Build GRCh38
rs780153438 Chr.17: 76009838-76009838 on Build GRCh38
rs761466929 Chr.17: 76009817-76009817 on Build GRCh38
rs779434566 Chr.17: 76009764-76009764 on Build GRCh38
rs933951508 Chr.17: 76009887-76009887 on Build GRCh38
rs931441818 Chr.17: 76009528-76009528 on Build GRCh38
rs774565127 Chr.17: 76009706-76009706 on Build GRCh38
rs373559433 Chr.17: 76009845-76009845 on Build GRCh38
rs143282482 Chr.17: 76009857-76009857 on Build GRCh38
rs757898505 Chr.17: 76009819-76009819 on Build GRCh38
rs367555955 Chr.17: 76009542-76009542 on Build GRCh38
rs140332207 Chr.17: 76009704-76009704 on Build GRCh38
rs370565249 Chr.17: 76009593-76009593 on Build GRCh38
rs779240634 Chr.17: 76009831-76009831 on Build GRCh38
rs139861901 Chr.17: 76009550-76009550 on Build GRCh38
rs749319043 Chr.17: 76009521-76009521 on Build GRCh38
rs554283625 Chr.17: 76009478-76009478 on Build GRCh38
rs749584898 Chr.17: 76009779-76009779 on Build GRCh38
rs763527200 Chr.17: 76009570-76009570 on Build GRCh38
rs200731297 Chr.17: 76009529-76009529 on Build GRCh38
rs777457834 Chr.17: 76009856-76009856 on Build GRCh38
rs754775757 Chr.17: 76009773-76009773 on Build GRCh38
rs762898947 Chr.17: 76009621-76009621 on Build GRCh38
rs868026762 Chr.17: 76009802-76009802 on Build GRCh38
rs779911374 Chr.17: 76009680-76009680 on Build GRCh38
rs920183082 Chr.17: 76009499-76009499 on Build GRCh38
rs372152002 Chr.17: 76009812-76009812 on Build GRCh38
rs776196580 Chr.17: 76009728-76009728 on Build GRCh38
rs760501345 Chr.17: 76009864-76009864 on Build GRCh38
rs961806696 Chr.17: 76009866-76009866 on Build GRCh38
rs200577963 Chr.17: 76009661-76009661 on Build GRCh38
rs749877204 Chr.17: 76009738-76009738 on Build GRCh38
rs970400848 Chr.17: 76009583-76009583 on Build GRCh38
rs767967549 Chr.17: 76009575-76009575 on Build GRCh38
rs140510922 Chr.17: 76009737-76009737 on Build GRCh38
rs146508997 Chr.17: 76009721-76009721 on Build GRCh38
rs780949433 Chr.17: 76009497-76009497 on Build GRCh38
rs201966558 Chr.17: 76009614-76009614 on Build GRCh38
rs770179994 Chr.17: 76009930-76009930 on Build GRCh38
rs745685608 Chr.17: 76009607-76009607 on Build GRCh38
rs747537304 Chr.17: 76009844-76009844 on Build GRCh38
rs267605057 Chr.17: 76009691-76009691 on Build GRCh38
rs980355073 Chr.17: 76009906-76009906 on Build GRCh38
rs200046392 Chr.17: 76009541-76009541 on Build GRCh38
rs761919888 Chr.17: 76009543-76009543 on Build GRCh38
rs768065645 Chr.17: 76009644-76009644 on Build GRCh38
rs747530259 Chr.17: 76009800-76009800 on Build GRCh38
rs374903579 Chr.17: 76009859-76009859 on Build GRCh38
rs769430799 Chr.17: 76009792-76009800 on Build GRCh38
rs143353839 Chr.17: 76009580-76009580 on Build GRCh38
rs548121579 Chr.17: 76009848-76009848 on Build GRCh38
rs769457633 Chr.17: 76009849-76009849 on Build GRCh38
rs770411485 Chr.17: 76009850-76009850 on Build GRCh38
rs759509889 Chr.17: 76009826-76009826 on Build GRCh38
rs376792152 Chr.17: 76009561-76009561 on Build GRCh38
rs569510252 Chr.17: 76009778-76009778 on Build GRCh38
rs747762061 Chr.17: 76009573-76009573 on Build GRCh38
rs139261978 Chr.17: 76009900-76009900 on Build GRCh38
rs760392340 Chr.17: 76009656-76009656 on Build GRCh38
rs748460268 Chr.17: 76009615-76009615 on Build GRCh38
rs149596756 Chr.17: 76009829-76009829 on Build GRCh38
rs763830732 Chr.17: 76009727-76009727 on Build GRCh38
rs777362032 Chr.17: 76009794-76009796 on Build GRCh38
rs766186081 Chr.17: 76009765-76009765 on Build GRCh38
rs376838915 Chr.17: 76009825-76009825 on Build GRCh38
rs878892963 Chr.17: 76009743-76009743 on Build GRCh38
rs770762901 Chr.17: 76009522-76009522 on Build GRCh38
rs752994504 Chr.17: 76009645-76009645 on Build GRCh38
rs375850675 Chr.17: 76009730-76009730 on Build GRCh38
rs145575003 Chr.17: 76009719-76009719 on Build GRCh38
rs148648557 Chr.17: 76009640-76009640 on Build GRCh38
rs751160453 Chr.17: 76009603-76009603 on Build GRCh38
rs759408395 Chr.17: 76009855-76009855 on Build GRCh38
rs777119774 Chr.17: 76009520-76009520 on Build GRCh38
rs755469583 Chr.17: 76009894-76009894 on Build GRCh38
rs759928902 Chr.17: 76009574-76009574 on Build GRCh38
rs755461318 Chr.17: 76009755-76009755 on Build GRCh38
rs368969011 Chr.17: 76009827-76009827 on Build GRCh38
rs138147066 Chr.17: 76009818-76009818 on Build GRCh38
rs200154594 Chr.17: 76009799-76009799 on Build GRCh38
rs376653928 Chr.17: 76009904-76009904 on Build GRCh38
rs778783709 Chr.17: 76009600-76009600 on Build GRCh38
rs781628056 Chr.17: 76009896-76009896 on Build GRCh38
rs745879063 Chr.17: 76009797-76009797 on Build GRCh38
rs763959931 Chr.17: 76009872-76009872 on Build GRCh38
rs376995768 Chr.17: 76009683-76009683 on Build GRCh38
rs371423033 Chr.17: 76009832-76009832 on Build GRCh38
rs755695873 Chr.17: 76009498-76009498 on Build GRCh38
rs769982483 Chr.17: 76009544-76009544 on Build GRCh38
rs200855427 Chr.17: 76009662-76009662 on Build GRCh38
rs559827414 Chr.17: 76009629-76009629 on Build GRCh38
rs1038473817 Chr.17: 76009920-76009920 on Build GRCh38
rs751390093 Chr.17: 76009772-76009772 on Build GRCh38
rs776976233 Chr.17: 76009813-76009813 on Build GRCh38
rs758334386 Chr.17: 76009756-76009756 on Build GRCh38
rs147527679 Chr.17: 76009586-76009586 on Build GRCh38
rs139479728 Chr.17: 76009604-76009604 on Build GRCh38
rs771170711 Chr.17: 76009705-76009705 on Build GRCh38
rs866389061 Chr.17: 76009482-76009482 on Build GRCh38
rs375245505 Chr.17: 76009620-76009620 on Build GRCh38
rs375552944 Chr.17: 76009496-76009496 on Build GRCh38
rs757543596 Chr.17: 76009788-76009788 on Build GRCh38
rs369142937 Chr.17: 76009879-76009879 on Build GRCh38
rs150391336 Chr.17: 76009790-76009790 on Build GRCh38
rs141948489 Chr.17: 76009481-76009481 on Build GRCh38
rs144978675 Chr.17: 76009619-76009619 on Build GRCh38
rs368498514 Chr.17: 76009684-76009684 on Build GRCh38
rs75019874 Chr.17: 76009530-76009530 on Build GRCh38
rs368923796 Chr.17: 76009613-76009613 on Build GRCh38
rs202053602 Chr.17: 76009892-76009892 on Build GRCh38
rs746438412 Chr.17: 76009714-76009714 on Build GRCh38
rs149505943 Chr.17: 76009702-76009702 on Build GRCh38
rs1014644565 Chr.17: 76009649-76009649 on Build GRCh38

View Full Product Details