Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76734028-76734531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRSF2
Entrez Gene ID
6427
Gene Name
serine and arginine rich splicing factor 2
Gene Aliases
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
UniGene
Hs.584801
Gene Chromosome Location
Chr.17: 76734115-76737431 on Build GRCh38
Transcripts
NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

SNP Detail

SNP ID Chromosome Location
rs1011426456 Chr.17: 76734203-76734203 on Build GRCh38
rs899481613 Chr.17: 76734443-76734445 on Build GRCh38
rs13588 Chr.17: 76734504-76734504 on Build GRCh38
rs1020755775 Chr.17: 76734270-76734270 on Build GRCh38
rs929456721 Chr.17: 76734312-76734312 on Build GRCh38
rs961125264 Chr.17: 76734361-76734361 on Build GRCh38
rs963747170 Chr.17: 76734189-76734189 on Build GRCh38
rs905852075 Chr.17: 76734142-76734142 on Build GRCh38
rs1039477044 Chr.17: 76734410-76734411 on Build GRCh38
rs866734007 Chr.17: 76734294-76734294 on Build GRCh38
rs1035777123 Chr.17: 76734241-76734244 on Build GRCh38
rs1052396070 Chr.17: 76734315-76734315 on Build GRCh38
rs747864928 Chr.17: 76734274-76734274 on Build GRCh38
rs1046828960 Chr.17: 76734113-76734113 on Build GRCh38
rs746390663 Chr.17: 76734325-76734325 on Build GRCh38
rs980374717 Chr.17: 76734473-76734473 on Build GRCh38
rs942226100 Chr.17: 76734091-76734091 on Build GRCh38
rs563363305 Chr.17: 76734212-76734212 on Build GRCh38
rs1804421 Chr.17: 76734349-76734349 on Build GRCh38
rs1000770126 Chr.17: 76734278-76734278 on Build GRCh38
rs1045066344 Chr.17: 76734122-76734122 on Build GRCh38
rs533202486 Chr.17: 76734451-76734451 on Build GRCh38
rs960436160 Chr.17: 76734248-76734251 on Build GRCh38
rs938918942 Chr.17: 76734499-76734499 on Build GRCh38
rs1804420 Chr.17: 76734337-76734337 on Build GRCh38
rs920102113 Chr.17: 76734107-76734107 on Build GRCh38
rs547395426 Chr.17: 76734453-76734453 on Build GRCh38
rs771433469 Chr.17: 76734275-76734275 on Build GRCh38
rs184319144 Chr.17: 76734310-76734310 on Build GRCh38
rs1021512001 Chr.17: 76734214-76734214 on Build GRCh38
rs552150735 Chr.17: 76734304-76734304 on Build GRCh38
rs977250633 Chr.17: 76734224-76734224 on Build GRCh38
rs527632466 Chr.17: 76734362-76734362 on Build GRCh38
rs1011227135 Chr.17: 76734445-76734445 on Build GRCh38
rs769020268 Chr.17: 76734151-76734151 on Build GRCh38
rs865998185 Chr.17: 76734292-76734292 on Build GRCh38
rs931558771 Chr.17: 76734115-76734115 on Build GRCh38
rs1002911686 Chr.17: 76734159-76734159 on Build GRCh38
rs1057237498 Chr.17: 76734199-76734199 on Build GRCh38
rs143754278 Chr.17: 76734091-76734091 on Build GRCh38
rs11087 Chr.17: 76734323-76734323 on Build GRCh38
rs953705596 Chr.17: 76734309-76734309 on Build GRCh38
rs1018320595 Chr.17: 76734152-76734152 on Build GRCh38
rs991596887 Chr.17: 76734248-76734248 on Build GRCh38
rs1028225254 Chr.17: 76734283-76734283 on Build GRCh38
rs897966633 Chr.17: 76734096-76734097 on Build GRCh38
rs1037976831 Chr.17: 76734096-76734096 on Build GRCh38
rs779413042 Chr.17: 76734146-76734149 on Build GRCh38
rs747368913 Chr.17: 76734286-76734286 on Build GRCh38
rs62086783 Chr.17: 76734071-76734071 on Build GRCh38
rs891848295 Chr.17: 76734205-76734205 on Build GRCh38
rs773807370 Chr.17: 76734260-76734260 on Build GRCh38
rs780366726 Chr.17: 76734076-76734076 on Build GRCh38
rs532418867 Chr.17: 76734257-76734257 on Build GRCh38
rs542035458 Chr.17: 76734450-76734450 on Build GRCh38
rs567412711 Chr.17: 76734480-76734480 on Build GRCh38
rs920088421 Chr.17: 76734379-76734379 on Build GRCh38
rs931502593 Chr.17: 76734444-76734444 on Build GRCh38
rs11550964 Chr.17: 76734393-76734393 on Build GRCh38
rs967172619 Chr.17: 76734216-76734216 on Build GRCh38
rs117994889 Chr.17: 76734065-76734065 on Build GRCh38
rs943744573 Chr.17: 76734363-76734363 on Build GRCh38
rs28693812 Chr.17: 76734106-76734106 on Build GRCh38
rs767271121 Chr.17: 76734095-76734095 on Build GRCh38
rs890908602 Chr.17: 76734140-76734140 on Build GRCh38

View Full Product Details