Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78426744-78427251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH17
Entrez Gene ID
8632
Gene Name
dynein axonemal heavy chain 17
Gene Aliases
DNAHL1, DNEL2
UniGene
Hs.375975
Gene Chromosome Location
Chr.17: 78423697-78577430 on Build GRCh38
Transcripts
NM_173628.3 XM_011525416.2 XM_017025261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755927 c.12666C>T p.Y4222Y Chr.17: 78427016-78427016 on Build GRCh38
3755929 c.12612G>A p.P4204P Chr.17: 78427070-78427070 on Build GRCh38
3755926 c.12750C>T p.Y4250Y Chr.17: 78427016-78427016 on Build GRCh38
3522822 c.12799G>A p.E4267K Chr.17: 78426967-78426967 on Build GRCh38
437655 c.12673G>T p.V4225F Chr.17: 78427009-78427009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369747701 Chr.17: 78426960-78426960 on Build GRCh38
rs945998092 Chr.17: 78426783-78426783 on Build GRCh38
rs7219936 Chr.17: 78427120-78427120 on Build GRCh38
rs369106569 Chr.17: 78427115-78427115 on Build GRCh38
rs372337837 Chr.17: 78426907-78426907 on Build GRCh38
rs776532870 Chr.17: 78427145-78427145 on Build GRCh38
rs377493055 Chr.17: 78427182-78427182 on Build GRCh38
rs142613771 Chr.17: 78427042-78427042 on Build GRCh38
rs566363952 Chr.17: 78426841-78426841 on Build GRCh38
rs1014639739 Chr.17: 78427054-78427054 on Build GRCh38
rs776126591 Chr.17: 78427071-78427071 on Build GRCh38
rs74001316 Chr.17: 78426897-78426897 on Build GRCh38
rs887701285 Chr.17: 78427216-78427216 on Build GRCh38
rs760533197 Chr.17: 78427056-78427056 on Build GRCh38
rs35604481 Chr.17: 78426961-78426961 on Build GRCh38
rs773086862 Chr.17: 78426984-78426984 on Build GRCh38
rs911759121 Chr.17: 78426844-78426859 on Build GRCh38
rs765544792 Chr.17: 78426958-78426958 on Build GRCh38
rs1032956187 Chr.17: 78427188-78427188 on Build GRCh38
rs529339411 Chr.17: 78427121-78427121 on Build GRCh38
rs771038744 Chr.17: 78427091-78427091 on Build GRCh38
rs960054344 Chr.17: 78426921-78426921 on Build GRCh38
rs144209606 Chr.17: 78427087-78427087 on Build GRCh38
rs765170521 Chr.17: 78427210-78427210 on Build GRCh38
rs752889400 Chr.17: 78427095-78427095 on Build GRCh38
rs59163554 Chr.17: 78427100-78427100 on Build GRCh38
rs35782078 Chr.17: 78426977-78426977 on Build GRCh38
rs1001793568 Chr.17: 78427011-78427011 on Build GRCh38
rs777291478 Chr.17: 78426908-78426908 on Build GRCh38
rs768408289 Chr.17: 78426984-78426985 on Build GRCh38
rs758437954 Chr.17: 78426795-78426795 on Build GRCh38
rs965768945 Chr.17: 78427057-78427057 on Build GRCh38
rs746572317 Chr.17: 78427131-78427131 on Build GRCh38
rs762938352 Chr.17: 78426919-78426919 on Build GRCh38
rs868309742 Chr.17: 78427041-78427041 on Build GRCh38
rs765818142 Chr.17: 78426920-78426920 on Build GRCh38
rs749184601 Chr.17: 78426986-78426986 on Build GRCh38
rs1001436675 Chr.17: 78426862-78426862 on Build GRCh38
rs752134298 Chr.17: 78427088-78427088 on Build GRCh38
rs770607295 Chr.17: 78427040-78427040 on Build GRCh38
rs1050177782 Chr.17: 78426823-78426823 on Build GRCh38
rs765598027 Chr.17: 78427012-78427012 on Build GRCh38
rs752983830 Chr.17: 78427031-78427031 on Build GRCh38
rs759905197 Chr.17: 78427090-78427090 on Build GRCh38
rs1027073635 Chr.17: 78426868-78426868 on Build GRCh38
rs771929306 Chr.17: 78427125-78427125 on Build GRCh38
rs763416624 Chr.17: 78427073-78427073 on Build GRCh38
rs747202307 Chr.17: 78426895-78426895 on Build GRCh38
rs374360300 Chr.17: 78426992-78426992 on Build GRCh38
rs746959699 Chr.17: 78426947-78426949 on Build GRCh38
rs911730049 Chr.17: 78427219-78427219 on Build GRCh38
rs3209030 Chr.17: 78427070-78427070 on Build GRCh38
rs755121094 Chr.17: 78427029-78427029 on Build GRCh38
rs750949715 Chr.17: 78426959-78426959 on Build GRCh38
rs749162191 Chr.17: 78427037-78427037 on Build GRCh38
rs147136333 Chr.17: 78426931-78426931 on Build GRCh38
rs758960368 Chr.17: 78426947-78426947 on Build GRCh38
rs759131475 Chr.17: 78427003-78427003 on Build GRCh38
rs978709231 Chr.17: 78426970-78426970 on Build GRCh38
rs768990159 Chr.17: 78426896-78426896 on Build GRCh38
rs751183775 Chr.17: 78426913-78426913 on Build GRCh38
rs763631400 Chr.17: 78426900-78426900 on Build GRCh38
rs769736657 Chr.17: 78426979-78426979 on Build GRCh38
rs568854600 Chr.17: 78426830-78426830 on Build GRCh38
rs754263873 Chr.17: 78426785-78426785 on Build GRCh38
rs181371014 Chr.17: 78426953-78426953 on Build GRCh38
rs564518198 Chr.17: 78427055-78427055 on Build GRCh38
rs72914885 Chr.17: 78426791-78426791 on Build GRCh38
rs754991384 Chr.17: 78426974-78426974 on Build GRCh38
rs756334417 Chr.17: 78427032-78427032 on Build GRCh38
rs747710526 Chr.17: 78427148-78427148 on Build GRCh38
rs765152057 Chr.17: 78426832-78426832 on Build GRCh38
rs779552330 Chr.17: 78427005-78427005 on Build GRCh38
rs535297612 Chr.17: 78426792-78426792 on Build GRCh38
rs1053167708 Chr.17: 78426833-78426833 on Build GRCh38
rs1009010038 Chr.17: 78426850-78426850 on Build GRCh38
rs976917461 Chr.17: 78426943-78426943 on Build GRCh38
rs913168828 Chr.17: 78426767-78426767 on Build GRCh38
rs190184449 Chr.17: 78426856-78426856 on Build GRCh38
rs372755956 Chr.17: 78427053-78427053 on Build GRCh38
rs149001523 Chr.17: 78427209-78427209 on Build GRCh38
rs754598879 Chr.17: 78426775-78426775 on Build GRCh38
rs773842617 Chr.17: 78426945-78426945 on Build GRCh38
rs776523015 Chr.17: 78427018-78427020 on Build GRCh38
rs897379096 Chr.17: 78426853-78426853 on Build GRCh38
rs757600591 Chr.17: 78426789-78426789 on Build GRCh38
rs766959912 Chr.17: 78427027-78427027 on Build GRCh38
rs771590353 Chr.17: 78427052-78427052 on Build GRCh38
rs751982638 Chr.17: 78427028-78427028 on Build GRCh38
rs780131093 Chr.17: 78426889-78426889 on Build GRCh38
rs373502157 Chr.17: 78427124-78427124 on Build GRCh38
rs190850315 Chr.17: 78427225-78427225 on Build GRCh38
rs775154419 Chr.17: 78427006-78427006 on Build GRCh38
rs376964362 Chr.17: 78427030-78427030 on Build GRCh38
rs771341971 Chr.17: 78427170-78427170 on Build GRCh38
rs760269542 Chr.17: 78427007-78427007 on Build GRCh38
rs780515103 Chr.17: 78426978-78426978 on Build GRCh38
rs145851504 Chr.17: 78427196-78427196 on Build GRCh38
rs751196497 Chr.17: 78426771-78426771 on Build GRCh38
rs1020696352 Chr.17: 78427147-78427147 on Build GRCh38
rs2271613 Chr.17: 78427016-78427016 on Build GRCh38
rs769613492 Chr.17: 78427173-78427173 on Build GRCh38
rs904492783 Chr.17: 78426831-78426831 on Build GRCh38
rs776381202 Chr.17: 78427135-78427135 on Build GRCh38
rs779955162 Chr.17: 78427130-78427130 on Build GRCh38
rs768124397 Chr.17: 78427134-78427134 on Build GRCh38
rs772213018 Chr.17: 78427049-78427049 on Build GRCh38
rs72914891 Chr.17: 78426852-78426852 on Build GRCh38
rs72914887 Chr.17: 78426822-78426822 on Build GRCh38
rs201985449 Chr.17: 78427099-78427099 on Build GRCh38
rs138740986 Chr.17: 78426954-78426954 on Build GRCh38
rs147406428 Chr.17: 78426990-78426990 on Build GRCh38
rs112803758 Chr.17: 78426860-78426860 on Build GRCh38
rs562678545 Chr.17: 78426957-78426957 on Build GRCh38
rs764367691 Chr.17: 78426776-78426776 on Build GRCh38
rs749954285 Chr.17: 78427156-78427156 on Build GRCh38
rs35081093 Chr.17: 78427044-78427044 on Build GRCh38
rs1049403700 Chr.17: 78427194-78427194 on Build GRCh38
rs200227498 Chr.17: 78427134-78427135 on Build GRCh38
rs372351110 Chr.17: 78427015-78427015 on Build GRCh38
rs967666308 Chr.17: 78426967-78426967 on Build GRCh38

View Full Product Details