Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36093016-36093520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL2
Entrez Gene ID
54936
Gene Name
ADP-ribosylhydrolase like 2
Gene Aliases
ARH3
UniGene
Hs.18021
Gene Chromosome Location
Chr.1: 36088852-36093932 on Build GRCh38
Transcripts
NM_017825.2 XM_011541636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299580 c.1072C>T p.R358C Chr.1: 36093366-36093366 on Build GRCh38
6224000 c.846C>T p.Y282Y Chr.1: 36093140-36093140 on Build GRCh38
6262998 c.808G>T p.G270C Chr.1: 36093102-36093102 on Build GRCh38
6595360 c.1019A>G p.Q340R Chr.1: 36093313-36093313 on Build GRCh38
459256 c.1009G>A p.E337K Chr.1: 36093303-36093303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560312697 Chr.1: 36093366-36093366 on Build GRCh38
rs139356242 Chr.1: 36093367-36093367 on Build GRCh38
rs375995036 Chr.1: 36093163-36093163 on Build GRCh38
rs760855749 Chr.1: 36093335-36093335 on Build GRCh38
rs372701793 Chr.1: 36093162-36093162 on Build GRCh38
rs750729168 Chr.1: 36093058-36093058 on Build GRCh38
rs775626747 Chr.1: 36093091-36093091 on Build GRCh38
rs769577354 Chr.1: 36093328-36093328 on Build GRCh38
rs558472367 Chr.1: 36093461-36093461 on Build GRCh38
rs752146820 Chr.1: 36093359-36093359 on Build GRCh38
rs571967173 Chr.1: 36093355-36093355 on Build GRCh38
rs766791824 Chr.1: 36093227-36093229 on Build GRCh38
rs369081688 Chr.1: 36093131-36093131 on Build GRCh38
rs199766545 Chr.1: 36093389-36093389 on Build GRCh38
rs200099878 Chr.1: 36093301-36093301 on Build GRCh38
rs1017590703 Chr.1: 36093159-36093159 on Build GRCh38
rs763109907 Chr.1: 36093121-36093121 on Build GRCh38
rs201735454 Chr.1: 36093298-36093298 on Build GRCh38
rs768450606 Chr.1: 36093191-36093191 on Build GRCh38
rs778705043 Chr.1: 36093263-36093263 on Build GRCh38
rs766568691 Chr.1: 36093227-36093227 on Build GRCh38
rs536830210 Chr.1: 36093443-36093443 on Build GRCh38
rs753145470 Chr.1: 36093143-36093143 on Build GRCh38
rs146218730 Chr.1: 36093225-36093225 on Build GRCh38
rs750787943 Chr.1: 36093414-36093414 on Build GRCh38
rs1044762077 Chr.1: 36093462-36093462 on Build GRCh38
rs183554661 Chr.1: 36093384-36093384 on Build GRCh38
rs1027988726 Chr.1: 36093309-36093309 on Build GRCh38
rs776682941 Chr.1: 36093305-36093305 on Build GRCh38
rs764494397 Chr.1: 36093340-36093340 on Build GRCh38
rs767594707 Chr.1: 36093237-36093237 on Build GRCh38
rs904731559 Chr.1: 36093116-36093116 on Build GRCh38
rs750603133 Chr.1: 36093107-36093107 on Build GRCh38
rs763802671 Chr.1: 36093103-36093103 on Build GRCh38
rs759541902 Chr.1: 36093233-36093233 on Build GRCh38
rs538677420 Chr.1: 36093365-36093365 on Build GRCh38
rs745401730 Chr.1: 36093266-36093266 on Build GRCh38
rs567716077 Chr.1: 36093422-36093422 on Build GRCh38
rs774494380 Chr.1: 36093330-36093330 on Build GRCh38
rs111850122 Chr.1: 36093075-36093075 on Build GRCh38
rs764701148 Chr.1: 36093253-36093253 on Build GRCh38
rs373961966 Chr.1: 36093190-36093190 on Build GRCh38
rs745358206 Chr.1: 36093164-36093164 on Build GRCh38
rs768003962 Chr.1: 36093132-36093132 on Build GRCh38
rs370323258 Chr.1: 36093392-36093392 on Build GRCh38
rs754383396 Chr.1: 36093430-36093430 on Build GRCh38
rs527784721 Chr.1: 36093160-36093160 on Build GRCh38
rs554615991 Chr.1: 36093397-36093397 on Build GRCh38
rs755555967 Chr.1: 36093403-36093404 on Build GRCh38
rs145383937 Chr.1: 36093371-36093371 on Build GRCh38
rs776323704 Chr.1: 36093196-36093196 on Build GRCh38
rs758157109 Chr.1: 36093359-36093359 on Build GRCh38
rs1034890606 Chr.1: 36093154-36093154 on Build GRCh38
rs549427499 Chr.1: 36093247-36093247 on Build GRCh38
rs760667264 Chr.1: 36093383-36093383 on Build GRCh38
rs756440961 Chr.1: 36093150-36093150 on Build GRCh38
rs118183123 Chr.1: 36093429-36093429 on Build GRCh38
rs778513008 Chr.1: 36093064-36093065 on Build GRCh38
rs560072611 Chr.1: 36093149-36093149 on Build GRCh38
rs905097396 Chr.1: 36093490-36093490 on Build GRCh38
rs756609050 Chr.1: 36093251-36093251 on Build GRCh38
rs144031883 Chr.1: 36093256-36093256 on Build GRCh38
rs1010296195 Chr.1: 36093285-36093285 on Build GRCh38
rs775431453 Chr.1: 36093168-36093168 on Build GRCh38
rs763426981 Chr.1: 36093421-36093421 on Build GRCh38
rs754435618 Chr.1: 36093345-36093345 on Build GRCh38
rs1043361664 Chr.1: 36093478-36093478 on Build GRCh38
rs371064687 Chr.1: 36093374-36093374 on Build GRCh38
rs551168109 Chr.1: 36093457-36093457 on Build GRCh38
rs750048109 Chr.1: 36093357-36093357 on Build GRCh38
rs779455271 Chr.1: 36093289-36093289 on Build GRCh38
rs577645269 Chr.1: 36093122-36093122 on Build GRCh38
rs760515017 Chr.1: 36093096-36093096 on Build GRCh38
rs765659747 Chr.1: 36093405-36093405 on Build GRCh38
rs927799465 Chr.1: 36093386-36093386 on Build GRCh38
rs759156973 Chr.1: 36093072-36093072 on Build GRCh38
rs531916765 Chr.1: 36093332-36093332 on Build GRCh38
rs757925811 Chr.1: 36093287-36093287 on Build GRCh38
rs375689539 Chr.1: 36093379-36093379 on Build GRCh38
rs771775723 Chr.1: 36093167-36093167 on Build GRCh38
rs780772725 Chr.1: 36093433-36093433 on Build GRCh38
rs550523074 Chr.1: 36093333-36093333 on Build GRCh38
rs898833280 Chr.1: 36093045-36093045 on Build GRCh38
rs543370113 Chr.1: 36093044-36093044 on Build GRCh38
rs375098574 Chr.1: 36093402-36093402 on Build GRCh38
rs561337860 Chr.1: 36093278-36093278 on Build GRCh38
rs930295798 Chr.1: 36093456-36093456 on Build GRCh38
rs779798854 Chr.1: 36093364-36093364 on Build GRCh38
rs778017668 Chr.1: 36093158-36093158 on Build GRCh38
rs771497461 Chr.1: 36093467-36093467 on Build GRCh38
rs774027150 Chr.1: 36093071-36093071 on Build GRCh38
rs747928436 Chr.1: 36093320-36093320 on Build GRCh38
rs200293603 Chr.1: 36093087-36093087 on Build GRCh38
rs774618602 Chr.1: 36093229-36093229 on Build GRCh38
rs545002185 Chr.1: 36093128-36093128 on Build GRCh38
rs754129757 Chr.1: 36093255-36093255 on Build GRCh38

View Full Product Details