Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39561981-39562488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PABPC4
Entrez Gene ID
8761
Gene Name
poly(A) binding protein cytoplasmic 4
Gene Aliases
APP-1, APP1, PABP4, iPABP
UniGene
Hs.169900
Gene Chromosome Location
Chr.1: 39560813-39576849 on Build GRCh38
Transcripts
NM_001135653.1 NM_001135654.1 NM_003819.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473947 c.1836C>T p.L612L Chr.1: 39562082-39562082 on Build GRCh38
4615539 c.1741_1742insC p.Q581fs*37 Chr.1: 39562343-39562344 on Build GRCh38
4622884 c.1770C>T p.R590R Chr.1: 39562196-39562196 on Build GRCh38
4471409 c.1670C>T p.S557F Chr.1: 39562367-39562367 on Build GRCh38
1342550 c.1693delC p.Q565fs*33 Chr.1: 39562344-39562344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770896374 Chr.1: 39562413-39562413 on Build GRCh38
rs781426604 Chr.1: 39562391-39562391 on Build GRCh38
rs571998042 Chr.1: 39562452-39562452 on Build GRCh38
rs747353016 Chr.1: 39562223-39562224 on Build GRCh38
rs758962869 Chr.1: 39562347-39562347 on Build GRCh38
rs773182596 Chr.1: 39562076-39562076 on Build GRCh38
rs143270321 Chr.1: 39562402-39562402 on Build GRCh38
rs762402081 Chr.1: 39562079-39562079 on Build GRCh38
rs996720628 Chr.1: 39562002-39562002 on Build GRCh38
rs769512992 Chr.1: 39562403-39562403 on Build GRCh38
rs758488606 Chr.1: 39562136-39562136 on Build GRCh38
rs779538157 Chr.1: 39562093-39562093 on Build GRCh38
rs763948657 Chr.1: 39562145-39562145 on Build GRCh38
rs767162555 Chr.1: 39562287-39562287 on Build GRCh38
rs767374388 Chr.1: 39562050-39562050 on Build GRCh38
rs200233849 Chr.1: 39562432-39562432 on Build GRCh38
rs201090188 Chr.1: 39562375-39562375 on Build GRCh38
rs765907104 Chr.1: 39562080-39562080 on Build GRCh38
rs777556247 Chr.1: 39562407-39562407 on Build GRCh38
rs201776412 Chr.1: 39562235-39562235 on Build GRCh38
rs747916449 Chr.1: 39562198-39562198 on Build GRCh38
rs781310791 Chr.1: 39562193-39562193 on Build GRCh38
rs776477021 Chr.1: 39562062-39562062 on Build GRCh38
rs756392541 Chr.1: 39562023-39562023 on Build GRCh38
rs775695699 Chr.1: 39562061-39562062 on Build GRCh38
rs531871119 Chr.1: 39562434-39562434 on Build GRCh38
rs1022495165 Chr.1: 39562019-39562019 on Build GRCh38
rs369685416 Chr.1: 39562023-39562024 on Build GRCh38
rs189716705 Chr.1: 39562216-39562216 on Build GRCh38
rs773710999 Chr.1: 39562449-39562449 on Build GRCh38
rs751631876 Chr.1: 39562349-39562349 on Build GRCh38
rs779626325 Chr.1: 39562039-39562039 on Build GRCh38
rs201997383 Chr.1: 39562345-39562345 on Build GRCh38
rs774872253 Chr.1: 39562118-39562118 on Build GRCh38
rs948003423 Chr.1: 39562178-39562178 on Build GRCh38
rs891018361 Chr.1: 39562459-39562459 on Build GRCh38
rs186210881 Chr.1: 39562437-39562437 on Build GRCh38
rs752029483 Chr.1: 39562457-39562457 on Build GRCh38
rs745357942 Chr.1: 39562420-39562420 on Build GRCh38
rs775523610 Chr.1: 39562053-39562053 on Build GRCh38
rs763900396 Chr.1: 39562445-39562445 on Build GRCh38
rs768279522 Chr.1: 39562056-39562056 on Build GRCh38
rs753417685 Chr.1: 39562190-39562190 on Build GRCh38
rs751249081 Chr.1: 39562022-39562022 on Build GRCh38
rs954500999 Chr.1: 39562163-39562163 on Build GRCh38
rs182344251 Chr.1: 39562233-39562233 on Build GRCh38
rs745907716 Chr.1: 39562044-39562044 on Build GRCh38
rs922879950 Chr.1: 39562127-39562127 on Build GRCh38
rs947922833 Chr.1: 39562390-39562390 on Build GRCh38
rs757732034 Chr.1: 39562092-39562092 on Build GRCh38
rs369911540 Chr.1: 39562082-39562082 on Build GRCh38
rs987165431 Chr.1: 39562165-39562165 on Build GRCh38
rs201782350 Chr.1: 39562055-39562055 on Build GRCh38
rs759033907 Chr.1: 39562272-39562272 on Build GRCh38
rs762324227 Chr.1: 39562391-39562392 on Build GRCh38
rs540067226 Chr.1: 39562314-39562314 on Build GRCh38
rs778202043 Chr.1: 39562026-39562026 on Build GRCh38
rs780498130 Chr.1: 39562348-39562348 on Build GRCh38
rs907615244 Chr.1: 39562214-39562214 on Build GRCh38
rs866020103 Chr.1: 39562180-39562180 on Build GRCh38
rs769601633 Chr.1: 39562213-39562213 on Build GRCh38
rs769689512 Chr.1: 39562070-39562070 on Build GRCh38
rs769044699 Chr.1: 39562343-39562344 on Build GRCh38
rs749700693 Chr.1: 39562031-39562031 on Build GRCh38
rs757825667 Chr.1: 39562033-39562033 on Build GRCh38
rs772325247 Chr.1: 39562046-39562046 on Build GRCh38
rs1043415923 Chr.1: 39562301-39562301 on Build GRCh38
rs751104610 Chr.1: 39562081-39562081 on Build GRCh38
rs1003926190 Chr.1: 39562464-39562464 on Build GRCh38
rs768922601 Chr.1: 39562187-39562187 on Build GRCh38
rs868182270 Chr.1: 39562181-39562181 on Build GRCh38
rs767132357 Chr.1: 39562090-39562090 on Build GRCh38
rs777058003 Chr.1: 39562344-39562344 on Build GRCh38
rs920597997 Chr.1: 39562175-39562175 on Build GRCh38
rs141368472 Chr.1: 39562174-39562174 on Build GRCh38
rs367597534 Chr.1: 39562302-39562302 on Build GRCh38
rs201212605 Chr.1: 39562218-39562218 on Build GRCh38
rs761582882 Chr.1: 39562067-39562067 on Build GRCh38
rs200762043 Chr.1: 39562091-39562091 on Build GRCh38
rs774195474 Chr.1: 39562415-39562415 on Build GRCh38

View Full Product Details