Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3496643-3497183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
UniGene
Hs.594640
Gene Chromosome Location
Chr.18: 3496032-4455307 on Build GRCh38
Transcripts
NM_001003809.2 NM_001242762.1 NM_001242763.1 NM_001242764.1 NM_001242766.1 NM_001308390.1 NM_004746.3 XM_005258171.2 XM_005258172.1 XM_005258173.3 XM_005258174.4 XM_006722367.3 XM_017026080.1 XM_017026081.1 XM_017026083.1

SNP Detail

SNP ID Chromosome Location
rs771823869 Chr.18: 3496981-3496981 on Build GRCh38
rs562436511 Chr.18: 3496947-3496947 on Build GRCh38
rs531483277 Chr.18: 3497105-3497105 on Build GRCh38
rs182393835 Chr.18: 3496966-3496966 on Build GRCh38
rs947301281 Chr.18: 3497104-3497104 on Build GRCh38
rs777194556 Chr.18: 3496767-3496767 on Build GRCh38
rs967223190 Chr.18: 3496671-3496671 on Build GRCh38
rs750707656 Chr.18: 3496803-3496806 on Build GRCh38
rs893090053 Chr.18: 3496746-3496746 on Build GRCh38
rs934416310 Chr.18: 3497063-3497063 on Build GRCh38
rs557257576 Chr.18: 3496772-3496772 on Build GRCh38
rs910151413 Chr.18: 3496925-3496925 on Build GRCh38
rs998658636 Chr.18: 3497094-3497094 on Build GRCh38
rs941571061 Chr.18: 3496936-3496936 on Build GRCh38
rs960811898 Chr.18: 3496896-3496896 on Build GRCh38
rs912335020 Chr.18: 3497040-3497040 on Build GRCh38
rs546379614 Chr.18: 3496760-3496761 on Build GRCh38
rs971561082 Chr.18: 3496857-3496857 on Build GRCh38
rs1051254178 Chr.18: 3496671-3496672 on Build GRCh38
rs1035551024 Chr.18: 3496893-3496893 on Build GRCh38
rs571356653 Chr.18: 3496735-3496735 on Build GRCh38
rs574192216 Chr.18: 3496839-3496839 on Build GRCh38
rs564982765 Chr.18: 3497145-3497145 on Build GRCh38
rs11447147 Chr.18: 3496742-3496743 on Build GRCh38
rs748824745 Chr.18: 3496903-3496903 on Build GRCh38
rs760463301 Chr.18: 3497141-3497141 on Build GRCh38
rs991131100 Chr.18: 3496780-3496780 on Build GRCh38
rs775372173 Chr.18: 3497135-3497135 on Build GRCh38
rs915536852 Chr.18: 3496804-3496804 on Build GRCh38
rs144373294 Chr.18: 3497127-3497127 on Build GRCh38
rs537189890 Chr.18: 3496762-3496762 on Build GRCh38
rs375496270 Chr.18: 3496918-3496918 on Build GRCh38
rs773917965 Chr.18: 3496944-3496944 on Build GRCh38
rs905656692 Chr.18: 3496837-3496837 on Build GRCh38
rs572859171 Chr.18: 3497075-3497075 on Build GRCh38
rs1052796337 Chr.18: 3497068-3497068 on Build GRCh38
rs919000635 Chr.18: 3496917-3496917 on Build GRCh38
rs962949874 Chr.18: 3496932-3496932 on Build GRCh38
rs868743482 Chr.18: 3496852-3496852 on Build GRCh38
rs770519071 Chr.18: 3496943-3496943 on Build GRCh38
rs986985593 Chr.18: 3497007-3497007 on Build GRCh38
rs542922703 Chr.18: 3496919-3496919 on Build GRCh38
rs554309992 Chr.18: 3496672-3496672 on Build GRCh38

View Full Product Details