Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 22936612-22937015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP8
Entrez Gene ID
5932
Gene Name
RB binding protein 8, endonuclease
Gene Aliases
COM1, CTIP, JWDS, RIM, SAE2, SCKL2
UniGene
Hs.546282
Gene Chromosome Location
Chr.18: 22914121-23026486 on Build GRCh38
Transcripts
NM_002894.2 NM_203291.1 NM_203292.1 XM_005258325.2 XM_005258326.3 XM_006722519.2 XM_006722520.2 XM_006722521.2 XM_011526132.2 XM_017025916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987003 c.85G>T p.E29* Chr.18: 22936936-22936936 on Build GRCh38
6763561 c.67C>A p.L23I Chr.18: 22936918-22936918 on Build GRCh38
5436404 c.29G>T p.S10I Chr.18: 22936880-22936880 on Build GRCh38
6763559 c.21C>A p.S7R Chr.18: 22936872-22936872 on Build GRCh38
7386984 c.91C>A p.H31N Chr.18: 22936942-22936942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765551179 Chr.18: 22936905-22936905 on Build GRCh38
rs771928161 Chr.18: 22936719-22936719 on Build GRCh38
rs766050259 Chr.18: 22936897-22936897 on Build GRCh38
rs181038099 Chr.18: 22936795-22936795 on Build GRCh38
rs758961826 Chr.18: 22936916-22936916 on Build GRCh38
rs145372887 Chr.18: 22936827-22936827 on Build GRCh38
rs539022022 Chr.18: 22936906-22936906 on Build GRCh38
rs754888476 Chr.18: 22936962-22936962 on Build GRCh38
rs997377545 Chr.18: 22936775-22936775 on Build GRCh38
rs759940206 Chr.18: 22936902-22936902 on Build GRCh38
rs772050923 Chr.18: 22936981-22936981 on Build GRCh38
rs746058802 Chr.18: 22936960-22936960 on Build GRCh38
rs764445306 Chr.18: 22936917-22936917 on Build GRCh38
rs374405394 Chr.18: 22936836-22936836 on Build GRCh38
rs757562015 Chr.18: 22936817-22936817 on Build GRCh38
rs141481899 Chr.18: 22936863-22936863 on Build GRCh38
rs746543292 Chr.18: 22936986-22936986 on Build GRCh38
rs779683662 Chr.18: 22936945-22936946 on Build GRCh38
rs757365480 Chr.18: 22936944-22936944 on Build GRCh38
rs772981216 Chr.18: 22936985-22936985 on Build GRCh38
rs901258373 Chr.18: 22936726-22936726 on Build GRCh38
rs747007818 Chr.18: 22936805-22936805 on Build GRCh38
rs772792358 Chr.18: 22936865-22936865 on Build GRCh38
rs751695183 Chr.18: 22936925-22936925 on Build GRCh38
rs781527990 Chr.18: 22936824-22936824 on Build GRCh38
rs963420782 Chr.18: 22936696-22936696 on Build GRCh38
rs372319361 Chr.18: 22936988-22936988 on Build GRCh38
rs770402777 Chr.18: 22936987-22936987 on Build GRCh38
rs149189755 Chr.18: 22936966-22936966 on Build GRCh38
rs200568560 Chr.18: 22936862-22936862 on Build GRCh38
rs993459625 Chr.18: 22936776-22936776 on Build GRCh38
rs781615993 Chr.18: 22936950-22936950 on Build GRCh38
rs1051416455 Chr.18: 22936904-22936904 on Build GRCh38
rs569980277 Chr.18: 22936899-22936899 on Build GRCh38
rs760036005 Chr.18: 22936896-22936896 on Build GRCh38
rs148708986 Chr.18: 22936851-22936851 on Build GRCh38
rs185646125 Chr.18: 22936797-22936797 on Build GRCh38
rs886053659 Chr.18: 22936769-22936769 on Build GRCh38
rs1015459982 Chr.18: 22936674-22936674 on Build GRCh38
rs368319773 Chr.18: 22936978-22936978 on Build GRCh38
rs747839477 Chr.18: 22936979-22936979 on Build GRCh38
rs958964768 Chr.18: 22936728-22936728 on Build GRCh38
rs887025370 Chr.18: 22936921-22936921 on Build GRCh38
rs527936377 Chr.18: 22936707-22936707 on Build GRCh38

View Full Product Details