Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45734212-45734740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
UniGene
Hs.101307
Gene Chromosome Location
Chr.18: 45724123-45752520 on Build GRCh38
Transcripts
NM_001128588.3 NM_001146036.2 NM_001146037.1 NM_001308278.1 NM_001308279.1 NM_015865.6 XM_005258329.1 XM_005258333.1 XM_006722526.3 XM_011526142.1 XM_011526144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821469 c.372C>A p.Y124* Chr.18: 45734304-45734304 on Build GRCh38
3821471 c.379A>C p.T127P Chr.18: 45734311-45734311 on Build GRCh38
6779668 c.553G>A p.V185M Chr.18: 45734317-45734317 on Build GRCh38
6394257 c.466A>G p.T156A Chr.18: 45734398-45734398 on Build GRCh38
6006177 c.551T>A p.L184Q Chr.18: 45734315-45734315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754797005 Chr.18: 45734275-45734275 on Build GRCh38
rs764949996 Chr.18: 45734353-45734353 on Build GRCh38
rs755014196 Chr.18: 45734342-45734343 on Build GRCh38
rs965751942 Chr.18: 45734470-45734470 on Build GRCh38
rs747951920 Chr.18: 45734291-45734291 on Build GRCh38
rs779085504 Chr.18: 45734345-45734346 on Build GRCh38
rs869292669 Chr.18: 45734437-45734438 on Build GRCh38
rs761143210 Chr.18: 45734449-45734449 on Build GRCh38
rs778632720 Chr.18: 45734284-45734284 on Build GRCh38
rs527786363 Chr.18: 45734457-45734457 on Build GRCh38
rs1017370612 Chr.18: 45734532-45734532 on Build GRCh38
rs1047632295 Chr.18: 45734507-45734507 on Build GRCh38
rs775752379 Chr.18: 45734308-45734308 on Build GRCh38
rs762349766 Chr.18: 45734252-45734252 on Build GRCh38
rs1005535831 Chr.18: 45734525-45734525 on Build GRCh38
rs377318707 Chr.18: 45734251-45734251 on Build GRCh38
rs761344869 Chr.18: 45734606-45734606 on Build GRCh38
rs775601456 Chr.18: 45734235-45734235 on Build GRCh38
rs758576124 Chr.18: 45734350-45734351 on Build GRCh38
rs773851667 Chr.18: 45734448-45734448 on Build GRCh38
rs762296350 Chr.18: 45734335-45734335 on Build GRCh38
rs1000107337 Chr.18: 45734368-45734368 on Build GRCh38
rs145134908 Chr.18: 45734343-45734343 on Build GRCh38
rs749005267 Chr.18: 45734548-45734548 on Build GRCh38
rs749908890 Chr.18: 45734260-45734260 on Build GRCh38
rs917362367 Chr.18: 45734662-45734662 on Build GRCh38
rs372865059 Chr.18: 45734447-45734447 on Build GRCh38
rs199975559 Chr.18: 45734255-45734255 on Build GRCh38
rs8096571 Chr.18: 45734508-45734508 on Build GRCh38
rs957875188 Chr.18: 45734674-45734674 on Build GRCh38
rs114641857 Chr.18: 45734414-45734414 on Build GRCh38
rs571444357 Chr.18: 45734666-45734666 on Build GRCh38
rs185283898 Chr.18: 45734269-45734269 on Build GRCh38
rs909892082 Chr.18: 45734601-45734601 on Build GRCh38
rs776564124 Chr.18: 45734575-45734578 on Build GRCh38
rs76940699 Chr.18: 45734436-45734436 on Build GRCh38
rs989802788 Chr.18: 45734684-45734684 on Build GRCh38
rs542004238 Chr.18: 45734603-45734603 on Build GRCh38
rs758213513 Chr.18: 45734373-45734373 on Build GRCh38
rs768933061 Chr.18: 45734331-45734331 on Build GRCh38
rs748500509 Chr.18: 45734429-45734429 on Build GRCh38
rs1054552945 Chr.18: 45734312-45734312 on Build GRCh38
rs745779013 Chr.18: 45734258-45734258 on Build GRCh38
rs75587592 Chr.18: 45734697-45734697 on Build GRCh38
rs756218171 Chr.18: 45734380-45734380 on Build GRCh38
rs563016158 Chr.18: 45734374-45734374 on Build GRCh38
rs765869708 Chr.18: 45734346-45734346 on Build GRCh38
rs777869206 Chr.18: 45734427-45734431 on Build GRCh38
rs549036324 Chr.18: 45734506-45734506 on Build GRCh38
rs16978473 Chr.18: 45734334-45734334 on Build GRCh38
rs758264934 Chr.18: 45734293-45734293 on Build GRCh38
rs58556761 Chr.18: 45734701-45734701 on Build GRCh38
rs958743199 Chr.18: 45734568-45734568 on Build GRCh38
rs779936392 Chr.18: 45734389-45734389 on Build GRCh38
rs777719635 Chr.18: 45734297-45734297 on Build GRCh38
rs188715513 Chr.18: 45734703-45734703 on Build GRCh38
rs202136116 Chr.18: 45734250-45734250 on Build GRCh38
rs978021676 Chr.18: 45734634-45734634 on Build GRCh38
rs374022751 Chr.18: 45734282-45734282 on Build GRCh38
rs773735683 Chr.18: 45734602-45734602 on Build GRCh38
rs1010416191 Chr.18: 45734400-45734400 on Build GRCh38
rs770310542 Chr.18: 45734439-45734440 on Build GRCh38
rs753514150 Chr.18: 45734350-45734350 on Build GRCh38
rs529739834 Chr.18: 45734650-45734650 on Build GRCh38
rs35812340 Chr.18: 45734436-45734437 on Build GRCh38
rs74597469 Chr.18: 45734435-45734435 on Build GRCh38
rs772654960 Chr.18: 45734337-45734337 on Build GRCh38
rs1044232885 Chr.18: 45734306-45734306 on Build GRCh38
rs538666106 Chr.18: 45734673-45734673 on Build GRCh38
rs149080548 Chr.18: 45734591-45734591 on Build GRCh38
rs397736857 Chr.18: 45734446-45734447 on Build GRCh38
rs1028439657 Chr.18: 45734542-45734542 on Build GRCh38
rs8095657 Chr.18: 45734580-45734580 on Build GRCh38
rs111352426 Chr.18: 45734268-45734268 on Build GRCh38
rs993331334 Chr.18: 45734415-45734415 on Build GRCh38
rs78937798 Chr.18: 45734273-45734273 on Build GRCh38
rs759054346 Chr.18: 45734351-45734351 on Build GRCh38
rs749387534 Chr.18: 45734395-45734395 on Build GRCh38
rs768718697 Chr.18: 45734403-45734403 on Build GRCh38
rs752749272 Chr.18: 45734347-45734348 on Build GRCh38
rs749543373 Chr.18: 45734323-45734323 on Build GRCh38
rs910227063 Chr.18: 45734694-45734694 on Build GRCh38
rs867798276 Chr.18: 45734287-45734287 on Build GRCh38
rs561973585 Chr.18: 45734613-45734613 on Build GRCh38
rs755707191 Chr.18: 45734262-45734262 on Build GRCh38
rs769817577 Chr.18: 45734307-45734307 on Build GRCh38
rs764408726 Chr.18: 45734248-45734248 on Build GRCh38
rs80044581 Chr.18: 45734371-45734371 on Build GRCh38
rs762065158 Chr.18: 45734702-45734702 on Build GRCh38
rs745902392 Chr.18: 45734304-45734304 on Build GRCh38
rs747309937 Chr.18: 45734437-45734437 on Build GRCh38
rs1054351394 Chr.18: 45734708-45734708 on Build GRCh38

View Full Product Details