Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74510721-74511229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNDP2
Entrez Gene ID
55748
Gene Name
CNDP dipeptidase 2 (metallopeptidase M20 family)
Gene Aliases
CN2, CPGL, HEL-S-13, HsT2298, PEPA
UniGene
Hs.149185
Gene Chromosome Location
Chr.18: 74496265-74523454 on Build GRCh38
Transcripts
NM_001168499.1 NM_018235.2 XM_005266728.2 XM_006722503.2 XM_011526071.2 XM_011526072.2 XM_011526073.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153880 c.583G>A p.D195N Chr.18: 74510939-74510939 on Build GRCh38
1241025 c.495G>T p.M165I Chr.18: 74510851-74510851 on Build GRCh38
3403647 c.536C>T p.A179V Chr.18: 74510892-74510892 on Build GRCh38
438377 c.655G>C p.E219Q Chr.18: 74511011-74511011 on Build GRCh38
4396241 c.552C>T p.F184F Chr.18: 74510908-74510908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770897034 Chr.18: 74510914-74510914 on Build GRCh38
rs780248302 Chr.18: 74510870-74510870 on Build GRCh38
rs750789500 Chr.18: 74510929-74510929 on Build GRCh38
rs575660878 Chr.18: 74511008-74511008 on Build GRCh38
rs754648445 Chr.18: 74510772-74510772 on Build GRCh38
rs202208596 Chr.18: 74510923-74510923 on Build GRCh38
rs769192423 Chr.18: 74511020-74511021 on Build GRCh38
rs781750509 Chr.18: 74511038-74511038 on Build GRCh38
rs753960343 Chr.18: 74510943-74510943 on Build GRCh38
rs145677677 Chr.18: 74510773-74510773 on Build GRCh38
rs767049173 Chr.18: 74510938-74510938 on Build GRCh38
rs758625244 Chr.18: 74511063-74511063 on Build GRCh38
rs748322327 Chr.18: 74510881-74510881 on Build GRCh38
rs371054265 Chr.18: 74510780-74510780 on Build GRCh38
rs777916953 Chr.18: 74511064-74511064 on Build GRCh38
rs773134724 Chr.18: 74510820-74510820 on Build GRCh38
rs897470051 Chr.18: 74510905-74510905 on Build GRCh38
rs758580568 Chr.18: 74510970-74510970 on Build GRCh38
rs868206104 Chr.18: 74510918-74510918 on Build GRCh38
rs548298258 Chr.18: 74511176-74511176 on Build GRCh38
rs995482530 Chr.18: 74510980-74510980 on Build GRCh38
rs767497569 Chr.18: 74510759-74510759 on Build GRCh38
rs8088885 Chr.18: 74511175-74511175 on Build GRCh38
rs912794687 Chr.18: 74510882-74510882 on Build GRCh38
rs767782122 Chr.18: 74510854-74510854 on Build GRCh38
rs755040750 Chr.18: 74510956-74510956 on Build GRCh38
rs928783235 Chr.18: 74511198-74511198 on Build GRCh38
rs559020663 Chr.18: 74510745-74510745 on Build GRCh38
rs566122589 Chr.18: 74511169-74511169 on Build GRCh38
rs370820197 Chr.18: 74511021-74511021 on Build GRCh38
rs139467921 Chr.18: 74510971-74510971 on Build GRCh38
rs2278158 Chr.18: 74511065-74511065 on Build GRCh38
rs948590491 Chr.18: 74511192-74511192 on Build GRCh38
rs200391267 Chr.18: 74510966-74510966 on Build GRCh38
rs1050159161 Chr.18: 74511040-74511065 on Build GRCh38
rs775646494 Chr.18: 74511077-74511077 on Build GRCh38
rs532245121 Chr.18: 74510791-74510791 on Build GRCh38
rs748905446 Chr.18: 74510832-74510832 on Build GRCh38
rs751859927 Chr.18: 74510842-74510842 on Build GRCh38
rs148388345 Chr.18: 74510819-74510819 on Build GRCh38
rs773991540 Chr.18: 74510817-74510817 on Build GRCh38
rs762632101 Chr.18: 74511028-74511028 on Build GRCh38
rs2278157 Chr.18: 74511133-74511133 on Build GRCh38
rs1012845107 Chr.18: 74510749-74510749 on Build GRCh38
rs34916325 Chr.18: 74510895-74510895 on Build GRCh38
rs868094700 Chr.18: 74511029-74511029 on Build GRCh38
rs771336985 Chr.18: 74510965-74510965 on Build GRCh38
rs747958571 Chr.18: 74510981-74510981 on Build GRCh38
rs147519744 Chr.18: 74510863-74510863 on Build GRCh38
rs547973029 Chr.18: 74510801-74510801 on Build GRCh38
rs35740490 Chr.18: 74510827-74510827 on Build GRCh38
rs958841216 Chr.18: 74510783-74510783 on Build GRCh38
rs373372409 Chr.18: 74510877-74510877 on Build GRCh38
rs907469842 Chr.18: 74510904-74510904 on Build GRCh38
rs971328283 Chr.18: 74510983-74510983 on Build GRCh38
rs139859713 Chr.18: 74511054-74511054 on Build GRCh38
rs781176395 Chr.18: 74510992-74510992 on Build GRCh38
rs199708812 Chr.18: 74510921-74510921 on Build GRCh38
rs765878565 Chr.18: 74510763-74510763 on Build GRCh38
rs563095367 Chr.18: 74511033-74511033 on Build GRCh38
rs138039302 Chr.18: 74510873-74510873 on Build GRCh38
rs373268995 Chr.18: 74511053-74511053 on Build GRCh38
rs768238613 Chr.18: 74511030-74511030 on Build GRCh38
rs561643002 Chr.18: 74511180-74511180 on Build GRCh38
rs746857976 Chr.18: 74510974-74510974 on Build GRCh38
rs765827173 Chr.18: 74510828-74510828 on Build GRCh38
rs181763245 Chr.18: 74510793-74510793 on Build GRCh38
rs780299501 Chr.18: 74510796-74510796 on Build GRCh38
rs759982823 Chr.18: 74511042-74511042 on Build GRCh38
rs376953227 Chr.18: 74510915-74510915 on Build GRCh38
rs756663501 Chr.18: 74510933-74510933 on Build GRCh38
rs946203220 Chr.18: 74510892-74510892 on Build GRCh38
rs368762481 Chr.18: 74510797-74510797 on Build GRCh38
rs2278159 Chr.18: 74510926-74510926 on Build GRCh38
rs543234302 Chr.18: 74511032-74511032 on Build GRCh38
rs756716500 Chr.18: 74510867-74510867 on Build GRCh38
rs989110828 Chr.18: 74511168-74511168 on Build GRCh38
rs945083718 Chr.18: 74511178-74511178 on Build GRCh38
rs763275334 Chr.18: 74510816-74510817 on Build GRCh38
rs764921857 Chr.18: 74510835-74510835 on Build GRCh38
rs745814103 Chr.18: 74511000-74511000 on Build GRCh38
rs940302360 Chr.18: 74511100-74511100 on Build GRCh38
rs374633647 Chr.18: 74510925-74510925 on Build GRCh38
rs751997697 Chr.18: 74510789-74510789 on Build GRCh38
rs770183692 Chr.18: 74511076-74511076 on Build GRCh38
rs201098707 Chr.18: 74510924-74510924 on Build GRCh38
rs373487786 Chr.18: 74510792-74510792 on Build GRCh38
rs753431269 Chr.18: 74510765-74510765 on Build GRCh38
rs770844139 Chr.18: 74510984-74510984 on Build GRCh38
rs79359709 Chr.18: 74510754-74510754 on Build GRCh38
rs754338607 Chr.18: 74511041-74511041 on Build GRCh38
rs771842560 Chr.18: 74510894-74510894 on Build GRCh38
rs757710471 Chr.18: 74510847-74510847 on Build GRCh38
rs774889715 Chr.18: 74511022-74511022 on Build GRCh38
rs920976830 Chr.18: 74511125-74511125 on Build GRCh38
rs151125709 Chr.18: 74510878-74510878 on Build GRCh38
rs776152098 Chr.18: 74510831-74510831 on Build GRCh38

View Full Product Details