Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 75410198-75410706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM21
Entrez Gene ID
284274
Gene Name
small integral membrane protein 21
Gene Aliases
C18orf62
UniGene
Hs.666744
Gene Chromosome Location
Chr.18: 75409476-75455653 on Build GRCh38
Transcripts
NM_001037331.3

SNP Detail

SNP ID Chromosome Location
rs546718522 Chr.18: 75410642-75410642 on Build GRCh38
rs754499471 Chr.18: 75410577-75410577 on Build GRCh38
rs576845687 Chr.18: 75410342-75410342 on Build GRCh38
rs1004400217 Chr.18: 75410509-75410509 on Build GRCh38
rs921201853 Chr.18: 75410235-75410235 on Build GRCh38
rs368737528 Chr.18: 75410242-75410242 on Build GRCh38
rs376694963 Chr.18: 75410227-75410227 on Build GRCh38
rs1006515437 Chr.18: 75410535-75410535 on Build GRCh38
rs528797147 Chr.18: 75410430-75410430 on Build GRCh38
rs989309049 Chr.18: 75410634-75410634 on Build GRCh38
rs943103432 Chr.18: 75410397-75410397 on Build GRCh38
rs188359794 Chr.18: 75410277-75410277 on Build GRCh38
rs1021638083 Chr.18: 75410624-75410624 on Build GRCh38
rs888186950 Chr.18: 75410484-75410484 on Build GRCh38
rs910424378 Chr.18: 75410369-75410369 on Build GRCh38
rs1006770580 Chr.18: 75410241-75410241 on Build GRCh38
rs545512376 Chr.18: 75410384-75410384 on Build GRCh38
rs970546709 Chr.18: 75410269-75410269 on Build GRCh38
rs988829596 Chr.18: 75410303-75410303 on Build GRCh38
rs906513767 Chr.18: 75410310-75410310 on Build GRCh38
rs906062061 Chr.18: 75410584-75410584 on Build GRCh38
rs993098292 Chr.18: 75410420-75410420 on Build GRCh38
rs887993048 Chr.18: 75410483-75410483 on Build GRCh38
rs533443587 Chr.18: 75410598-75410598 on Build GRCh38
rs554851759 Chr.18: 75410266-75410266 on Build GRCh38
rs556798105 Chr.18: 75410327-75410327 on Build GRCh38
rs531018694 Chr.18: 75410591-75410591 on Build GRCh38
rs766399431 Chr.18: 75410414-75410414 on Build GRCh38
rs561996186 Chr.18: 75410567-75410567 on Build GRCh38
rs913791642 Chr.18: 75410324-75410324 on Build GRCh38
rs192244121 Chr.18: 75410592-75410592 on Build GRCh38
rs965020236 Chr.18: 75410539-75410539 on Build GRCh38
rs1043910145 Chr.18: 75410358-75410358 on Build GRCh38
rs988777150 Chr.18: 75410610-75410610 on Build GRCh38
rs1023548048 Chr.18: 75410551-75410551 on Build GRCh38
rs112618211 Chr.18: 75410596-75410596 on Build GRCh38
rs1047396002 Chr.18: 75410452-75410452 on Build GRCh38
rs559288700 Chr.18: 75410416-75410416 on Build GRCh38
rs1040151720 Chr.18: 75410399-75410399 on Build GRCh38
rs1055263611 Chr.18: 75410347-75410347 on Build GRCh38
rs995006170 Chr.18: 75410404-75410404 on Build GRCh38
rs543563526 Chr.18: 75410302-75410302 on Build GRCh38
rs867999364 Chr.18: 75410280-75410280 on Build GRCh38
rs912388241 Chr.18: 75410491-75410491 on Build GRCh38
rs954133411 Chr.18: 75410632-75410632 on Build GRCh38
rs1054037425 Chr.18: 75410248-75410248 on Build GRCh38
rs542110820 Chr.18: 75410521-75410521 on Build GRCh38
rs981584681 Chr.18: 75410272-75410272 on Build GRCh38
rs1023137824 Chr.18: 75410244-75410244 on Build GRCh38
rs1015366961 Chr.18: 75410513-75410513 on Build GRCh38
rs11660753 Chr.18: 75410643-75410643 on Build GRCh38
rs570878366 Chr.18: 75410595-75410595 on Build GRCh38
rs956121529 Chr.18: 75410604-75410604 on Build GRCh38

View Full Product Details