Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2944749-2945254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF77
Entrez Gene ID
58492
Gene Name
zinc finger protein 77
Gene Aliases
pT1
UniGene
Hs.38004
Gene Chromosome Location
Chr.19: 2933218-2944971 on Build GRCh38
Transcripts
NM_021217.2 XM_017027081.1 XM_017027082.1

SNP Detail

SNP ID Chromosome Location
rs570442676 Chr.19: 2945171-2945171 on Build GRCh38
rs915914390 Chr.19: 2944963-2944963 on Build GRCh38
rs906213958 Chr.19: 2945135-2945135 on Build GRCh38
rs796672622 Chr.19: 2944828-2944828 on Build GRCh38
rs558306479 Chr.19: 2945159-2945159 on Build GRCh38
rs571405967 Chr.19: 2944770-2944770 on Build GRCh38
rs1026938970 Chr.19: 2944919-2944919 on Build GRCh38
rs375553228 Chr.19: 2945021-2945021 on Build GRCh38
rs578220032 Chr.19: 2944969-2944969 on Build GRCh38
rs540315178 Chr.19: 2945069-2945069 on Build GRCh38
rs767139099 Chr.19: 2945173-2945173 on Build GRCh38
rs145708822 Chr.19: 2945001-2945001 on Build GRCh38
rs890838612 Chr.19: 2945005-2945005 on Build GRCh38
rs902453061 Chr.19: 2944794-2944794 on Build GRCh38
rs535968955 Chr.19: 2944908-2944908 on Build GRCh38
rs951181993 Chr.19: 2944929-2944929 on Build GRCh38
rs372280798 Chr.19: 2944820-2944820 on Build GRCh38
rs528491028 Chr.19: 2945065-2945065 on Build GRCh38
rs566476995 Chr.19: 2944944-2944944 on Build GRCh38
rs977887826 Chr.19: 2945226-2945226 on Build GRCh38
rs953650551 Chr.19: 2945197-2945197 on Build GRCh38
rs968791233 Chr.19: 2945187-2945187 on Build GRCh38
rs1048105946 Chr.19: 2945118-2945118 on Build GRCh38
rs921887856 Chr.19: 2944911-2944911 on Build GRCh38
rs4375782 Chr.19: 2945101-2945101 on Build GRCh38
rs991586931 Chr.19: 2945082-2945082 on Build GRCh38
rs938793977 Chr.19: 2944946-2944946 on Build GRCh38
rs182393623 Chr.19: 2945080-2945080 on Build GRCh38
rs911977378 Chr.19: 2945076-2945076 on Build GRCh38
rs1032459904 Chr.19: 2944802-2944802 on Build GRCh38
rs532305829 Chr.19: 2945088-2945088 on Build GRCh38
rs560605656 Chr.19: 2945224-2945224 on Build GRCh38
rs1004997955 Chr.19: 2945033-2945033 on Build GRCh38
rs56243325 Chr.19: 2945203-2945203 on Build GRCh38
rs1024699263 Chr.19: 2944868-2944868 on Build GRCh38
rs974039596 Chr.19: 2945047-2945047 on Build GRCh38
rs148940274 Chr.19: 2945002-2945002 on Build GRCh38
rs763416445 Chr.19: 2945003-2945003 on Build GRCh38
rs111380042 Chr.19: 2944955-2944955 on Build GRCh38
rs550438123 Chr.19: 2945090-2945090 on Build GRCh38
rs978703007 Chr.19: 2945195-2945195 on Build GRCh38
rs1000534960 Chr.19: 2944800-2944800 on Build GRCh38
rs566396840 Chr.19: 2945125-2945125 on Build GRCh38
rs924840391 Chr.19: 2945095-2945095 on Build GRCh38
rs1029259388 Chr.19: 2945056-2945056 on Build GRCh38
rs1011913963 Chr.19: 2944848-2944848 on Build GRCh38
rs1004549779 Chr.19: 2945152-2945152 on Build GRCh38
rs1036055857 Chr.19: 2945163-2945163 on Build GRCh38
rs1029642322 Chr.19: 2945181-2945181 on Build GRCh38
rs899609755 Chr.19: 2945117-2945117 on Build GRCh38
rs781184731 Chr.19: 2944899-2944899 on Build GRCh38
rs916028876 Chr.19: 2945090-2945090 on Build GRCh38
rs4338902 Chr.19: 2945104-2945104 on Build GRCh38
rs995043878 Chr.19: 2945178-2945178 on Build GRCh38
rs906330020 Chr.19: 2945014-2945014 on Build GRCh38
rs140057887 Chr.19: 2944808-2944808 on Build GRCh38
rs143699675 Chr.19: 2944984-2944984 on Build GRCh38
rs568696283 Chr.19: 2944902-2944902 on Build GRCh38
rs570803495 Chr.19: 2945118-2945119 on Build GRCh38
rs943569621 Chr.19: 2945109-2945109 on Build GRCh38
rs1026887030 Chr.19: 2945060-2945060 on Build GRCh38
rs924641009 Chr.19: 2945196-2945196 on Build GRCh38
rs983606161 Chr.19: 2944888-2944888 on Build GRCh38
rs751936795 Chr.19: 2944872-2944872 on Build GRCh38
rs1022986033 Chr.19: 2945183-2945183 on Build GRCh38
rs550401145 Chr.19: 2944882-2944882 on Build GRCh38
rs912063644 Chr.19: 2945105-2945105 on Build GRCh38
rs912162291 Chr.19: 2945218-2945218 on Build GRCh38
rs987735332 Chr.19: 2945214-2945214 on Build GRCh38
rs764532964 Chr.19: 2944897-2944897 on Build GRCh38
rs561185888 Chr.19: 2945036-2945036 on Build GRCh38
rs936538431 Chr.19: 2945092-2945092 on Build GRCh38
rs755361513 Chr.19: 2944878-2944878 on Build GRCh38
rs563902767 Chr.19: 2945231-2945231 on Build GRCh38
rs768740446 Chr.19: 2944887-2944887 on Build GRCh38
rs1031153725 Chr.19: 2944894-2944894 on Build GRCh38
rs987494369 Chr.19: 2945074-2945074 on Build GRCh38
rs781621350 Chr.19: 2944879-2944879 on Build GRCh38
rs75614126 Chr.19: 2945093-2945093 on Build GRCh38
rs912010048 Chr.19: 2944923-2944923 on Build GRCh38
rs553629271 Chr.19: 2944965-2944965 on Build GRCh38
rs1043167498 Chr.19: 2944866-2944866 on Build GRCh38
rs982994595 Chr.19: 2945071-2945071 on Build GRCh38
rs1039461090 Chr.19: 2945114-2945114 on Build GRCh38
rs899494754 Chr.19: 2945038-2945038 on Build GRCh38
rs1015469933 Chr.19: 2945034-2945034 on Build GRCh38
rs890899953 Chr.19: 2944901-2944901 on Build GRCh38
rs936553869 Chr.19: 2944964-2944964 on Build GRCh38
rs932018986 Chr.19: 2944913-2944913 on Build GRCh38
rs79257218 Chr.19: 2944940-2944940 on Build GRCh38
rs1036131457 Chr.19: 2945153-2945153 on Build GRCh38
rs1018923966 Chr.19: 2945216-2945216 on Build GRCh38
rs766842119 Chr.19: 2944829-2944829 on Build GRCh38
rs1007983304 Chr.19: 2945035-2945035 on Build GRCh38
rs943835188 Chr.19: 2944939-2944939 on Build GRCh38
rs187879602 Chr.19: 2945227-2945227 on Build GRCh38
rs543161268 Chr.19: 2945013-2945013 on Build GRCh38
rs143596298 Chr.19: 2945137-2945137 on Build GRCh38
rs547089897 Chr.19: 2944893-2944893 on Build GRCh38
rs987523928 Chr.19: 2944916-2944916 on Build GRCh38

View Full Product Details