Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3599763-3600254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBXA2R
Entrez Gene ID
6915
Gene Name
thromboxane A2 receptor
Gene Aliases
BDPLT13, TXA2-R
UniGene
Hs.442530
Gene Chromosome Location
Chr.19: 3594506-3608749 on Build GRCh38
Transcripts
NM_001060.5 NM_201636.2 XM_011528214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7131527 c.537G>T p.P179P Chr.19: 3600098-3600098 on Build GRCh38
6150490 c.568G>A p.E190K Chr.19: 3600067-3600067 on Build GRCh38
5575021 c.534C>A p.Y178* Chr.19: 3600101-3600101 on Build GRCh38
6798034 c.557C>T p.T186M Chr.19: 3600078-3600078 on Build GRCh38
4732573 c.625G>A p.G209R Chr.19: 3600010-3600010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993137867 Chr.19: 3599790-3599790 on Build GRCh38
rs767661627 Chr.19: 3600007-3600007 on Build GRCh38
rs754308869 Chr.19: 3599934-3599934 on Build GRCh38
rs748835821 Chr.19: 3600176-3600176 on Build GRCh38
rs34881364 Chr.19: 3600077-3600077 on Build GRCh38
rs780858879 Chr.19: 3599969-3599969 on Build GRCh38
rs202053248 Chr.19: 3599872-3599872 on Build GRCh38
rs201418594 Chr.19: 3600086-3600086 on Build GRCh38
rs200152113 Chr.19: 3600205-3600205 on Build GRCh38
rs372994525 Chr.19: 3599983-3599983 on Build GRCh38
rs774389147 Chr.19: 3600098-3600098 on Build GRCh38
rs201461454 Chr.19: 3599982-3599982 on Build GRCh38
rs5750 Chr.19: 3600108-3600108 on Build GRCh38
rs776787597 Chr.19: 3600211-3600211 on Build GRCh38
rs770941864 Chr.19: 3599929-3599929 on Build GRCh38
rs200638504 Chr.19: 3599853-3599853 on Build GRCh38
rs771248487 Chr.19: 3599813-3599815 on Build GRCh38
rs200936803 Chr.19: 3599891-3599891 on Build GRCh38
rs769254342 Chr.19: 3600067-3600067 on Build GRCh38
rs781614025 Chr.19: 3600017-3600019 on Build GRCh38
rs1037940718 Chr.19: 3600005-3600005 on Build GRCh38
rs5748 Chr.19: 3600200-3600200 on Build GRCh38
rs1025762347 Chr.19: 3599930-3599930 on Build GRCh38
rs762854873 Chr.19: 3599894-3599894 on Build GRCh38
rs200390929 Chr.19: 3599936-3599936 on Build GRCh38
rs200945430 Chr.19: 3600149-3600149 on Build GRCh38
rs765231710 Chr.19: 3600226-3600226 on Build GRCh38
rs781162737 Chr.19: 3600093-3600093 on Build GRCh38
rs764234244 Chr.19: 3600014-3600014 on Build GRCh38
rs952849503 Chr.19: 3599956-3599956 on Build GRCh38
rs760943192 Chr.19: 3600128-3600128 on Build GRCh38
rs985466939 Chr.19: 3600193-3600193 on Build GRCh38
rs376762511 Chr.19: 3600124-3600124 on Build GRCh38
rs766209508 Chr.19: 3599895-3599895 on Build GRCh38
rs373503496 Chr.19: 3599840-3599840 on Build GRCh38
rs748733057 Chr.19: 3599987-3599987 on Build GRCh38
rs780720942 Chr.19: 3600055-3600055 on Build GRCh38
rs533734665 Chr.19: 3599915-3599915 on Build GRCh38
rs201067295 Chr.19: 3599958-3599958 on Build GRCh38
rs768957393 Chr.19: 3600210-3600210 on Build GRCh38
rs775983717 Chr.19: 3599870-3599870 on Build GRCh38
rs750819182 Chr.19: 3600046-3600046 on Build GRCh38
rs201798582 Chr.19: 3599906-3599906 on Build GRCh38
rs768214380 Chr.19: 3599869-3599869 on Build GRCh38
rs773027867 Chr.19: 3599984-3599984 on Build GRCh38
rs777185021 Chr.19: 3600170-3600170 on Build GRCh38
rs752909002 Chr.19: 3600009-3600009 on Build GRCh38
rs200163011 Chr.19: 3600022-3600022 on Build GRCh38
rs746714945 Chr.19: 3599962-3599962 on Build GRCh38
rs779291000 Chr.19: 3600029-3600029 on Build GRCh38
rs541642445 Chr.19: 3600158-3600158 on Build GRCh38
rs759875711 Chr.19: 3600003-3600003 on Build GRCh38
rs199952923 Chr.19: 3599918-3599918 on Build GRCh38
rs1001860165 Chr.19: 3600060-3600060 on Build GRCh38
rs200600231 Chr.19: 3600122-3600122 on Build GRCh38
rs906238607 Chr.19: 3599849-3599849 on Build GRCh38
rs5752 Chr.19: 3599873-3599873 on Build GRCh38
rs779235776 Chr.19: 3599954-3599954 on Build GRCh38
rs200460580 Chr.19: 3599966-3599966 on Build GRCh38
rs749640187 Chr.19: 3599805-3599805 on Build GRCh38
rs747631604 Chr.19: 3599981-3599981 on Build GRCh38
rs200172414 Chr.19: 3600167-3600167 on Build GRCh38
rs771836447 Chr.19: 3600118-3600118 on Build GRCh38
rs545650041 Chr.19: 3600033-3600033 on Build GRCh38
rs771232220 Chr.19: 3599832-3599832 on Build GRCh38
rs976720076 Chr.19: 3599933-3599933 on Build GRCh38
rs201984682 Chr.19: 3599963-3599963 on Build GRCh38
rs964354069 Chr.19: 3600008-3600008 on Build GRCh38
rs774262525 Chr.19: 3600002-3600002 on Build GRCh38
rs5749 Chr.19: 3600157-3600157 on Build GRCh38
rs747768808 Chr.19: 3600056-3600056 on Build GRCh38
rs760677961 Chr.19: 3600013-3600013 on Build GRCh38
rs954017459 Chr.19: 3600178-3600178 on Build GRCh38
rs746660127 Chr.19: 3599857-3599857 on Build GRCh38
rs761892417 Chr.19: 3600221-3600221 on Build GRCh38
rs370280465 Chr.19: 3600203-3600203 on Build GRCh38
rs770630880 Chr.19: 3600017-3600017 on Build GRCh38
rs5751 Chr.19: 3599986-3599986 on Build GRCh38
rs987854613 Chr.19: 3600070-3600070 on Build GRCh38
rs759419835 Chr.19: 3600116-3600116 on Build GRCh38
rs202109452 Chr.19: 3600233-3600233 on Build GRCh38
rs199672500 Chr.19: 3600071-3600071 on Build GRCh38
rs752350415 Chr.19: 3600161-3600161 on Build GRCh38
rs764130321 Chr.19: 3600133-3600133 on Build GRCh38
rs552434490 Chr.19: 3599789-3599789 on Build GRCh38
rs201736798 Chr.19: 3600023-3600023 on Build GRCh38
rs199581802 Chr.19: 3600099-3600099 on Build GRCh38
rs554342940 Chr.19: 3599896-3599896 on Build GRCh38
rs777642650 Chr.19: 3599802-3599802 on Build GRCh38
rs756775879 Chr.19: 3600177-3600177 on Build GRCh38
rs866409065 Chr.19: 3600001-3600001 on Build GRCh38
rs1024614381 Chr.19: 3599812-3599812 on Build GRCh38
rs932661365 Chr.19: 3599973-3599973 on Build GRCh38
rs758711772 Chr.19: 3600051-3600051 on Build GRCh38
rs200718962 Chr.19: 3600109-3600109 on Build GRCh38
rs34604531 Chr.19: 3600206-3600206 on Build GRCh38
rs746206573 Chr.19: 3600179-3600188 on Build GRCh38
rs397514542 Chr.19: 3599913-3599913 on Build GRCh38
rs922331694 Chr.19: 3599820-3599820 on Build GRCh38
rs776662673 Chr.19: 3600136-3600136 on Build GRCh38
rs199995921 Chr.19: 3599831-3599831 on Build GRCh38
rs201310328 Chr.19: 3599922-3599922 on Build GRCh38
rs145699053 Chr.19: 3600006-3600006 on Build GRCh38
rs201295847 Chr.19: 3600197-3600197 on Build GRCh38
rs866763548 Chr.19: 3600004-3600004 on Build GRCh38
rs62130599 Chr.19: 3599852-3599852 on Build GRCh38
rs1047097779 Chr.19: 3600202-3600202 on Build GRCh38
rs202065690 Chr.19: 3599925-3599925 on Build GRCh38
rs1033904590 Chr.19: 3600061-3600061 on Build GRCh38
rs895856931 Chr.19: 3600184-3600184 on Build GRCh38
rs976803993 Chr.19: 3600015-3600015 on Build GRCh38
rs199654283 Chr.19: 3600143-3600143 on Build GRCh38

View Full Product Details