Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11576152-11576652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP5
Entrez Gene ID
54
Gene Name
acid phosphatase 5, tartrate resistant
Gene Aliases
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
UniGene
Hs.1211
Gene Chromosome Location
Chr.19: 11574660-11579008 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Gene Symbol
ZNF627
Entrez Gene ID
199692
Gene Name
zinc finger protein 627
Gene Aliases
-
UniGene
Hs.526665
Gene Chromosome Location
Chr.19: 11575255-11619159 on Build GRCh38
Transcripts
NM_001111034.2 NM_001111035.2 NM_001111036.2 NM_001322023.1 NM_001611.4 XM_005259938.1 XM_011528069.2 XM_011527780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509218 c.413G>A p.R138H Chr.19: 11576565-11576565 on Build GRCh38
5854633 c.619C>T p.P207S Chr.19: 11576359-11576359 on Build GRCh38
3528588 c.620C>T p.P207L Chr.19: 11576358-11576358 on Build GRCh38
4909382 c.555G>A p.L185L Chr.19: 11576423-11576423 on Build GRCh38
3672731 c.412C>T p.R138C Chr.19: 11576566-11576566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746697489 Chr.19: 11576439-11576439 on Build GRCh38
rs748235353 Chr.19: 11576336-11576336 on Build GRCh38
rs779109113 Chr.19: 11576199-11576199 on Build GRCh38
rs756201795 Chr.19: 11576400-11576400 on Build GRCh38
rs778966885 Chr.19: 11576497-11576497 on Build GRCh38
rs368852148 Chr.19: 11576342-11576342 on Build GRCh38
rs751723542 Chr.19: 11576322-11576322 on Build GRCh38
rs761983850 Chr.19: 11576609-11576609 on Build GRCh38
rs367699467 Chr.19: 11576396-11576396 on Build GRCh38
rs887425978 Chr.19: 11576462-11576462 on Build GRCh38
rs752902742 Chr.19: 11576468-11576468 on Build GRCh38
rs764203884 Chr.19: 11576604-11576604 on Build GRCh38
rs371719196 Chr.19: 11576430-11576430 on Build GRCh38
rs766688951 Chr.19: 11576393-11576393 on Build GRCh38
rs387906672 Chr.19: 11576376-11576376 on Build GRCh38
rs765756315 Chr.19: 11576305-11576305 on Build GRCh38
rs978061454 Chr.19: 11576333-11576333 on Build GRCh38
rs764197394 Chr.19: 11576478-11576478 on Build GRCh38
rs775336126 Chr.19: 11576367-11576367 on Build GRCh38
rs371717424 Chr.19: 11576248-11576248 on Build GRCh38
rs779391766 Chr.19: 11576463-11576463 on Build GRCh38
rs756292294 Chr.19: 11576473-11576473 on Build GRCh38
rs755385110 Chr.19: 11576397-11576397 on Build GRCh38
rs757294445 Chr.19: 11576420-11576420 on Build GRCh38
rs758739987 Chr.19: 11576222-11576222 on Build GRCh38
rs568957938 Chr.19: 11576377-11576377 on Build GRCh38
rs144617874 Chr.19: 11576446-11576446 on Build GRCh38
rs547734259 Chr.19: 11576276-11576276 on Build GRCh38
rs760906463 Chr.19: 11576601-11576601 on Build GRCh38
rs765472713 Chr.19: 11576613-11576613 on Build GRCh38
rs142179752 Chr.19: 11576341-11576341 on Build GRCh38
rs2229531 Chr.19: 11576380-11576380 on Build GRCh38
rs915517618 Chr.19: 11576546-11576546 on Build GRCh38
rs750825481 Chr.19: 11576209-11576209 on Build GRCh38
rs387906669 Chr.19: 11576311-11576311 on Build GRCh38
rs774878498 Chr.19: 11576451-11576451 on Build GRCh38
rs2305799 Chr.19: 11576536-11576536 on Build GRCh38
rs753889410 Chr.19: 11576480-11576480 on Build GRCh38
rs373684791 Chr.19: 11576498-11576498 on Build GRCh38
rs925299596 Chr.19: 11576624-11576624 on Build GRCh38
rs774823139 Chr.19: 11576566-11576566 on Build GRCh38
rs199512530 Chr.19: 11576264-11576264 on Build GRCh38
rs754135147 Chr.19: 11576605-11576605 on Build GRCh38
rs747299112 Chr.19: 11576230-11576230 on Build GRCh38
rs764216271 Chr.19: 11576238-11576238 on Build GRCh38
rs552474987 Chr.19: 11576304-11576304 on Build GRCh38
rs978324600 Chr.19: 11576608-11576608 on Build GRCh38
rs750328133 Chr.19: 11576619-11576619 on Build GRCh38
rs376767095 Chr.19: 11576456-11576456 on Build GRCh38
rs770788576 Chr.19: 11576346-11576346 on Build GRCh38
rs201716955 Chr.19: 11576557-11576557 on Build GRCh38
rs752926629 Chr.19: 11576399-11576399 on Build GRCh38
rs575728569 Chr.19: 11576440-11576440 on Build GRCh38
rs773070944 Chr.19: 11576262-11576262 on Build GRCh38
rs776392911 Chr.19: 11576544-11576544 on Build GRCh38
rs374180791 Chr.19: 11576284-11576284 on Build GRCh38
rs767694832 Chr.19: 11576398-11576398 on Build GRCh38
rs771877821 Chr.19: 11576364-11576364 on Build GRCh38
rs535257196 Chr.19: 11576356-11576356 on Build GRCh38
rs749180050 Chr.19: 11576411-11576411 on Build GRCh38
rs143362935 Chr.19: 11576565-11576565 on Build GRCh38
rs1025967277 Chr.19: 11576452-11576452 on Build GRCh38
rs951775339 Chr.19: 11576201-11576202 on Build GRCh38
rs202169550 Chr.19: 11576434-11576434 on Build GRCh38
rs139822120 Chr.19: 11576435-11576435 on Build GRCh38
rs762602282 Chr.19: 11576202-11576202 on Build GRCh38
rs369197756 Chr.19: 11576447-11576447 on Build GRCh38
rs781199182 Chr.19: 11576335-11576335 on Build GRCh38
rs142623076 Chr.19: 11576296-11576296 on Build GRCh38
rs1030101526 Chr.19: 11576181-11576181 on Build GRCh38
rs778805198 Chr.19: 11576433-11576433 on Build GRCh38
rs200679613 Chr.19: 11576628-11576628 on Build GRCh38
rs186246910 Chr.19: 11576627-11576627 on Build GRCh38
rs145419216 Chr.19: 11576517-11576517 on Build GRCh38
rs540751831 Chr.19: 11576188-11576188 on Build GRCh38
rs372107388 Chr.19: 11576357-11576357 on Build GRCh38
rs141555832 Chr.19: 11576485-11576485 on Build GRCh38
rs373030121 Chr.19: 11576285-11576285 on Build GRCh38
rs766846705 Chr.19: 11576449-11576449 on Build GRCh38
rs777726227 Chr.19: 11576404-11576404 on Build GRCh38
rs2229532 Chr.19: 11576317-11576317 on Build GRCh38
rs754840152 Chr.19: 11576631-11576631 on Build GRCh38
rs767853623 Chr.19: 11576585-11576585 on Build GRCh38
rs936765239 Chr.19: 11576370-11576370 on Build GRCh38
rs762775321 Chr.19: 11576263-11576263 on Build GRCh38
rs745457954 Chr.19: 11576436-11576436 on Build GRCh38
rs866992772 Chr.19: 11576458-11576458 on Build GRCh38
rs746606561 Chr.19: 11576519-11576519 on Build GRCh38
rs147105591 Chr.19: 11576501-11576501 on Build GRCh38
rs559438510 Chr.19: 11576197-11576197 on Build GRCh38
rs768709730 Chr.19: 11576231-11576231 on Build GRCh38
rs760044774 Chr.19: 11576568-11576568 on Build GRCh38
rs781252304 Chr.19: 11576235-11576235 on Build GRCh38
rs535962234 Chr.19: 11576389-11576389 on Build GRCh38
rs147864731 Chr.19: 11576457-11576457 on Build GRCh38
rs780454598 Chr.19: 11576320-11576320 on Build GRCh38
rs148656335 Chr.19: 11576326-11576326 on Build GRCh38
rs750930124 Chr.19: 11576381-11576381 on Build GRCh38
rs780220020 Chr.19: 11576224-11576224 on Build GRCh38
rs750934725 Chr.19: 11576308-11576308 on Build GRCh38
rs182019242 Chr.19: 11576599-11576599 on Build GRCh38
rs757465602 Chr.19: 11576483-11576483 on Build GRCh38
rs374919506 Chr.19: 11576258-11576258 on Build GRCh38

View Full Product Details