Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12578083-12578590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF490
Entrez Gene ID
57474
Gene Name
zinc finger protein 490
Gene Aliases
-
UniGene
Hs.655860
Gene Chromosome Location
Chr.19: 12576106-12610809 on Build GRCh38
Transcripts
NM_020714.2

SNP Detail

SNP ID Chromosome Location
rs537735250 Chr.19: 12578546-12578546 on Build GRCh38
rs532536361 Chr.19: 12578215-12578215 on Build GRCh38
rs565264983 Chr.19: 12578198-12578198 on Build GRCh38
rs1045352232 Chr.19: 12578177-12578177 on Build GRCh38
rs1026944688 Chr.19: 12578314-12578314 on Build GRCh38
rs948208340 Chr.19: 12578171-12578171 on Build GRCh38
rs558310205 Chr.19: 12578550-12578550 on Build GRCh38
rs982688458 Chr.19: 12578374-12578374 on Build GRCh38
rs745859180 Chr.19: 12578513-12578513 on Build GRCh38
rs952742430 Chr.19: 12578320-12578320 on Build GRCh38
rs1052538401 Chr.19: 12578187-12578187 on Build GRCh38
rs899582092 Chr.19: 12578551-12578551 on Build GRCh38
rs953950987 Chr.19: 12578446-12578446 on Build GRCh38
rs779198275 Chr.19: 12578346-12578346 on Build GRCh38
rs963904094 Chr.19: 12578233-12578233 on Build GRCh38
rs147813484 Chr.19: 12578350-12578350 on Build GRCh38
rs772249063 Chr.19: 12578472-12578472 on Build GRCh38
rs905220270 Chr.19: 12578522-12578522 on Build GRCh38
rs968210831 Chr.19: 12578395-12578395 on Build GRCh38
rs907832350 Chr.19: 12578118-12578118 on Build GRCh38
rs1037282066 Chr.19: 12578509-12578509 on Build GRCh38
rs1056447059 Chr.19: 12578126-12578126 on Build GRCh38
rs948260545 Chr.19: 12578481-12578481 on Build GRCh38
rs940789249 Chr.19: 12578124-12578124 on Build GRCh38
rs769748547 Chr.19: 12578539-12578539 on Build GRCh38
rs939842893 Chr.19: 12578503-12578503 on Build GRCh38
rs985196213 Chr.19: 12578117-12578117 on Build GRCh38
rs1040851785 Chr.19: 12578526-12578526 on Build GRCh38
rs986704701 Chr.19: 12578461-12578461 on Build GRCh38
rs992229759 Chr.19: 12578420-12578420 on Build GRCh38
rs912488568 Chr.19: 12578462-12578462 on Build GRCh38
rs145988194 Chr.19: 12578185-12578185 on Build GRCh38
rs1015355028 Chr.19: 12578393-12578393 on Build GRCh38
rs1002487854 Chr.19: 12578305-12578305 on Build GRCh38
rs917991529 Chr.19: 12578477-12578477 on Build GRCh38
rs563538621 Chr.19: 12578147-12578147 on Build GRCh38
rs547825803 Chr.19: 12578232-12578232 on Build GRCh38
rs777687337 Chr.19: 12578498-12578498 on Build GRCh38
rs531759221 Chr.19: 12578180-12578180 on Build GRCh38
rs780612346 Chr.19: 12578210-12578210 on Build GRCh38
rs542154440 Chr.19: 12578133-12578133 on Build GRCh38
rs1002437011 Chr.19: 12578545-12578545 on Build GRCh38
rs1010167416 Chr.19: 12578373-12578373 on Build GRCh38
rs115620662 Chr.19: 12578404-12578404 on Build GRCh38
rs796201028 Chr.19: 12578199-12578199 on Build GRCh38
rs192495802 Chr.19: 12578425-12578425 on Build GRCh38
rs1035172081 Chr.19: 12578307-12578307 on Build GRCh38
rs570042919 Chr.19: 12578501-12578501 on Build GRCh38
rs925439664 Chr.19: 12578500-12578500 on Build GRCh38
rs892161071 Chr.19: 12578516-12578516 on Build GRCh38
rs755351515 Chr.19: 12578283-12578283 on Build GRCh38
rs943672897 Chr.19: 12578518-12578518 on Build GRCh38
rs754275493 Chr.19: 12578253-12578253 on Build GRCh38

View Full Product Details