Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12580554-12581056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF490
Entrez Gene ID
57474
Gene Name
zinc finger protein 490
Gene Aliases
-
UniGene
Hs.655860
Gene Chromosome Location
Chr.19: 12576106-12610809 on Build GRCh38
Transcripts
NM_020714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6022770 c.1145G>T p.R382I Chr.19: 12580930-12580930 on Build GRCh38
991334 c.1300C>T p.R434* Chr.19: 12580775-12580775 on Build GRCh38
991335 c.1140C>T p.H380H Chr.19: 12580935-12580935 on Build GRCh38
991332 c.1357C>T p.R453W Chr.19: 12580718-12580718 on Build GRCh38
4644168 c.1212C>T p.Y404Y Chr.19: 12580863-12580863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76668661 Chr.19: 12580833-12580833 on Build GRCh38
rs1045891744 Chr.19: 12580675-12580675 on Build GRCh38
rs746653328 Chr.19: 12580750-12580750 on Build GRCh38
rs771373675 Chr.19: 12580780-12580780 on Build GRCh38
rs762040850 Chr.19: 12580983-12580983 on Build GRCh38
rs562509194 Chr.19: 12580821-12580821 on Build GRCh38
rs780413037 Chr.19: 12580938-12580938 on Build GRCh38
rs754610970 Chr.19: 12580876-12580876 on Build GRCh38
rs774653522 Chr.19: 12580682-12580682 on Build GRCh38
rs866708203 Chr.19: 12580886-12580886 on Build GRCh38
rs199847080 Chr.19: 12580665-12580665 on Build GRCh38
rs755016610 Chr.19: 12580813-12580813 on Build GRCh38
rs534425378 Chr.19: 12580718-12580718 on Build GRCh38
rs140333364 Chr.19: 12581004-12581004 on Build GRCh38
rs753788913 Chr.19: 12580603-12580603 on Build GRCh38
rs755672983 Chr.19: 12581021-12581021 on Build GRCh38
rs769971579 Chr.19: 12580989-12580989 on Build GRCh38
rs941344639 Chr.19: 12580918-12580918 on Build GRCh38
rs977114346 Chr.19: 12580635-12580635 on Build GRCh38
rs918815091 Chr.19: 12580993-12580993 on Build GRCh38
rs1040612761 Chr.19: 12580691-12580691 on Build GRCh38
rs762024644 Chr.19: 12580893-12580894 on Build GRCh38
rs147278541 Chr.19: 12581002-12581002 on Build GRCh38
rs752140802 Chr.19: 12580898-12580898 on Build GRCh38
rs200277578 Chr.19: 12580651-12580651 on Build GRCh38
rs140858103 Chr.19: 12580610-12580610 on Build GRCh38
rs867334373 Chr.19: 12580737-12580737 on Build GRCh38
rs776989207 Chr.19: 12580972-12580972 on Build GRCh38
rs766167854 Chr.19: 12580850-12580850 on Build GRCh38
rs934103212 Chr.19: 12580756-12580756 on Build GRCh38
rs376962945 Chr.19: 12580986-12580986 on Build GRCh38
rs374620519 Chr.19: 12581003-12581003 on Build GRCh38
rs767270705 Chr.19: 12581018-12581018 on Build GRCh38
rs755619749 Chr.19: 12580904-12580904 on Build GRCh38
rs747206114 Chr.19: 12580955-12580955 on Build GRCh38
rs779629101 Chr.19: 12580743-12580743 on Build GRCh38
rs771316199 Chr.19: 12580681-12580681 on Build GRCh38
rs758261428 Chr.19: 12580720-12580720 on Build GRCh38
rs143593697 Chr.19: 12580715-12580715 on Build GRCh38
rs144688605 Chr.19: 12580670-12580670 on Build GRCh38
rs762891577 Chr.19: 12580849-12580849 on Build GRCh38
rs974067760 Chr.19: 12580936-12580936 on Build GRCh38
rs148830742 Chr.19: 12580690-12580690 on Build GRCh38
rs557928943 Chr.19: 12580687-12580687 on Build GRCh38
rs752335248 Chr.19: 12581019-12581019 on Build GRCh38
rs200930919 Chr.19: 12580808-12580808 on Build GRCh38
rs569152894 Chr.19: 12580617-12580617 on Build GRCh38
rs765137320 Chr.19: 12580717-12580717 on Build GRCh38
rs975885913 Chr.19: 12580818-12580818 on Build GRCh38
rs1013049910 Chr.19: 12580824-12580824 on Build GRCh38
rs772473720 Chr.19: 12580816-12580816 on Build GRCh38
rs377377938 Chr.19: 12580772-12580772 on Build GRCh38
rs887128871 Chr.19: 12580793-12580793 on Build GRCh38
rs759119432 Chr.19: 12581010-12581010 on Build GRCh38
rs779618603 Chr.19: 12580654-12580654 on Build GRCh38
rs796743533 Chr.19: 12580909-12580909 on Build GRCh38
rs1020197303 Chr.19: 12580794-12580794 on Build GRCh38
rs143896527 Chr.19: 12580851-12580851 on Build GRCh38
rs73497762 Chr.19: 12580820-12580820 on Build GRCh38
rs376032343 Chr.19: 12580695-12580695 on Build GRCh38
rs767662256 Chr.19: 12580685-12580685 on Build GRCh38
rs746566213 Chr.19: 12580656-12580656 on Build GRCh38
rs377726915 Chr.19: 12580629-12580629 on Build GRCh38
rs780584425 Chr.19: 12580895-12580895 on Build GRCh38
rs1005572997 Chr.19: 12580797-12580797 on Build GRCh38
rs761845059 Chr.19: 12580832-12580832 on Build GRCh38
rs751822142 Chr.19: 12580931-12580931 on Build GRCh38
rs768855346 Chr.19: 12580966-12580966 on Build GRCh38
rs144564838 Chr.19: 12580758-12580758 on Build GRCh38
rs140620156 Chr.19: 12580683-12580683 on Build GRCh38
rs141732776 Chr.19: 12580799-12580799 on Build GRCh38
rs750370813 Chr.19: 12580838-12580838 on Build GRCh38
rs775611926 Chr.19: 12580817-12580817 on Build GRCh38
rs531353057 Chr.19: 12580910-12580910 on Build GRCh38
rs924315276 Chr.19: 12580636-12580636 on Build GRCh38
rs779457890 Chr.19: 12580925-12580925 on Build GRCh38
rs370729346 Chr.19: 12580830-12580830 on Build GRCh38
rs777221425 Chr.19: 12581027-12581027 on Build GRCh38
rs757208480 Chr.19: 12580716-12580716 on Build GRCh38
rs757155250 Chr.19: 12580608-12580608 on Build GRCh38
rs866791610 Chr.19: 12580801-12580801 on Build GRCh38
rs764007344 Chr.19: 12580581-12580581 on Build GRCh38
rs768201159 Chr.19: 12580659-12580659 on Build GRCh38
rs749210086 Chr.19: 12580785-12580785 on Build GRCh38
rs751310789 Chr.19: 12581006-12581006 on Build GRCh38
rs767516361 Chr.19: 12581019-12581020 on Build GRCh38
rs368983745 Chr.19: 12580646-12580646 on Build GRCh38
rs749817479 Chr.19: 12580774-12580774 on Build GRCh38
rs553045052 Chr.19: 12580827-12580827 on Build GRCh38
rs559788529 Chr.19: 12580935-12580935 on Build GRCh38
rs373290922 Chr.19: 12580663-12580663 on Build GRCh38

View Full Product Details