Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43975968-43976473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0B
Entrez Gene ID
533
Gene Name
ATPase H+ transporting V0 subunit b
Gene Aliases
ATP6F, HATPL, VMA16
UniGene
Hs.596514
Gene Chromosome Location
Chr.1: 43974648-43978300 on Build GRCh38
Transcripts
NM_001039457.2 NM_001294333.1 NM_004047.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262454 c.149C>G p.S50C Chr.1: 43976122-43976122 on Build GRCh38
5025263 c.276C>T p.V92V Chr.1: 43976377-43976377 on Build GRCh38
6875791 c.225C>T p.S75S Chr.1: 43976326-43976326 on Build GRCh38
4664463 c.199T>C p.W67R Chr.1: 43976172-43976172 on Build GRCh38
214141 c.271C>A p.L91M Chr.1: 43976372-43976372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773503779 Chr.1: 43976115-43976115 on Build GRCh38
rs768805395 Chr.1: 43976409-43976409 on Build GRCh38
rs72890653 Chr.1: 43975990-43975990 on Build GRCh38
rs757115025 Chr.1: 43976161-43976161 on Build GRCh38
rs761741458 Chr.1: 43976426-43976426 on Build GRCh38
rs1016111717 Chr.1: 43976308-43976308 on Build GRCh38
rs1052620335 Chr.1: 43976126-43976126 on Build GRCh38
rs749598470 Chr.1: 43976212-43976212 on Build GRCh38
rs372238517 Chr.1: 43976254-43976254 on Build GRCh38
rs368220457 Chr.1: 43976252-43976252 on Build GRCh38
rs746037587 Chr.1: 43976223-43976223 on Build GRCh38
rs371622392 Chr.1: 43976195-43976195 on Build GRCh38
rs369879383 Chr.1: 43976291-43976291 on Build GRCh38
rs368096009 Chr.1: 43976185-43976185 on Build GRCh38
rs561149186 Chr.1: 43976193-43976193 on Build GRCh38
rs756321064 Chr.1: 43976375-43976383 on Build GRCh38
rs1000495113 Chr.1: 43976088-43976088 on Build GRCh38
rs541391906 Chr.1: 43976014-43976014 on Build GRCh38
rs751042342 Chr.1: 43976298-43976298 on Build GRCh38
rs896191108 Chr.1: 43976145-43976145 on Build GRCh38
rs763379367 Chr.1: 43976117-43976117 on Build GRCh38
rs774613455 Chr.1: 43976222-43976222 on Build GRCh38
rs753681254 Chr.1: 43976160-43976160 on Build GRCh38
rs776822634 Chr.1: 43976421-43976421 on Build GRCh38
rs1056858580 Chr.1: 43976237-43976237 on Build GRCh38
rs760632445 Chr.1: 43976153-43976153 on Build GRCh38
rs374437044 Chr.1: 43976207-43976207 on Build GRCh38
rs750186065 Chr.1: 43976288-43976288 on Build GRCh38
rs772138028 Chr.1: 43976181-43976181 on Build GRCh38
rs776757840 Chr.1: 43976268-43976268 on Build GRCh38
rs370347873 Chr.1: 43976190-43976191 on Build GRCh38
rs1002115685 Chr.1: 43976243-43976243 on Build GRCh38
rs780705853 Chr.1: 43976311-43976311 on Build GRCh38
rs770267854 Chr.1: 43976197-43976197 on Build GRCh38
rs200691197 Chr.1: 43976270-43976270 on Build GRCh38
rs752866602 Chr.1: 43976282-43976283 on Build GRCh38
rs947478879 Chr.1: 43976387-43976387 on Build GRCh38
rs188542011 Chr.1: 43976218-43976218 on Build GRCh38
rs772418698 Chr.1: 43976385-43976385 on Build GRCh38
rs757711676 Chr.1: 43976320-43976320 on Build GRCh38
rs771660499 Chr.1: 43976085-43976085 on Build GRCh38
rs764092536 Chr.1: 43976265-43976265 on Build GRCh38
rs765350813 Chr.1: 43976355-43976355 on Build GRCh38
rs764184905 Chr.1: 43976157-43976157 on Build GRCh38
rs746276423 Chr.1: 43976365-43976365 on Build GRCh38
rs754984634 Chr.1: 43976151-43976152 on Build GRCh38
rs770286215 Chr.1: 43976109-43976109 on Build GRCh38
rs368947160 Chr.1: 43976070-43976070 on Build GRCh38
rs749764900 Chr.1: 43976317-43976317 on Build GRCh38
rs976819968 Chr.1: 43976340-43976340 on Build GRCh38
rs921246807 Chr.1: 43976350-43976350 on Build GRCh38
rs747061836 Chr.1: 43976404-43976404 on Build GRCh38
rs767595857 Chr.1: 43976147-43976147 on Build GRCh38
rs373170197 Chr.1: 43976394-43976394 on Build GRCh38
rs776584876 Chr.1: 43976263-43976268 on Build GRCh38
rs370541094 Chr.1: 43976073-43976073 on Build GRCh38
rs750042041 Chr.1: 43976179-43976179 on Build GRCh38
rs766074669 Chr.1: 43976292-43976292 on Build GRCh38
rs754535852 Chr.1: 43976299-43976299 on Build GRCh38
rs1027133819 Chr.1: 43976236-43976236 on Build GRCh38
rs373645584 Chr.1: 43976172-43976172 on Build GRCh38
rs951534601 Chr.1: 43976247-43976247 on Build GRCh38
rs779560069 Chr.1: 43976098-43976098 on Build GRCh38
rs765040488 Chr.1: 43976287-43976287 on Build GRCh38
rs752830314 Chr.1: 43976151-43976151 on Build GRCh38
rs774686750 Chr.1: 43976134-43976134 on Build GRCh38
rs771030788 Chr.1: 43976125-43976125 on Build GRCh38
rs552595219 Chr.1: 43975997-43975997 on Build GRCh38
rs921466399 Chr.1: 43976428-43976428 on Build GRCh38
rs776603858 Chr.1: 43976419-43976419 on Build GRCh38
rs760904235 Chr.1: 43976261-43976261 on Build GRCh38
rs759702603 Chr.1: 43976136-43976136 on Build GRCh38
rs758016310 Chr.1: 43976183-43976183 on Build GRCh38
rs377561163 Chr.1: 43976055-43976055 on Build GRCh38
rs779280296 Chr.1: 43976323-43976323 on Build GRCh38
rs373131262 Chr.1: 43976042-43976042 on Build GRCh38

View Full Product Details