Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45627932-45628435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPBP1L1
Entrez Gene ID
60313
Gene Name
GC-rich promoter binding protein 1 like 1
Gene Aliases
SP192
UniGene
Hs.238432
Gene Chromosome Location
Chr.1: 45627304-45686691 on Build GRCh38
Transcripts
NM_021639.4 XM_017002019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936881 c.1334C>T p.S445F Chr.1: 45628347-45628347 on Build GRCh38
3735357 c.1389T>A p.S463R Chr.1: 45628292-45628292 on Build GRCh38
910214 c.1380C>T p.T460T Chr.1: 45628301-45628301 on Build GRCh38
4186799 c.1332C>A p.F444L Chr.1: 45628349-45628349 on Build GRCh38
4687752 c.1381G>A p.E461K Chr.1: 45628300-45628300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779916534 Chr.1: 45628328-45628328 on Build GRCh38
rs982021465 Chr.1: 45627999-45627999 on Build GRCh38
rs368807443 Chr.1: 45628364-45628364 on Build GRCh38
rs897965862 Chr.1: 45628027-45628027 on Build GRCh38
rs369909760 Chr.1: 45627978-45627978 on Build GRCh38
rs983455957 Chr.1: 45628132-45628132 on Build GRCh38
rs988012580 Chr.1: 45627982-45627982 on Build GRCh38
rs757351983 Chr.1: 45628224-45628224 on Build GRCh38
rs144842150 Chr.1: 45628362-45628362 on Build GRCh38
rs562233813 Chr.1: 45628068-45628071 on Build GRCh38
rs775895943 Chr.1: 45628026-45628026 on Build GRCh38
rs551889993 Chr.1: 45628083-45628083 on Build GRCh38
rs535795471 Chr.1: 45628198-45628198 on Build GRCh38
rs766518015 Chr.1: 45628215-45628216 on Build GRCh38
rs148623746 Chr.1: 45628296-45628296 on Build GRCh38
rs889351495 Chr.1: 45628051-45628054 on Build GRCh38
rs146975550 Chr.1: 45628029-45628029 on Build GRCh38
rs745794836 Chr.1: 45628226-45628226 on Build GRCh38
rs768406667 Chr.1: 45628330-45628330 on Build GRCh38
rs775295703 Chr.1: 45628248-45628248 on Build GRCh38
rs753068618 Chr.1: 45628007-45628008 on Build GRCh38
rs970611427 Chr.1: 45628216-45628216 on Build GRCh38
rs753138793 Chr.1: 45628297-45628297 on Build GRCh38
rs760098178 Chr.1: 45628293-45628293 on Build GRCh38
rs761981853 Chr.1: 45628166-45628166 on Build GRCh38
rs368615457 Chr.1: 45627991-45627991 on Build GRCh38
rs1052044983 Chr.1: 45628050-45628050 on Build GRCh38
rs952283596 Chr.1: 45628396-45628396 on Build GRCh38
rs970233562 Chr.1: 45628060-45628060 on Build GRCh38
rs755360795 Chr.1: 45628319-45628320 on Build GRCh38
rs769319262 Chr.1: 45628276-45628276 on Build GRCh38
rs780084622 Chr.1: 45627983-45627983 on Build GRCh38
rs1135850 Chr.1: 45628020-45628020 on Build GRCh38
rs926515787 Chr.1: 45628005-45628006 on Build GRCh38
rs775194402 Chr.1: 45628046-45628046 on Build GRCh38
rs375147861 Chr.1: 45628345-45628345 on Build GRCh38
rs1030659590 Chr.1: 45628312-45628312 on Build GRCh38
rs768870611 Chr.1: 45628097-45628097 on Build GRCh38
rs114265929 Chr.1: 45628197-45628197 on Build GRCh38
rs1036498981 Chr.1: 45628023-45628024 on Build GRCh38
rs900359954 Chr.1: 45628204-45628204 on Build GRCh38
rs576674137 Chr.1: 45628366-45628366 on Build GRCh38
rs554199240 Chr.1: 45628245-45628245 on Build GRCh38
rs999165070 Chr.1: 45628023-45628023 on Build GRCh38
rs367989046 Chr.1: 45628300-45628300 on Build GRCh38
rs767898317 Chr.1: 45628185-45628185 on Build GRCh38
rs1250 Chr.1: 45628355-45628355 on Build GRCh38
rs761298668 Chr.1: 45628273-45628273 on Build GRCh38
rs759279789 Chr.1: 45628239-45628239 on Build GRCh38
rs1009285333 Chr.1: 45628172-45628172 on Build GRCh38
rs181427044 Chr.1: 45628108-45628108 on Build GRCh38
rs751125930 Chr.1: 45628372-45628372 on Build GRCh38
rs757341252 Chr.1: 45628310-45628310 on Build GRCh38
rs138129534 Chr.1: 45628182-45628182 on Build GRCh38
rs779064488 Chr.1: 45628313-45628313 on Build GRCh38
rs776070470 Chr.1: 45628270-45628270 on Build GRCh38
rs991916151 Chr.1: 45628032-45628032 on Build GRCh38
rs541428578 Chr.1: 45628225-45628225 on Build GRCh38
rs12093126 Chr.1: 45628365-45628365 on Build GRCh38
rs957764657 Chr.1: 45627979-45627979 on Build GRCh38
rs944810068 Chr.1: 45628194-45628194 on Build GRCh38
rs138602356 Chr.1: 45628079-45628080 on Build GRCh38
rs765032571 Chr.1: 45628301-45628301 on Build GRCh38
rs867961522 Chr.1: 45627963-45627963 on Build GRCh38
rs760625835 Chr.1: 45628048-45628051 on Build GRCh38
rs756740059 Chr.1: 45628378-45628378 on Build GRCh38
rs1027787816 Chr.1: 45628390-45628390 on Build GRCh38
rs774773127 Chr.1: 45628280-45628280 on Build GRCh38
rs768102444 Chr.1: 45628295-45628295 on Build GRCh38
rs555877653 Chr.1: 45628113-45628113 on Build GRCh38
rs747819985 Chr.1: 45628347-45628347 on Build GRCh38
rs980793757 Chr.1: 45628074-45628074 on Build GRCh38
rs1032008692 Chr.1: 45628090-45628090 on Build GRCh38
rs996518564 Chr.1: 45628006-45628006 on Build GRCh38
rs370823552 Chr.1: 45628322-45628322 on Build GRCh38
rs60972541 Chr.1: 45628271-45628271 on Build GRCh38
rs1024934672 Chr.1: 45628038-45628038 on Build GRCh38
rs751866667 Chr.1: 45628218-45628219 on Build GRCh38
rs556590492 Chr.1: 45628006-45628007 on Build GRCh38
rs754110031 Chr.1: 45628302-45628302 on Build GRCh38
rs749910703 Chr.1: 45628008-45628009 on Build GRCh38
rs1022196980 Chr.1: 45628008-45628008 on Build GRCh38

View Full Product Details