Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14409458-14409962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
UniGene
Hs.311609
Gene Chromosome Location
Chr.19: 14408798-14419383 on Build GRCh38
Transcripts
NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160373 c.771G>A p.T257T Chr.19: 14409835-14409835 on Build GRCh38
4818191 c.803G>A p.*268* Chr.19: 14409911-14409911 on Build GRCh38
5519621 c.756_758delCGA p.D252delD Chr.19: 14409848-14409850 on Build GRCh38
4676810 c.959T>C p.M320T Chr.19: 14409551-14409551 on Build GRCh38
6641724 c.943G>A p.A315T Chr.19: 14409567-14409567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375946796 Chr.19: 14409711-14409711 on Build GRCh38
rs1005877340 Chr.19: 14409553-14409553 on Build GRCh38
rs777924695 Chr.19: 14409492-14409492 on Build GRCh38
rs773047847 Chr.19: 14409538-14409538 on Build GRCh38
rs1055630597 Chr.19: 14409745-14409745 on Build GRCh38
rs180836789 Chr.19: 14409574-14409574 on Build GRCh38
rs372469313 Chr.19: 14409728-14409728 on Build GRCh38
rs887889499 Chr.19: 14409655-14409655 on Build GRCh38
rs149114372 Chr.19: 14409633-14409633 on Build GRCh38
rs779430327 Chr.19: 14409806-14409806 on Build GRCh38
rs759084693 Chr.19: 14409722-14409722 on Build GRCh38
rs747213514 Chr.19: 14409493-14409493 on Build GRCh38
rs371615674 Chr.19: 14409634-14409634 on Build GRCh38
rs777803217 Chr.19: 14409589-14409589 on Build GRCh38
rs2302994 Chr.19: 14409522-14409522 on Build GRCh38
rs754881959 Chr.19: 14409568-14409568 on Build GRCh38
rs778914622 Chr.19: 14409483-14409483 on Build GRCh38
rs775811775 Chr.19: 14409672-14409673 on Build GRCh38
rs184600463 Chr.19: 14409901-14409901 on Build GRCh38
rs370457029 Chr.19: 14409523-14409523 on Build GRCh38
rs372750051 Chr.19: 14409702-14409702 on Build GRCh38
rs759329953 Chr.19: 14409664-14409664 on Build GRCh38
rs1002266758 Chr.19: 14409660-14409660 on Build GRCh38
rs945851723 Chr.19: 14409488-14409488 on Build GRCh38
rs375628786 Chr.19: 14409835-14409835 on Build GRCh38
rs148270304 Chr.19: 14409782-14409782 on Build GRCh38
rs752674619 Chr.19: 14409679-14409679 on Build GRCh38
rs373132705 Chr.19: 14409486-14409486 on Build GRCh38
rs747261610 Chr.19: 14409591-14409591 on Build GRCh38
rs771241139 Chr.19: 14409494-14409494 on Build GRCh38
rs780768591 Chr.19: 14409930-14409930 on Build GRCh38
rs776088660 Chr.19: 14409617-14409617 on Build GRCh38
rs751601500 Chr.19: 14409698-14409698 on Build GRCh38
rs750697093 Chr.19: 14409773-14409773 on Build GRCh38
rs755445718 Chr.19: 14409916-14409916 on Build GRCh38
rs765490744 Chr.19: 14409919-14409919 on Build GRCh38
rs761902909 Chr.19: 14409751-14409751 on Build GRCh38
rs770515371 Chr.19: 14409714-14409714 on Build GRCh38
rs1034232766 Chr.19: 14409542-14409542 on Build GRCh38
rs777049345 Chr.19: 14409503-14409503 on Build GRCh38
rs771535082 Chr.19: 14409735-14409736 on Build GRCh38
rs957135947 Chr.19: 14409825-14409825 on Build GRCh38
rs772315826 Chr.19: 14409490-14409490 on Build GRCh38
rs769817457 Chr.19: 14409609-14409609 on Build GRCh38
rs756399705 Chr.19: 14409781-14409781 on Build GRCh38
rs770362905 Chr.19: 14409487-14409488 on Build GRCh38
rs560901519 Chr.19: 14409853-14409853 on Build GRCh38
rs556046710 Chr.19: 14409829-14409829 on Build GRCh38
rs371297874 Chr.19: 14409514-14409514 on Build GRCh38
rs753317889 Chr.19: 14409926-14409926 on Build GRCh38
rs201894828 Chr.19: 14409615-14409615 on Build GRCh38
rs910223775 Chr.19: 14409894-14409894 on Build GRCh38
rs758526835 Chr.19: 14409587-14409587 on Build GRCh38
rs143209973 Chr.19: 14409756-14409756 on Build GRCh38
rs897653601 Chr.19: 14409575-14409575 on Build GRCh38
rs775269512 Chr.19: 14409852-14409852 on Build GRCh38
rs141278207 Chr.19: 14409810-14409810 on Build GRCh38
rs780909815 Chr.19: 14409880-14409880 on Build GRCh38
rs746421482 Chr.19: 14409518-14409518 on Build GRCh38
rs756595849 Chr.19: 14409705-14409705 on Build GRCh38
rs58961341 Chr.19: 14409527-14409527 on Build GRCh38
rs35237200 Chr.19: 14409866-14409867 on Build GRCh38
rs750453454 Chr.19: 14409898-14409898 on Build GRCh38
rs769379830 Chr.19: 14409850-14409850 on Build GRCh38
rs146936895 Chr.19: 14409579-14409579 on Build GRCh38
rs750773177 Chr.19: 14409703-14409703 on Build GRCh38
rs371219917 Chr.19: 14409783-14409783 on Build GRCh38
rs376030027 Chr.19: 14409593-14409593 on Build GRCh38
rs985813372 Chr.19: 14409841-14409841 on Build GRCh38
rs367978289 Chr.19: 14409821-14409821 on Build GRCh38
rs768601340 Chr.19: 14409859-14409859 on Build GRCh38
rs199873900 Chr.19: 14409900-14409900 on Build GRCh38
rs373060190 Chr.19: 14409865-14409865 on Build GRCh38
rs748162385 Chr.19: 14409487-14409487 on Build GRCh38
rs766149625 Chr.19: 14409559-14409559 on Build GRCh38
rs757427136 Chr.19: 14409701-14409701 on Build GRCh38
rs761623802 Chr.19: 14409870-14409870 on Build GRCh38
rs147553508 Chr.19: 14409595-14409595 on Build GRCh38
rs979655571 Chr.19: 14409724-14409724 on Build GRCh38
rs770279031 Chr.19: 14409521-14409521 on Build GRCh38
rs776396984 Chr.19: 14409715-14409715 on Build GRCh38
rs764998809 Chr.19: 14409672-14409672 on Build GRCh38
rs1014258868 Chr.19: 14409697-14409697 on Build GRCh38
rs772912455 Chr.19: 14409877-14409879 on Build GRCh38
rs374387661 Chr.19: 14409790-14409790 on Build GRCh38
rs767701980 Chr.19: 14409896-14409896 on Build GRCh38
rs763954815 Chr.19: 14409735-14409735 on Build GRCh38
rs762891875 Chr.19: 14409732-14409732 on Build GRCh38
rs141207973 Chr.19: 14409567-14409567 on Build GRCh38
rs751624181 Chr.19: 14409736-14409736 on Build GRCh38
rs758821810 Chr.19: 14409927-14409927 on Build GRCh38
rs769646984 Chr.19: 14409725-14409725 on Build GRCh38
rs913922343 Chr.19: 14409687-14409687 on Build GRCh38
rs749790285 Chr.19: 14409630-14409630 on Build GRCh38
rs200339346 Chr.19: 14409592-14409592 on Build GRCh38
rs765996110 Chr.19: 14409639-14409639 on Build GRCh38
rs745318006 Chr.19: 14409936-14409936 on Build GRCh38
rs369375673 Chr.19: 14409713-14409713 on Build GRCh38
rs764216943 Chr.19: 14409683-14409683 on Build GRCh38
rs760339680 Chr.19: 14409552-14409552 on Build GRCh38
rs761435009 Chr.19: 14409731-14409731 on Build GRCh38
rs376132514 Chr.19: 14409682-14409682 on Build GRCh38
rs780534617 Chr.19: 14409710-14409710 on Build GRCh38
rs776325434 Chr.19: 14409659-14409659 on Build GRCh38

View Full Product Details