Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17601983-17602475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC13A
Entrez Gene ID
23025
Gene Name
unc-13 homolog A
Gene Aliases
Munc13-1
UniGene
Hs.164502
Gene Chromosome Location
Chr.19: 17601328-17688344 on Build GRCh38
Transcripts
NM_001080421.2 XM_011527810.2 XM_011527811.2 XM_017026502.1

SNP Detail

SNP ID Chromosome Location
rs945546427 Chr.19: 17602241-17602241 on Build GRCh38
rs552925372 Chr.19: 17602193-17602193 on Build GRCh38
rs73922142 Chr.19: 17602252-17602252 on Build GRCh38
rs1024403581 Chr.19: 17602371-17602371 on Build GRCh38
rs976185529 Chr.19: 17602452-17602452 on Build GRCh38
rs901610476 Chr.19: 17602142-17602142 on Build GRCh38
rs943417915 Chr.19: 17602445-17602445 on Build GRCh38
rs892601510 Chr.19: 17602236-17602236 on Build GRCh38
rs1012643202 Chr.19: 17602359-17602359 on Build GRCh38
rs1039545472 Chr.19: 17602302-17602302 on Build GRCh38
rs886734383 Chr.19: 17602332-17602332 on Build GRCh38
rs1016586995 Chr.19: 17602337-17602337 on Build GRCh38
rs980816337 Chr.19: 17602034-17602034 on Build GRCh38
rs541402882 Chr.19: 17602382-17602382 on Build GRCh38
rs1039600150 Chr.19: 17602066-17602066 on Build GRCh38
rs758259071 Chr.19: 17602184-17602184 on Build GRCh38
rs765528204 Chr.19: 17602146-17602146 on Build GRCh38
rs975922588 Chr.19: 17602352-17602352 on Build GRCh38
rs181117570 Chr.19: 17602111-17602111 on Build GRCh38
rs901032258 Chr.19: 17602318-17602318 on Build GRCh38
rs914096116 Chr.19: 17602229-17602229 on Build GRCh38
rs990142356 Chr.19: 17602414-17602414 on Build GRCh38
rs889054685 Chr.19: 17602440-17602440 on Build GRCh38
rs893612154 Chr.19: 17602043-17602043 on Build GRCh38
rs139575379 Chr.19: 17602329-17602329 on Build GRCh38
rs560717155 Chr.19: 17602420-17602420 on Build GRCh38
rs977718719 Chr.19: 17602218-17602218 on Build GRCh38
rs752068647 Chr.19: 17602072-17602072 on Build GRCh38
rs537800196 Chr.19: 17602199-17602199 on Build GRCh38
rs936349221 Chr.19: 17602444-17602444 on Build GRCh38
rs1054910818 Chr.19: 17602237-17602237 on Build GRCh38
rs971601258 Chr.19: 17602197-17602197 on Build GRCh38
rs1050640506 Chr.19: 17602427-17602427 on Build GRCh38
rs73922285 Chr.19: 17602127-17602127 on Build GRCh38
rs1050493180 Chr.19: 17602434-17602434 on Build GRCh38
rs73922140 Chr.19: 17602165-17602165 on Build GRCh38
rs1026982476 Chr.19: 17602313-17602318 on Build GRCh38
rs997196371 Chr.19: 17602425-17602425 on Build GRCh38
rs189996040 Chr.19: 17602031-17602031 on Build GRCh38
rs969482327 Chr.19: 17602253-17602253 on Build GRCh38
rs922394104 Chr.19: 17602453-17602453 on Build GRCh38
rs917441899 Chr.19: 17602189-17602189 on Build GRCh38
rs936058848 Chr.19: 17602020-17602020 on Build GRCh38
rs185323574 Chr.19: 17602246-17602246 on Build GRCh38
rs971601531 Chr.19: 17602381-17602381 on Build GRCh38
rs188172335 Chr.19: 17602338-17602338 on Build GRCh38
rs1016641118 Chr.19: 17602171-17602171 on Build GRCh38
rs958488121 Chr.19: 17602183-17602183 on Build GRCh38
rs564779882 Chr.19: 17602336-17602336 on Build GRCh38
rs903551879 Chr.19: 17602170-17602170 on Build GRCh38
rs924891552 Chr.19: 17602392-17602392 on Build GRCh38
rs572593484 Chr.19: 17602230-17602230 on Build GRCh38
rs981295167 Chr.19: 17602279-17602279 on Build GRCh38
rs73922141 Chr.19: 17602247-17602247 on Build GRCh38
rs957897956 Chr.19: 17602395-17602395 on Build GRCh38
rs1006733481 Chr.19: 17602044-17602044 on Build GRCh38
rs566930298 Chr.19: 17602030-17602030 on Build GRCh38
rs569280513 Chr.19: 17602098-17602098 on Build GRCh38
rs370187049 Chr.19: 17602342-17602342 on Build GRCh38
rs907667313 Chr.19: 17602068-17602068 on Build GRCh38
rs1005678813 Chr.19: 17602166-17602166 on Build GRCh38
rs537299040 Chr.19: 17602099-17602099 on Build GRCh38
rs539348168 Chr.19: 17602319-17602319 on Build GRCh38
rs936025327 Chr.19: 17602234-17602234 on Build GRCh38

View Full Product Details