Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 30443833-30444321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF536
Entrez Gene ID
9745
Gene Name
zinc finger protein 536
Gene Aliases
-
UniGene
Hs.378901
Gene Chromosome Location
Chr.19: 30224236-30713538 on Build GRCh38
Transcripts
NM_014717.2 XM_011527554.2 XM_011527555.2 XM_011527557.2 XM_011527558.2 XM_011527560.2 XM_017027527.1 XM_017027528.1 XM_017027529.1 XM_017027530.1 XM_017027531.1 XM_017027532.1 XM_017027533.1 XM_017027534.1 XM_017027535.1 XM_017027536.1 XM_017027537.1 XM_017027538.1 XM_017027539.1 XM_017027540.1 XM_017027541.1 XM_017027542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507440 c.405C>A p.C135* Chr.19: 30443967-30443967 on Build GRCh38
4656283 c.701C>T p.P234L Chr.19: 30444263-30444263 on Build GRCh38
401825 c.471C>A p.P157P Chr.19: 30444033-30444033 on Build GRCh38
6831050 c.684G>A p.P228P Chr.19: 30444246-30444246 on Build GRCh38
2750201 c.374G>A p.R125H Chr.19: 30443936-30443936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029087352 Chr.19: 30443913-30443913 on Build GRCh38
rs778595563 Chr.19: 30443962-30443962 on Build GRCh38
rs529033826 Chr.19: 30444289-30444289 on Build GRCh38
rs768351285 Chr.19: 30443877-30443877 on Build GRCh38
rs768839903 Chr.19: 30444069-30444069 on Build GRCh38
rs757128655 Chr.19: 30444270-30444270 on Build GRCh38
rs781063881 Chr.19: 30444271-30444271 on Build GRCh38
rs771752100 Chr.19: 30443888-30443888 on Build GRCh38
rs202070946 Chr.19: 30444293-30444293 on Build GRCh38
rs761872698 Chr.19: 30444093-30444093 on Build GRCh38
rs574116352 Chr.19: 30444108-30444108 on Build GRCh38
rs547189963 Chr.19: 30444066-30444066 on Build GRCh38
rs201984847 Chr.19: 30443967-30443967 on Build GRCh38
rs767953644 Chr.19: 30444094-30444094 on Build GRCh38
rs762293077 Chr.19: 30443923-30443923 on Build GRCh38
rs563610630 Chr.19: 30444288-30444288 on Build GRCh38
rs542393059 Chr.19: 30444225-30444225 on Build GRCh38
rs754333227 Chr.19: 30444126-30444126 on Build GRCh38
rs1040675243 Chr.19: 30444178-30444178 on Build GRCh38
rs745738230 Chr.19: 30444177-30444177 on Build GRCh38
rs750879270 Chr.19: 30443853-30443853 on Build GRCh38
rs771316228 Chr.19: 30444299-30444299 on Build GRCh38
rs756533351 Chr.19: 30443934-30443934 on Build GRCh38
rs747168364 Chr.19: 30444297-30444297 on Build GRCh38
rs767712502 Chr.19: 30443929-30443929 on Build GRCh38
rs749435165 Chr.19: 30444141-30444141 on Build GRCh38
rs543213590 Chr.19: 30444145-30444145 on Build GRCh38
rs768856443 Chr.19: 30444188-30444188 on Build GRCh38
rs375960895 Chr.19: 30444153-30444153 on Build GRCh38
rs748244776 Chr.19: 30444292-30444292 on Build GRCh38
rs1046845420 Chr.19: 30444148-30444148 on Build GRCh38
rs764450173 Chr.19: 30443916-30443916 on Build GRCh38
rs745829931 Chr.19: 30444280-30444280 on Build GRCh38
rs757892372 Chr.19: 30444263-30444263 on Build GRCh38
rs757112692 Chr.19: 30444154-30444154 on Build GRCh38
rs573585108 Chr.19: 30443914-30443914 on Build GRCh38
rs780520045 Chr.19: 30443954-30443954 on Build GRCh38
rs368903327 Chr.19: 30444045-30444045 on Build GRCh38
rs766672931 Chr.19: 30444243-30444243 on Build GRCh38
rs773216743 Chr.19: 30443891-30443891 on Build GRCh38
rs766942588 Chr.19: 30444114-30444114 on Build GRCh38
rs201200479 Chr.19: 30443901-30443901 on Build GRCh38
rs994581539 Chr.19: 30443904-30443904 on Build GRCh38
rs777458781 Chr.19: 30444151-30444151 on Build GRCh38
rs778509421 Chr.19: 30443880-30443880 on Build GRCh38
rs771139840 Chr.19: 30444210-30444210 on Build GRCh38
rs771040026 Chr.19: 30444015-30444015 on Build GRCh38
rs555455357 Chr.19: 30444166-30444166 on Build GRCh38
rs752428408 Chr.19: 30444134-30444134 on Build GRCh38
rs992138232 Chr.19: 30444007-30444007 on Build GRCh38
rs139989586 Chr.19: 30443854-30443854 on Build GRCh38
rs748239710 Chr.19: 30444208-30444208 on Build GRCh38
rs919746440 Chr.19: 30444123-30444123 on Build GRCh38
rs144245375 Chr.19: 30443883-30443883 on Build GRCh38
rs904025179 Chr.19: 30444224-30444224 on Build GRCh38
rs543321339 Chr.19: 30444264-30444264 on Build GRCh38
rs201076928 Chr.19: 30444199-30444199 on Build GRCh38
rs773509386 Chr.19: 30444136-30444136 on Build GRCh38
rs774642001 Chr.19: 30444075-30444075 on Build GRCh38
rs780013345 Chr.19: 30444291-30444291 on Build GRCh38
rs572216856 Chr.19: 30444176-30444176 on Build GRCh38
rs777391713 Chr.19: 30443974-30443974 on Build GRCh38
rs763191371 Chr.19: 30444054-30444054 on Build GRCh38
rs752197056 Chr.19: 30444260-30444260 on Build GRCh38
rs149579708 Chr.19: 30443861-30443861 on Build GRCh38
rs868226035 Chr.19: 30444025-30444025 on Build GRCh38
rs760463207 Chr.19: 30443892-30443892 on Build GRCh38
rs148725432 Chr.19: 30444105-30444105 on Build GRCh38
rs866467288 Chr.19: 30443936-30443936 on Build GRCh38
rs771905828 Chr.19: 30443973-30443973 on Build GRCh38
rs557658131 Chr.19: 30444058-30444058 on Build GRCh38
rs764660596 Chr.19: 30444132-30444132 on Build GRCh38
rs577844999 Chr.19: 30444240-30444240 on Build GRCh38
rs769949653 Chr.19: 30443896-30443896 on Build GRCh38
rs750940266 Chr.19: 30443931-30443931 on Build GRCh38
rs751825343 Chr.19: 30443917-30443917 on Build GRCh38
rs867380993 Chr.19: 30444010-30444010 on Build GRCh38
rs776933304 Chr.19: 30444301-30444301 on Build GRCh38
rs746988855 Chr.19: 30443980-30443980 on Build GRCh38
rs956913563 Chr.19: 30443999-30443999 on Build GRCh38
rs896581213 Chr.19: 30443932-30443932 on Build GRCh38
rs537372243 Chr.19: 30443961-30443961 on Build GRCh38
rs899516990 Chr.19: 30444191-30444191 on Build GRCh38
rs79971318 Chr.19: 30444294-30444294 on Build GRCh38
rs764827458 Chr.19: 30444250-30444250 on Build GRCh38
rs528598169 Chr.19: 30444304-30444304 on Build GRCh38
rs759997778 Chr.19: 30444245-30444245 on Build GRCh38

View Full Product Details