Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40071417-40071921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF780A
Entrez Gene ID
284323
Gene Name
zinc finger protein 780A
Gene Aliases
ZNF780
UniGene
Hs.115140
Gene Chromosome Location
Chr.19: 40069152-40091706 on Build GRCh38
Transcripts
NM_001142579.1 XM_005258773.2 XM_006723150.3 XM_011526771.2 XM_011526772.2 XM_017026611.1 XM_017026612.1 XM_017026613.1 XM_017026614.1 XM_017026615.1 XM_017026616.1 XM_017026617.1 XM_017026618.1 XM_017026619.1

SNP Detail

SNP ID Chromosome Location
rs749814855 Chr.19: 40071818-40071818 on Build GRCh38
rs932806293 Chr.19: 40071831-40071831 on Build GRCh38
rs944138097 Chr.19: 40071616-40071616 on Build GRCh38
rs929993339 Chr.19: 40071580-40071580 on Build GRCh38
rs867949641 Chr.19: 40071861-40071861 on Build GRCh38
rs1047512462 Chr.19: 40071595-40071595 on Build GRCh38
rs891171014 Chr.19: 40071607-40071607 on Build GRCh38
rs974321425 Chr.19: 40071763-40071763 on Build GRCh38
rs1035355305 Chr.19: 40071610-40071610 on Build GRCh38
rs977709429 Chr.19: 40071720-40071720 on Build GRCh38
rs922862691 Chr.19: 40071821-40071821 on Build GRCh38
rs1009872947 Chr.19: 40071705-40071705 on Build GRCh38
rs542254176 Chr.19: 40071663-40071663 on Build GRCh38
rs539390487 Chr.19: 40071443-40071443 on Build GRCh38
rs892798176 Chr.19: 40071658-40071658 on Build GRCh38
rs971140243 Chr.19: 40071714-40071714 on Build GRCh38
rs1049811296 Chr.19: 40071495-40071495 on Build GRCh38
rs1003495391 Chr.19: 40071513-40071513 on Build GRCh38
rs967802724 Chr.19: 40071659-40071659 on Build GRCh38
rs752808256 Chr.19: 40071644-40071644 on Build GRCh38
rs974342906 Chr.19: 40071527-40071527 on Build GRCh38
rs997425142 Chr.19: 40071463-40071463 on Build GRCh38
rs146236625 Chr.19: 40071631-40071631 on Build GRCh38
rs991080013 Chr.19: 40071655-40071655 on Build GRCh38
rs971564253 Chr.19: 40071450-40071450 on Build GRCh38
rs888545483 Chr.19: 40071496-40071496 on Build GRCh38
rs1034513192 Chr.19: 40071754-40071754 on Build GRCh38
rs927473549 Chr.19: 40071497-40071497 on Build GRCh38
rs1003051759 Chr.19: 40071752-40071752 on Build GRCh38
rs868179994 Chr.19: 40071865-40071865 on Build GRCh38
rs764292123 Chr.19: 40071788-40071788 on Build GRCh38
rs1005542135 Chr.19: 40071505-40071505 on Build GRCh38
rs961279748 Chr.19: 40071511-40071511 on Build GRCh38
rs337781 Chr.19: 40071457-40071457 on Build GRCh38
rs1020375570 Chr.19: 40071707-40071707 on Build GRCh38
rs77224922 Chr.19: 40071760-40071760 on Build GRCh38
rs183388669 Chr.19: 40071656-40071656 on Build GRCh38
rs553528493 Chr.19: 40071646-40071646 on Build GRCh38
rs73549804 Chr.19: 40071510-40071510 on Build GRCh38
rs919921102 Chr.19: 40071578-40071578 on Build GRCh38

View Full Product Details