Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40422455-40422981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERTAD1
Entrez Gene ID
29950
Gene Name
SERTA domain containing 1
Gene Aliases
SEI1, TRIP-Br1
UniGene
Hs.269898
Gene Chromosome Location
Chr.19: 40422501-40426025 on Build GRCh38
Transcripts
NM_013376.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239506 c.671G>A p.G224D Chr.19: 40422876-40422876 on Build GRCh38
1225314 c.586C>G p.P196A Chr.19: 40422961-40422961 on Build GRCh38
6839913 c.628C>G p.L210V Chr.19: 40422919-40422919 on Build GRCh38
1225313 c.578G>A p.G193D Chr.19: 40422969-40422969 on Build GRCh38
247261 c.693G>T p.P231P Chr.19: 40422854-40422854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781481801 Chr.19: 40422859-40422859 on Build GRCh38
rs922790236 Chr.19: 40422654-40422654 on Build GRCh38
rs529683769 Chr.19: 40422823-40422823 on Build GRCh38
rs773175741 Chr.19: 40422941-40422941 on Build GRCh38
rs922709114 Chr.19: 40422864-40422864 on Build GRCh38
rs934144586 Chr.19: 40422881-40422881 on Build GRCh38
rs200693640 Chr.19: 40422852-40422852 on Build GRCh38
rs565488211 Chr.19: 40422683-40422683 on Build GRCh38
rs750226128 Chr.19: 40422655-40422655 on Build GRCh38
rs765361452 Chr.19: 40422839-40422839 on Build GRCh38
rs758512962 Chr.19: 40422830-40422830 on Build GRCh38
rs200723077 Chr.19: 40422829-40422829 on Build GRCh38
rs755975301 Chr.19: 40422691-40422691 on Build GRCh38
rs745420561 Chr.19: 40422904-40422904 on Build GRCh38
rs889348752 Chr.19: 40422813-40422814 on Build GRCh38
rs778221847 Chr.19: 40422810-40422810 on Build GRCh38
rs201246504 Chr.19: 40422813-40422813 on Build GRCh38
rs763072155 Chr.19: 40422848-40422848 on Build GRCh38
rs778330919 Chr.19: 40422886-40422886 on Build GRCh38
rs770491400 Chr.19: 40422804-40422804 on Build GRCh38
rs1038592974 Chr.19: 40422925-40422925 on Build GRCh38
rs372247621 Chr.19: 40422924-40422924 on Build GRCh38
rs777968444 Chr.19: 40422845-40422845 on Build GRCh38
rs993222535 Chr.19: 40422719-40422719 on Build GRCh38
rs149466244 Chr.19: 40422542-40422542 on Build GRCh38
rs572433735 Chr.19: 40422498-40422498 on Build GRCh38
rs750532808 Chr.19: 40422840-40422840 on Build GRCh38
rs780080872 Chr.19: 40422755-40422755 on Build GRCh38
rs898829090 Chr.19: 40422927-40422927 on Build GRCh38
rs898835040 Chr.19: 40422702-40422702 on Build GRCh38
rs978733758 Chr.19: 40422556-40422556 on Build GRCh38
rs984704481 Chr.19: 40422517-40422517 on Build GRCh38
rs200028938 Chr.19: 40422907-40422907 on Build GRCh38
rs941582459 Chr.19: 40422919-40422919 on Build GRCh38
rs368033046 Chr.19: 40422908-40422908 on Build GRCh38
rs890061658 Chr.19: 40422682-40422682 on Build GRCh38
rs183707175 Chr.19: 40422707-40422707 on Build GRCh38
rs1036561607 Chr.19: 40422748-40422748 on Build GRCh38
rs576810872 Chr.19: 40422587-40422587 on Build GRCh38
rs774572011 Chr.19: 40422955-40422955 on Build GRCh38
rs916453589 Chr.19: 40422665-40422666 on Build GRCh38
rs746610064 Chr.19: 40422909-40422909 on Build GRCh38
rs372415034 Chr.19: 40422794-40422794 on Build GRCh38
rs771680640 Chr.19: 40422814-40422814 on Build GRCh38
rs934189826 Chr.19: 40422665-40422665 on Build GRCh38
rs989301672 Chr.19: 40422610-40422610 on Build GRCh38
rs563199092 Chr.19: 40422851-40422851 on Build GRCh38
rs769674082 Chr.19: 40422923-40422923 on Build GRCh38
rs1014118587 Chr.19: 40422772-40422772 on Build GRCh38
rs552052722 Chr.19: 40422883-40422883 on Build GRCh38
rs530723602 Chr.19: 40422855-40422855 on Build GRCh38
rs538736763 Chr.19: 40422913-40422913 on Build GRCh38
rs144833504 Chr.19: 40422552-40422552 on Build GRCh38
rs938703491 Chr.19: 40422611-40422611 on Build GRCh38
rs967578214 Chr.19: 40422849-40422849 on Build GRCh38
rs139610928 Chr.19: 40422858-40422858 on Build GRCh38
rs761661919 Chr.19: 40422914-40422914 on Build GRCh38
rs375132769 Chr.19: 40422865-40422865 on Build GRCh38
rs562618721 Chr.19: 40422942-40422942 on Build GRCh38
rs1032972167 Chr.19: 40422841-40422841 on Build GRCh38
rs992074433 Chr.19: 40422653-40422653 on Build GRCh38
rs1049955461 Chr.19: 40422680-40422680 on Build GRCh38
rs543989748 Chr.19: 40422592-40422592 on Build GRCh38
rs766436934 Chr.19: 40422946-40422946 on Build GRCh38
rs759690841 Chr.19: 40422957-40422957 on Build GRCh38
rs895637074 Chr.19: 40422759-40422759 on Build GRCh38
rs997621470 Chr.19: 40422778-40422778 on Build GRCh38
rs753209821 Chr.19: 40422877-40422877 on Build GRCh38
rs997777271 Chr.19: 40422547-40422547 on Build GRCh38
rs967421494 Chr.19: 40422572-40422572 on Build GRCh38
rs541146103 Chr.19: 40422762-40422762 on Build GRCh38
rs768402366 Chr.19: 40422716-40422716 on Build GRCh38
rs925958442 Chr.19: 40422557-40422557 on Build GRCh38

View Full Product Details