Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40423039-40423551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERTAD1
Entrez Gene ID
29950
Gene Name
SERTA domain containing 1
Gene Aliases
SEI1, TRIP-Br1
UniGene
Hs.269898
Gene Chromosome Location
Chr.19: 40422501-40426025 on Build GRCh38
Transcripts
NM_013376.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3225633 c.96G>A p.A32A Chr.19: 40423451-40423451 on Build GRCh38
6857782 c.447G>A p.A149A Chr.19: 40423100-40423100 on Build GRCh38
355435 c.439G>A p.G147R Chr.19: 40423108-40423108 on Build GRCh38
7200369 c.394C>T p.P132S Chr.19: 40423153-40423153 on Build GRCh38
3534071 c.254C>T p.A85V Chr.19: 40423293-40423293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201866343 Chr.19: 40423452-40423452 on Build GRCh38
rs745988772 Chr.19: 40423148-40423148 on Build GRCh38
rs771521516 Chr.19: 40423444-40423444 on Build GRCh38
rs770915429 Chr.19: 40423127-40423127 on Build GRCh38
rs868367130 Chr.19: 40423197-40423197 on Build GRCh38
rs765833634 Chr.19: 40423101-40423101 on Build GRCh38
rs772101025 Chr.19: 40423155-40423155 on Build GRCh38
rs775597230 Chr.19: 40423163-40423163 on Build GRCh38
rs763220495 Chr.19: 40423502-40423507 on Build GRCh38
rs778120948 Chr.19: 40423513-40423513 on Build GRCh38
rs142064816 Chr.19: 40423305-40423305 on Build GRCh38
rs760082961 Chr.19: 40423364-40423364 on Build GRCh38
rs753449567 Chr.19: 40423394-40423394 on Build GRCh38
rs566138367 Chr.19: 40423457-40423457 on Build GRCh38
rs11558326 Chr.19: 40423303-40423303 on Build GRCh38
rs761292442 Chr.19: 40423523-40423523 on Build GRCh38
rs755864752 Chr.19: 40423266-40423266 on Build GRCh38
rs763491575 Chr.19: 40423357-40423357 on Build GRCh38
rs757714537 Chr.19: 40423100-40423100 on Build GRCh38
rs202084984 Chr.19: 40423361-40423361 on Build GRCh38
rs754512069 Chr.19: 40423110-40423110 on Build GRCh38
rs111302483 Chr.19: 40423298-40423298 on Build GRCh38
rs984656597 Chr.19: 40423281-40423281 on Build GRCh38
rs752301347 Chr.19: 40423236-40423236 on Build GRCh38
rs1010169757 Chr.19: 40423415-40423415 on Build GRCh38
rs776691949 Chr.19: 40423323-40423323 on Build GRCh38
rs1054362899 Chr.19: 40423376-40423376 on Build GRCh38
rs1013009204 Chr.19: 40423208-40423208 on Build GRCh38
rs139675217 Chr.19: 40423064-40423064 on Build GRCh38
rs115956398 Chr.19: 40423525-40423525 on Build GRCh38
rs1010678046 Chr.19: 40423066-40423066 on Build GRCh38
rs559448634 Chr.19: 40423313-40423313 on Build GRCh38
rs918774554 Chr.19: 40423465-40423465 on Build GRCh38
rs760978076 Chr.19: 40423060-40423060 on Build GRCh38
rs766377309 Chr.19: 40423474-40423474 on Build GRCh38
rs776280103 Chr.19: 40423453-40423453 on Build GRCh38
rs149858128 Chr.19: 40423113-40423113 on Build GRCh38
rs530688337 Chr.19: 40423418-40423418 on Build GRCh38
rs761387596 Chr.19: 40423454-40423454 on Build GRCh38
rs537760535 Chr.19: 40423195-40423195 on Build GRCh38
rs745614340 Chr.19: 40423292-40423293 on Build GRCh38
rs747793204 Chr.19: 40423112-40423112 on Build GRCh38
rs143056697 Chr.19: 40423434-40423434 on Build GRCh38
rs759144744 Chr.19: 40423227-40423227 on Build GRCh38
rs773784248 Chr.19: 40423438-40423438 on Build GRCh38
rs769620014 Chr.19: 40423480-40423481 on Build GRCh38
rs574200748 Chr.19: 40423328-40423328 on Build GRCh38
rs774691219 Chr.19: 40423446-40423446 on Build GRCh38
rs768585777 Chr.19: 40423432-40423432 on Build GRCh38
rs757067120 Chr.19: 40423399-40423399 on Build GRCh38
rs867317282 Chr.19: 40423156-40423156 on Build GRCh38
rs781029127 Chr.19: 40423111-40423111 on Build GRCh38
rs768979696 Chr.19: 40423321-40423321 on Build GRCh38
rs747158875 Chr.19: 40423430-40423430 on Build GRCh38
rs540762016 Chr.19: 40423310-40423310 on Build GRCh38
rs750924856 Chr.19: 40423226-40423226 on Build GRCh38
rs773874319 Chr.19: 40423346-40423346 on Build GRCh38
rs960358763 Chr.19: 40423169-40423169 on Build GRCh38
rs552907396 Chr.19: 40423358-40423358 on Build GRCh38
rs770291578 Chr.19: 40423435-40423435 on Build GRCh38
rs752312191 Chr.19: 40423362-40423362 on Build GRCh38
rs756467050 Chr.19: 40423502-40423502 on Build GRCh38
rs373618032 Chr.19: 40423429-40423429 on Build GRCh38
rs528036940 Chr.19: 40423339-40423339 on Build GRCh38
rs576772456 Chr.19: 40423104-40423104 on Build GRCh38
rs949742679 Chr.19: 40423311-40423311 on Build GRCh38
rs777118791 Chr.19: 40423200-40423200 on Build GRCh38
rs781216600 Chr.19: 40423155-40423155 on Build GRCh38
rs891684715 Chr.19: 40423387-40423387 on Build GRCh38
rs572956804 Chr.19: 40423416-40423416 on Build GRCh38
rs755104501 Chr.19: 40423481-40423481 on Build GRCh38
rs372266997 Chr.19: 40423518-40423518 on Build GRCh38
rs781504064 Chr.19: 40423433-40423433 on Build GRCh38
rs367871638 Chr.19: 40423136-40423136 on Build GRCh38
rs749659665 Chr.19: 40423440-40423440 on Build GRCh38
rs151146859 Chr.19: 40423451-40423451 on Build GRCh38
rs748237161 Chr.19: 40423484-40423484 on Build GRCh38
rs768719985 Chr.19: 40423501-40423502 on Build GRCh38
rs905684702 Chr.19: 40423336-40423336 on Build GRCh38
rs962889600 Chr.19: 40423223-40423223 on Build GRCh38
rs765892300 Chr.19: 40423222-40423222 on Build GRCh38
rs747053056 Chr.19: 40423320-40423320 on Build GRCh38
rs773895593 Chr.19: 40423478-40423478 on Build GRCh38
rs4150991 Chr.19: 40423061-40423061 on Build GRCh38
rs952756001 Chr.19: 40423248-40423248 on Build GRCh38
rs930261032 Chr.19: 40423296-40423296 on Build GRCh38
rs775670147 Chr.19: 40423502-40423504 on Build GRCh38
rs145548575 Chr.19: 40423297-40423297 on Build GRCh38
rs749035388 Chr.19: 40423120-40423120 on Build GRCh38
rs370352296 Chr.19: 40423231-40423231 on Build GRCh38
rs140122572 Chr.19: 40423291-40423291 on Build GRCh38
rs147951593 Chr.19: 40423119-40423119 on Build GRCh38
rs368916899 Chr.19: 40423483-40423483 on Build GRCh38
rs888436810 Chr.19: 40423524-40423524 on Build GRCh38
rs268687 Chr.19: 40423456-40423456 on Build GRCh38
rs771161571 Chr.19: 40423519-40423519 on Build GRCh38
rs751409419 Chr.19: 40423476-40423476 on Build GRCh38
rs762251739 Chr.19: 40423217-40423217 on Build GRCh38
rs1006926950 Chr.19: 40423449-40423449 on Build GRCh38
rs763849190 Chr.19: 40423385-40423385 on Build GRCh38
rs760894157 Chr.19: 40423165-40423165 on Build GRCh38

View Full Product Details