Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42763760-42764274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG8
Entrez Gene ID
440533
Gene Name
pregnancy specific beta-1-glycoprotein 8
Gene Aliases
PSG1
UniGene
Hs.466843
Gene Chromosome Location
Chr.19: 42752687-42765692 on Build GRCh38
Transcripts
NM_001130167.1 NM_001130168.1 NM_182707.2 XM_011526986.1 XM_011526988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590110 c.308C>A p.S103Y Chr.19: 42764038-42764038 on Build GRCh38
3534946 c.306T>C p.Y102Y Chr.19: 42764040-42764040 on Build GRCh38
6288762 c.341A>G p.Q114R Chr.19: 42764005-42764005 on Build GRCh38
5933929 c.200G>A p.W67* Chr.19: 42764146-42764146 on Build GRCh38
5855993 c.234_235CA>TG p.I79V Chr.19: 42764111-42764112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398102313 Chr.19: 42763990-42763990 on Build GRCh38
rs772994342 Chr.19: 42764003-42764003 on Build GRCh38
rs770223171 Chr.19: 42764153-42764153 on Build GRCh38
rs150385493 Chr.19: 42763915-42763915 on Build GRCh38
rs747139040 Chr.19: 42764102-42764102 on Build GRCh38
rs114913961 Chr.19: 42764075-42764075 on Build GRCh38
rs767582675 Chr.19: 42763882-42763882 on Build GRCh38
rs142689447 Chr.19: 42764236-42764236 on Build GRCh38
rs1005433361 Chr.19: 42763972-42763972 on Build GRCh38
rs527880082 Chr.19: 42763938-42763938 on Build GRCh38
rs573917968 Chr.19: 42763834-42763834 on Build GRCh38
rs759484117 Chr.19: 42763979-42763979 on Build GRCh38
rs553890173 Chr.19: 42764034-42764034 on Build GRCh38
rs571272164 Chr.19: 42764188-42764188 on Build GRCh38
rs556036611 Chr.19: 42764235-42764235 on Build GRCh38
rs756202280 Chr.19: 42764242-42764242 on Build GRCh38
rs76352186 Chr.19: 42764090-42764090 on Build GRCh38
rs773915950 Chr.19: 42763872-42763872 on Build GRCh38
rs767156163 Chr.19: 42764077-42764077 on Build GRCh38
rs758194328 Chr.19: 42764183-42764183 on Build GRCh38
rs149961067 Chr.19: 42764045-42764045 on Build GRCh38
rs376789797 Chr.19: 42763926-42763926 on Build GRCh38
rs141265232 Chr.19: 42764093-42764093 on Build GRCh38
rs751580404 Chr.19: 42763937-42763937 on Build GRCh38
rs750586023 Chr.19: 42764070-42764070 on Build GRCh38
rs368446510 Chr.19: 42764072-42764072 on Build GRCh38
rs138853785 Chr.19: 42764172-42764172 on Build GRCh38
rs542565104 Chr.19: 42763853-42763853 on Build GRCh38
rs148036395 Chr.19: 42764217-42764217 on Build GRCh38
rs1049355172 Chr.19: 42764040-42764040 on Build GRCh38
rs770646877 Chr.19: 42763907-42763907 on Build GRCh38
rs75257969 Chr.19: 42764083-42764083 on Build GRCh38
rs768673447 Chr.19: 42763958-42763958 on Build GRCh38
rs767124095 Chr.19: 42764128-42764128 on Build GRCh38
rs750549965 Chr.19: 42763895-42763895 on Build GRCh38
rs770369152 Chr.19: 42764123-42764123 on Build GRCh38
rs201774677 Chr.19: 42764159-42764159 on Build GRCh38
rs77134863 Chr.19: 42764005-42764005 on Build GRCh38
rs781630576 Chr.19: 42763900-42763900 on Build GRCh38
rs572751671 Chr.19: 42764199-42764199 on Build GRCh38
rs761266960 Chr.19: 42764175-42764175 on Build GRCh38
rs753125487 Chr.19: 42763939-42763939 on Build GRCh38
rs138118337 Chr.19: 42764142-42764142 on Build GRCh38
rs201478462 Chr.19: 42763909-42763909 on Build GRCh38
rs79378816 Chr.19: 42764001-42764001 on Build GRCh38
rs768825721 Chr.19: 42764106-42764106 on Build GRCh38
rs146950952 Chr.19: 42764054-42764054 on Build GRCh38
rs755597560 Chr.19: 42764087-42764087 on Build GRCh38
rs771163131 Chr.19: 42764006-42764006 on Build GRCh38
rs367918383 Chr.19: 42764059-42764059 on Build GRCh38
rs751141317 Chr.19: 42764044-42764044 on Build GRCh38
rs560183732 Chr.19: 42763833-42763833 on Build GRCh38
rs780555542 Chr.19: 42764051-42764051 on Build GRCh38
rs768322602 Chr.19: 42764221-42764221 on Build GRCh38
rs906538567 Chr.19: 42763800-42763800 on Build GRCh38
rs756187432 Chr.19: 42764204-42764204 on Build GRCh38
rs761475929 Chr.19: 42763924-42763924 on Build GRCh38
rs938657320 Chr.19: 42764085-42764085 on Build GRCh38
rs762961901 Chr.19: 42764232-42764232 on Build GRCh38
rs776491877 Chr.19: 42763922-42763922 on Build GRCh38
rs141872267 Chr.19: 42763969-42763969 on Build GRCh38
rs190921232 Chr.19: 42763990-42763990 on Build GRCh38
rs749822114 Chr.19: 42764213-42764213 on Build GRCh38
rs781513768 Chr.19: 42764203-42764203 on Build GRCh38
rs768915559 Chr.19: 42764145-42764145 on Build GRCh38
rs756565747 Chr.19: 42763985-42763985 on Build GRCh38
rs781232973 Chr.19: 42763998-42763998 on Build GRCh38
rs775439993 Chr.19: 42764061-42764061 on Build GRCh38
rs7255518 Chr.19: 42764099-42764099 on Build GRCh38
rs778663769 Chr.19: 42763946-42763946 on Build GRCh38
rs750621396 Chr.19: 42763929-42763929 on Build GRCh38
rs7255419 Chr.19: 42764100-42764100 on Build GRCh38
rs757110395 Chr.19: 42763886-42763886 on Build GRCh38
rs775542496 Chr.19: 42763883-42763883 on Build GRCh38
rs764803115 Chr.19: 42764228-42764228 on Build GRCh38
rs142736244 Chr.19: 42764053-42764053 on Build GRCh38
rs774618783 Chr.19: 42764057-42764057 on Build GRCh38
rs141761918 Chr.19: 42764219-42764219 on Build GRCh38
rs148091598 Chr.19: 42763967-42763967 on Build GRCh38
rs778209745 Chr.19: 42764208-42764208 on Build GRCh38
rs757187752 Chr.19: 42764135-42764135 on Build GRCh38
rs765140265 Chr.19: 42764176-42764176 on Build GRCh38
rs760330897 Chr.19: 42764166-42764166 on Build GRCh38
rs763266798 Chr.19: 42764157-42764157 on Build GRCh38
rs778872603 Chr.19: 42764136-42764136 on Build GRCh38
rs763061504 Chr.19: 42763931-42763931 on Build GRCh38
rs779728700 Chr.19: 42763956-42763956 on Build GRCh38
rs773081290 Chr.19: 42764229-42764229 on Build GRCh38
rs770678423 Chr.19: 42764056-42764056 on Build GRCh38
rs746456815 Chr.19: 42763957-42763957 on Build GRCh38
rs570388501 Chr.19: 42764150-42764150 on Build GRCh38
rs779816799 Chr.19: 42763993-42763993 on Build GRCh38
rs756254977 Chr.19: 42764081-42764084 on Build GRCh38
rs950852259 Chr.19: 42763839-42763839 on Build GRCh38
rs777344894 Chr.19: 42764108-42764108 on Build GRCh38
rs551436858 Chr.19: 42764178-42764178 on Build GRCh38
rs773480083 Chr.19: 42764124-42764124 on Build GRCh38
rs373148587 Chr.19: 42764067-42764067 on Build GRCh38
rs765313788 Chr.19: 42763894-42763894 on Build GRCh38
rs749785146 Chr.19: 42764213-42764213 on Build GRCh38
rs768115204 Chr.19: 42763982-42763982 on Build GRCh38
rs945498790 Chr.19: 42763787-42763787 on Build GRCh38
rs116767543 Chr.19: 42763941-42763941 on Build GRCh38
rs11355507 Chr.19: 42763989-42763989 on Build GRCh38
rs147359969 Chr.19: 42764243-42764243 on Build GRCh38
rs771673487 Chr.19: 42763911-42763911 on Build GRCh38
rs770429833 Chr.19: 42763870-42763870 on Build GRCh38
rs751670103 Chr.19: 42763976-42763976 on Build GRCh38
rs372958378 Chr.19: 42763921-42763921 on Build GRCh38
rs766184721 Chr.19: 42764035-42764035 on Build GRCh38
rs753285468 Chr.19: 42764131-42764131 on Build GRCh38
rs747483891 Chr.19: 42763897-42763897 on Build GRCh38
rs778265919 Chr.19: 42763986-42763986 on Build GRCh38
rs764632234 Chr.19: 42764033-42764033 on Build GRCh38
rs562874456 Chr.19: 42763862-42763862 on Build GRCh38
rs757517518 Chr.19: 42763892-42763892 on Build GRCh38
rs763949093 Chr.19: 42763887-42763887 on Build GRCh38
rs762398728 Chr.19: 42764122-42764122 on Build GRCh38
rs370490968 Chr.19: 42764027-42764027 on Build GRCh38
rs577892665 Chr.19: 42763819-42763819 on Build GRCh38
rs748032206 Chr.19: 42763999-42763999 on Build GRCh38
rs74660622 Chr.19: 42764196-42764196 on Build GRCh38
rs765694653 Chr.19: 42764070-42764070 on Build GRCh38
rs753766127 Chr.19: 42764091-42764091 on Build GRCh38
rs895132510 Chr.19: 42763961-42763961 on Build GRCh38
rs7256456 Chr.19: 42764064-42764064 on Build GRCh38
rs1015726719 Chr.19: 42763852-42763853 on Build GRCh38
rs758194763 Chr.19: 42764138-42764138 on Build GRCh38
rs150588080 Chr.19: 42764225-42764225 on Build GRCh38
rs761102569 Chr.19: 42764032-42764032 on Build GRCh38
rs75687580 Chr.19: 42764233-42764233 on Build GRCh38
rs7255164 Chr.19: 42763917-42763917 on Build GRCh38
rs753108890 Chr.19: 42763984-42763984 on Build GRCh38
rs748391978 Chr.19: 42763901-42763901 on Build GRCh38
rs1027851771 Chr.19: 42763837-42763837 on Build GRCh38
rs1042864066 Chr.19: 42763790-42763790 on Build GRCh38
rs760771531 Chr.19: 42764062-42764062 on Build GRCh38
rs146093517 Chr.19: 42763970-42763970 on Build GRCh38
rs1003919458 Chr.19: 42763807-42763807 on Build GRCh38
rs115679481 Chr.19: 42764007-42764007 on Build GRCh38
rs752363136 Chr.19: 42764130-42764130 on Build GRCh38
rs531614033 Chr.19: 42763889-42763889 on Build GRCh38
rs771769482 Chr.19: 42763954-42763954 on Build GRCh38
rs149135211 Chr.19: 42764148-42764148 on Build GRCh38
rs763326638 Chr.19: 42764127-42764127 on Build GRCh38
rs750210420 Chr.19: 42764137-42764137 on Build GRCh38
rs746061706 Chr.19: 42764058-42764058 on Build GRCh38
rs773746520 Chr.19: 42764156-42764156 on Build GRCh38
rs746544244 Chr.19: 42763997-42763997 on Build GRCh38
rs369659318 Chr.19: 42763951-42763951 on Build GRCh38
rs982590256 Chr.19: 42763851-42763851 on Build GRCh38
rs370832692 Chr.19: 42764250-42764250 on Build GRCh38
rs547950171 Chr.19: 42764142-42764142 on Build GRCh38
rs375085910 Chr.19: 42763896-42763896 on Build GRCh38
rs766432917 Chr.19: 42763934-42763934 on Build GRCh38
rs371799367 Chr.19: 42763874-42763874 on Build GRCh38
rs540460277 Chr.19: 42763832-42763832 on Build GRCh38
rs138900265 Chr.19: 42763974-42763974 on Build GRCh38
rs745417777 Chr.19: 42763910-42763910 on Build GRCh38
rs372792521 Chr.19: 42763846-42763846 on Build GRCh38

View Full Product Details