Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44274362-44274874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF233
Entrez Gene ID
353355
Gene Name
zinc finger protein 233
Gene Aliases
-
UniGene
Hs.466891
Gene Chromosome Location
Chr.19: 44259880-44275317 on Build GRCh38
Transcripts
NM_001207005.1 NM_181756.2 XM_017026758.1 XM_017026759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644222 c.1795G>T p.E599* Chr.19: 44274509-44274509 on Build GRCh38
5686576 c.1860T>A p.H620Q Chr.19: 44274574-44274574 on Build GRCh38
1721051 c.1967G>A p.G656D Chr.19: 44274627-44274627 on Build GRCh38
3535503 c.1939G>A p.D647N Chr.19: 44274653-44274653 on Build GRCh38
6024483 c.1885_1886insT p.S630fs*26 Chr.19: 44274545-44274546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779905529 Chr.19: 44274699-44274699 on Build GRCh38
rs1009398833 Chr.19: 44274459-44274459 on Build GRCh38
rs763181841 Chr.19: 44274727-44274727 on Build GRCh38
rs765488975 Chr.19: 44274504-44274504 on Build GRCh38
rs373216791 Chr.19: 44274707-44274707 on Build GRCh38
rs950652646 Chr.19: 44274534-44274534 on Build GRCh38
rs768132633 Chr.19: 44274607-44274607 on Build GRCh38
rs776107423 Chr.19: 44274714-44274714 on Build GRCh38
rs767558704 Chr.19: 44274642-44274643 on Build GRCh38
rs7256897 Chr.19: 44274717-44274717 on Build GRCh38
rs752172200 Chr.19: 44274708-44274708 on Build GRCh38
rs183401131 Chr.19: 44274831-44274831 on Build GRCh38
rs867943193 Chr.19: 44274552-44274552 on Build GRCh38
rs759840941 Chr.19: 44274621-44274632 on Build GRCh38
rs758546863 Chr.19: 44274393-44274393 on Build GRCh38
rs776284890 Chr.19: 44274596-44274596 on Build GRCh38
rs772069307 Chr.19: 44274687-44274687 on Build GRCh38
rs762839651 Chr.19: 44274644-44274644 on Build GRCh38
rs1024568460 Chr.19: 44274616-44274616 on Build GRCh38
rs200295413 Chr.19: 44274519-44274519 on Build GRCh38
rs767620287 Chr.19: 44274646-44274646 on Build GRCh38
rs750697604 Chr.19: 44274622-44274622 on Build GRCh38
rs997234652 Chr.19: 44274511-44274511 on Build GRCh38
rs746919413 Chr.19: 44274703-44274703 on Build GRCh38
rs1047474794 Chr.19: 44274758-44274758 on Build GRCh38
rs1032372828 Chr.19: 44274541-44274541 on Build GRCh38
rs1008393644 Chr.19: 44274774-44274774 on Build GRCh38
rs2884016 Chr.19: 44274646-44274646 on Build GRCh38
rs760128534 Chr.19: 44274602-44274602 on Build GRCh38
rs757703669 Chr.19: 44274720-44274721 on Build GRCh38
rs745829504 Chr.19: 44274588-44274588 on Build GRCh38
rs199781533 Chr.19: 44274604-44274604 on Build GRCh38
rs867961626 Chr.19: 44274635-44274635 on Build GRCh38
rs767111168 Chr.19: 44274454-44274454 on Build GRCh38
rs781609934 Chr.19: 44274542-44274542 on Build GRCh38
rs776306581 Chr.19: 44274612-44274612 on Build GRCh38
rs751008846 Chr.19: 44274505-44274505 on Build GRCh38
rs386809645 Chr.19: 44274643-44274646 on Build GRCh38
rs75921463 Chr.19: 44274643-44274643 on Build GRCh38
rs753024279 Chr.19: 44274653-44274653 on Build GRCh38
rs764699921 Chr.19: 44274702-44274703 on Build GRCh38
rs758821581 Chr.19: 44274525-44274525 on Build GRCh38
rs928975040 Chr.19: 44274745-44274745 on Build GRCh38
rs367738951 Chr.19: 44274494-44274494 on Build GRCh38
rs768538023 Chr.19: 44274385-44274385 on Build GRCh38
rs148499228 Chr.19: 44274402-44274402 on Build GRCh38
rs766252720 Chr.19: 44274576-44274577 on Build GRCh38
rs746339529 Chr.19: 44274391-44274391 on Build GRCh38
rs749056334 Chr.19: 44274436-44274436 on Build GRCh38
rs774382974 Chr.19: 44274449-44274449 on Build GRCh38
rs1041240233 Chr.19: 44274816-44274816 on Build GRCh38
rs926532142 Chr.19: 44274697-44274697 on Build GRCh38
rs1047527928 Chr.19: 44274442-44274442 on Build GRCh38
rs202091669 Chr.19: 44274528-44274528 on Build GRCh38
rs747316136 Chr.19: 44274400-44274400 on Build GRCh38
rs779995466 Chr.19: 44274394-44274394 on Build GRCh38
rs59660444 Chr.19: 44274643-44274643 on Build GRCh38
rs796944595 Chr.19: 44274390-44274390 on Build GRCh38
rs912064350 Chr.19: 44274766-44274766 on Build GRCh38
rs756231013 Chr.19: 44274655-44274655 on Build GRCh38
rs868442545 Chr.19: 44274642-44274643 on Build GRCh38
rs763695399 Chr.19: 44274483-44274483 on Build GRCh38
rs935659515 Chr.19: 44274386-44274386 on Build GRCh38
rs781653514 Chr.19: 44274727-44274728 on Build GRCh38
rs140083467 Chr.19: 44274477-44274477 on Build GRCh38
rs76164141 Chr.19: 44274585-44274585 on Build GRCh38
rs780198618 Chr.19: 44274532-44274532 on Build GRCh38
rs192675509 Chr.19: 44274387-44274387 on Build GRCh38
rs761290468 Chr.19: 44274782-44274782 on Build GRCh38
rs141851681 Chr.19: 44274452-44274452 on Build GRCh38
rs1042604766 Chr.19: 44274763-44274763 on Build GRCh38
rs945627970 Chr.19: 44274406-44274406 on Build GRCh38
rs770823324 Chr.19: 44274440-44274440 on Build GRCh38
rs386809644 Chr.19: 44274643-44274644 on Build GRCh38
rs760475674 Chr.19: 44274408-44274409 on Build GRCh38
rs1042565740 Chr.19: 44274425-44274425 on Build GRCh38
rs76961325 Chr.19: 44274762-44274762 on Build GRCh38
rs752842706 Chr.19: 44274644-44274645 on Build GRCh38
rs371654157 Chr.19: 44274619-44274619 on Build GRCh38
rs375321862 Chr.19: 44274631-44274631 on Build GRCh38
rs146078644 Chr.19: 44274475-44274475 on Build GRCh38
rs755286316 Chr.19: 44274533-44274533 on Build GRCh38
rs773148336 Chr.19: 44274431-44274431 on Build GRCh38
rs761470775 Chr.19: 44274715-44274715 on Build GRCh38
rs761452350 Chr.19: 44274615-44274615 on Build GRCh38
rs200527384 Chr.19: 44274591-44274591 on Build GRCh38
rs2884015 Chr.19: 44274644-44274644 on Build GRCh38
rs758509166 Chr.19: 44274694-44274696 on Build GRCh38
rs377690459 Chr.19: 44274660-44274660 on Build GRCh38
rs768222818 Chr.19: 44274712-44274712 on Build GRCh38
rs775095585 Chr.19: 44274599-44274599 on Build GRCh38
rs754134732 Chr.19: 44274384-44274384 on Build GRCh38
rs748345426 Chr.19: 44274545-44274545 on Build GRCh38
rs745608564 Chr.19: 44274674-44274674 on Build GRCh38
rs76295615 Chr.19: 44274645-44274645 on Build GRCh38
rs770913114 Chr.19: 44274550-44274550 on Build GRCh38
rs957098359 Chr.19: 44274695-44274695 on Build GRCh38

View Full Product Details