Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44758106-44758636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL3
Entrez Gene ID
602
Gene Name
B-cell CLL/lymphoma 3
Gene Aliases
BCL4, D19S37
UniGene
Hs.31210
Gene Chromosome Location
Chr.19: 44742621-44760044 on Build GRCh38
Transcripts
NM_005178.4 XM_011527197.2 XM_011527198.2 XM_011527200.2 XM_017027109.1 XM_017027110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5952108 c.1009C>T p.L337F Chr.19: 44758363-44758363 on Build GRCh38
5787932 c.911C>T p.A304V Chr.19: 44758265-44758265 on Build GRCh38
4886930 c.1012A>C p.K338Q Chr.19: 44758366-44758366 on Build GRCh38
7327978 c.1002C>T p.D334D Chr.19: 44758356-44758356 on Build GRCh38
6379769 c.921C>A p.Y307* Chr.19: 44758275-44758275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2306148 Chr.19: 44758174-44758174 on Build GRCh38
rs568986481 Chr.19: 44758208-44758208 on Build GRCh38
rs373579508 Chr.19: 44758488-44758488 on Build GRCh38
rs374794948 Chr.19: 44758552-44758552 on Build GRCh38
rs958682100 Chr.19: 44758421-44758421 on Build GRCh38
rs932053725 Chr.19: 44758518-44758518 on Build GRCh38
rs922016661 Chr.19: 44758305-44758305 on Build GRCh38
rs947306162 Chr.19: 44758589-44758589 on Build GRCh38
rs781540511 Chr.19: 44758302-44758302 on Build GRCh38
rs370500131 Chr.19: 44758462-44758462 on Build GRCh38
rs1025217932 Chr.19: 44758198-44758198 on Build GRCh38
rs894982538 Chr.19: 44758172-44758172 on Build GRCh38
rs539810994 Chr.19: 44758209-44758210 on Build GRCh38
rs923400771 Chr.19: 44758499-44758499 on Build GRCh38
rs1012404250 Chr.19: 44758538-44758538 on Build GRCh38
rs754245032 Chr.19: 44758239-44758239 on Build GRCh38
rs778783165 Chr.19: 44758335-44758335 on Build GRCh38
rs34585458 Chr.19: 44758315-44758315 on Build GRCh38
rs912055738 Chr.19: 44758434-44758434 on Build GRCh38
rs777776029 Chr.19: 44758201-44758201 on Build GRCh38
rs536904635 Chr.19: 44758570-44758570 on Build GRCh38
rs760204437 Chr.19: 44758410-44758410 on Build GRCh38
rs1050458856 Chr.19: 44758563-44758563 on Build GRCh38
rs771584240 Chr.19: 44758420-44758420 on Build GRCh38
rs770880267 Chr.19: 44758318-44758318 on Build GRCh38
rs780052702 Chr.19: 44758276-44758276 on Build GRCh38
rs34967149 Chr.19: 44758448-44758448 on Build GRCh38
rs768349474 Chr.19: 44758446-44758446 on Build GRCh38
rs894925970 Chr.19: 44758528-44758528 on Build GRCh38
rs34401216 Chr.19: 44758263-44758263 on Build GRCh38
rs766876091 Chr.19: 44758504-44758504 on Build GRCh38
rs893088899 Chr.19: 44758138-44758138 on Build GRCh38
rs1026698864 Chr.19: 44758168-44758168 on Build GRCh38
rs995256226 Chr.19: 44758157-44758157 on Build GRCh38
rs987686260 Chr.19: 44758424-44758424 on Build GRCh38
rs745806788 Chr.19: 44758392-44758392 on Build GRCh38
rs952336913 Chr.19: 44758454-44758454 on Build GRCh38
rs896820529 Chr.19: 44758135-44758135 on Build GRCh38
rs771039737 Chr.19: 44758210-44758210 on Build GRCh38
rs1048634087 Chr.19: 44758522-44758522 on Build GRCh38
rs774450925 Chr.19: 44758213-44758213 on Build GRCh38
rs778852971 Chr.19: 44758236-44758236 on Build GRCh38
rs199807623 Chr.19: 44758312-44758312 on Build GRCh38
rs1012107382 Chr.19: 44758176-44758176 on Build GRCh38
rs372815971 Chr.19: 44758266-44758266 on Build GRCh38
rs765595926 Chr.19: 44758471-44758471 on Build GRCh38
rs991209952 Chr.19: 44758432-44758432 on Build GRCh38
rs755193601 Chr.19: 44758289-44758289 on Build GRCh38
rs143644908 Chr.19: 44758409-44758409 on Build GRCh38
rs35844547 Chr.19: 44758209-44758209 on Build GRCh38
rs773609630 Chr.19: 44758495-44758495 on Build GRCh38
rs147211387 Chr.19: 44758498-44758498 on Build GRCh38
rs761419662 Chr.19: 44758450-44758450 on Build GRCh38
rs2306147 Chr.19: 44758220-44758220 on Build GRCh38
rs571056210 Chr.19: 44758277-44758277 on Build GRCh38
rs748330939 Chr.19: 44758310-44758310 on Build GRCh38
rs760305515 Chr.19: 44758237-44758237 on Build GRCh38
rs763734130 Chr.19: 44758238-44758238 on Build GRCh38
rs953569437 Chr.19: 44758320-44758320 on Build GRCh38
rs758878538 Chr.19: 44758269-44758269 on Build GRCh38

View Full Product Details