Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50361571-50362064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPSA
Entrez Gene ID
9476
Gene Name
napsin A aspartic peptidase
Gene Aliases
KAP, Kdap, NAP1, NAPA, SNAPA
UniGene
Hs.512843
Gene Chromosome Location
Chr.19: 50358472-50369331 on Build GRCh38
Transcripts
NM_004851.2 XM_011527540.1 XM_017027512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6386506 c.277G>T p.V93F Chr.19: 50362041-50362041 on Build GRCh38
5056854 c.327C>T p.C109C Chr.19: 50361991-50361991 on Build GRCh38
713310 c.306C>G p.L102L Chr.19: 50362012-50362012 on Build GRCh38
4080351 c.434G>A p.R145Q Chr.19: 50361697-50361697 on Build GRCh38
6381193 c.347G>T p.C116F Chr.19: 50361971-50361971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780917700 Chr.19: 50361936-50361936 on Build GRCh38
rs752954713 Chr.19: 50361647-50361647 on Build GRCh38
rs750429905 Chr.19: 50361806-50361806 on Build GRCh38
rs745914617 Chr.19: 50361722-50361722 on Build GRCh38
rs878987940 Chr.19: 50361935-50361935 on Build GRCh38
rs898399448 Chr.19: 50361769-50361769 on Build GRCh38
rs757657519 Chr.19: 50361958-50361958 on Build GRCh38
rs371514076 Chr.19: 50361797-50361797 on Build GRCh38
rs757962918 Chr.19: 50361809-50361809 on Build GRCh38
rs376501854 Chr.19: 50361658-50361658 on Build GRCh38
rs750981539 Chr.19: 50362042-50362042 on Build GRCh38
rs988735982 Chr.19: 50361855-50361855 on Build GRCh38
rs1032048298 Chr.19: 50361927-50361927 on Build GRCh38
rs765176103 Chr.19: 50362025-50362025 on Build GRCh38
rs970071402 Chr.19: 50361746-50361746 on Build GRCh38
rs779368805 Chr.19: 50361969-50361969 on Build GRCh38
rs775766641 Chr.19: 50361781-50361781 on Build GRCh38
rs990364436 Chr.19: 50361980-50361980 on Build GRCh38
rs993722721 Chr.19: 50361592-50361592 on Build GRCh38
rs374073747 Chr.19: 50361813-50361813 on Build GRCh38
rs957600557 Chr.19: 50361957-50361957 on Build GRCh38
rs370440200 Chr.19: 50361802-50361802 on Build GRCh38
rs746431183 Chr.19: 50361973-50361973 on Build GRCh38
rs767684648 Chr.19: 50361644-50361644 on Build GRCh38
rs749672946 Chr.19: 50361701-50361701 on Build GRCh38
rs536594316 Chr.19: 50361892-50361892 on Build GRCh38
rs768712881 Chr.19: 50362004-50362004 on Build GRCh38
rs367651287 Chr.19: 50362003-50362003 on Build GRCh38
rs775366354 Chr.19: 50362001-50362001 on Build GRCh38
rs754526185 Chr.19: 50361827-50361827 on Build GRCh38
rs902290832 Chr.19: 50361917-50361917 on Build GRCh38
rs749905064 Chr.19: 50361677-50361677 on Build GRCh38
rs566480672 Chr.19: 50361978-50361978 on Build GRCh38
rs763054627 Chr.19: 50362032-50362032 on Build GRCh38
rs532220200 Chr.19: 50361784-50361784 on Build GRCh38
rs751081503 Chr.19: 50361821-50361821 on Build GRCh38
rs761672934 Chr.19: 50362024-50362024 on Build GRCh38
rs138101768 Chr.19: 50361662-50361662 on Build GRCh38
rs186910155 Chr.19: 50361698-50361698 on Build GRCh38
rs749574007 Chr.19: 50361956-50361956 on Build GRCh38
rs201994232 Chr.19: 50361712-50361712 on Build GRCh38
rs773392864 Chr.19: 50361678-50361678 on Build GRCh38
rs999581053 Chr.19: 50361918-50361918 on Build GRCh38
rs1035713106 Chr.19: 50361786-50361786 on Build GRCh38
rs191752567 Chr.19: 50361870-50361870 on Build GRCh38
rs554866074 Chr.19: 50361913-50361913 on Build GRCh38
rs760494965 Chr.19: 50361650-50361650 on Build GRCh38
rs533744414 Chr.19: 50362028-50362028 on Build GRCh38
rs368961358 Chr.19: 50361690-50361690 on Build GRCh38
rs1019755795 Chr.19: 50361837-50361837 on Build GRCh38
rs34576633 Chr.19: 50361877-50361878 on Build GRCh38
rs759631878 Chr.19: 50361630-50361630 on Build GRCh38
rs968997598 Chr.19: 50361988-50361988 on Build GRCh38
rs376308207 Chr.19: 50361800-50361800 on Build GRCh38
rs765280362 Chr.19: 50361674-50361674 on Build GRCh38
rs772187022 Chr.19: 50361770-50361770 on Build GRCh38
rs1002898790 Chr.19: 50361754-50361754 on Build GRCh38
rs746693112 Chr.19: 50361697-50361697 on Build GRCh38
rs774353721 Chr.19: 50361629-50361629 on Build GRCh38
rs776231658 Chr.19: 50361801-50361801 on Build GRCh38
rs776944945 Chr.19: 50362022-50362022 on Build GRCh38
rs771375487 Chr.19: 50361704-50361704 on Build GRCh38

View Full Product Details