Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55761661-55762177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFPL4A
Entrez Gene ID
342931
Gene Name
ret finger protein like 4A
Gene Aliases
RFPL4, RNF210
UniGene
Hs.631553
Gene Chromosome Location
Chr.19: 55757298-55763388 on Build GRCh38
Transcripts
NM_001145014.1 XM_011526915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963663 c.286+10C>A p.? Chr.19: 55762096-55762096 on Build GRCh38
5785057 c.183C>G p.I61M Chr.19: 55761983-55761983 on Build GRCh38
7231990 c.277A>G p.K93E Chr.19: 55762077-55762077 on Build GRCh38
148753 c.127G>A p.D43N Chr.19: 55761927-55761927 on Build GRCh38
5892394 c.52C>T p.L18F Chr.19: 55761852-55761852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952500155 Chr.19: 55762020-55762020 on Build GRCh38
rs371547624 Chr.19: 55761723-55761723 on Build GRCh38
rs778020941 Chr.19: 55761778-55761778 on Build GRCh38
rs550132468 Chr.19: 55761948-55761948 on Build GRCh38
rs908011055 Chr.19: 55761698-55761698 on Build GRCh38
rs9710441 Chr.19: 55761910-55761910 on Build GRCh38
rs780789451 Chr.19: 55761978-55761978 on Build GRCh38
rs765688290 Chr.19: 55762108-55762108 on Build GRCh38
rs751235715 Chr.19: 55761706-55761706 on Build GRCh38
rs918100969 Chr.19: 55762122-55762122 on Build GRCh38
rs62126990 Chr.19: 55761789-55761789 on Build GRCh38
rs111497074 Chr.19: 55761744-55761744 on Build GRCh38
rs372205326 Chr.19: 55762103-55762103 on Build GRCh38
rs2020385 Chr.19: 55761770-55761770 on Build GRCh38
rs762725015 Chr.19: 55761775-55761775 on Build GRCh38
rs756932030 Chr.19: 55761949-55761949 on Build GRCh38
rs559641281 Chr.19: 55761771-55761771 on Build GRCh38
rs1045039182 Chr.19: 55761863-55761863 on Build GRCh38
rs767807828 Chr.19: 55761864-55761864 on Build GRCh38
rs772473069 Chr.19: 55762098-55762098 on Build GRCh38
rs528943290 Chr.19: 55762006-55762006 on Build GRCh38
rs760108267 Chr.19: 55762104-55762104 on Build GRCh38
rs1056683290 Chr.19: 55761785-55761785 on Build GRCh38
rs780702048 Chr.19: 55761777-55761778 on Build GRCh38
rs537368511 Chr.19: 55761715-55761715 on Build GRCh38
rs903865593 Chr.19: 55761876-55761876 on Build GRCh38
rs766483880 Chr.19: 55762100-55762100 on Build GRCh38
rs1019444913 Chr.19: 55761925-55761925 on Build GRCh38
rs768430378 Chr.19: 55762074-55762074 on Build GRCh38
rs745615757 Chr.19: 55761924-55761924 on Build GRCh38
rs956676544 Chr.19: 55761907-55761907 on Build GRCh38
rs761671155 Chr.19: 55762094-55762094 on Build GRCh38
rs774319697 Chr.19: 55762092-55762092 on Build GRCh38
rs866155671 Chr.19: 55762007-55762007 on Build GRCh38
rs948090325 Chr.19: 55762123-55762123 on Build GRCh38
rs143775947 Chr.19: 55761707-55761707 on Build GRCh38
rs755937277 Chr.19: 55762058-55762061 on Build GRCh38
rs1000767045 Chr.19: 55761898-55761898 on Build GRCh38
rs200422093 Chr.19: 55761919-55761919 on Build GRCh38
rs940819190 Chr.19: 55761709-55761709 on Build GRCh38
rs573733109 Chr.19: 55761754-55761754 on Build GRCh38
rs556917186 Chr.19: 55761729-55761729 on Build GRCh38
rs867378181 Chr.19: 55761834-55761834 on Build GRCh38
rs1031318812 Chr.19: 55761902-55761902 on Build GRCh38
rs919571428 Chr.19: 55761992-55761992 on Build GRCh38
rs773553386 Chr.19: 55762099-55762099 on Build GRCh38
rs2019946 Chr.19: 55761927-55761927 on Build GRCh38
rs202117713 Chr.19: 55761843-55761843 on Build GRCh38
rs548963378 Chr.19: 55762061-55762061 on Build GRCh38
rs533241266 Chr.19: 55761936-55761936 on Build GRCh38
rs982490611 Chr.19: 55762045-55762045 on Build GRCh38
rs371808615 Chr.19: 55761884-55761884 on Build GRCh38
rs878991567 Chr.19: 55761778-55761778 on Build GRCh38
rs750573187 Chr.19: 55761889-55761889 on Build GRCh38
rs753097729 Chr.19: 55762111-55762111 on Build GRCh38
rs908113432 Chr.19: 55762084-55762084 on Build GRCh38
rs375134584 Chr.19: 55761692-55761692 on Build GRCh38
rs1833784 Chr.19: 55762136-55762136 on Build GRCh38
rs963725104 Chr.19: 55761926-55761926 on Build GRCh38
rs774233991 Chr.19: 55761836-55761836 on Build GRCh38
rs749265868 Chr.19: 55762046-55762046 on Build GRCh38
rs202102526 Chr.19: 55761991-55761991 on Build GRCh38
rs754364089 Chr.19: 55761928-55761928 on Build GRCh38
rs192959272 Chr.19: 55761700-55761700 on Build GRCh38
rs1049789243 Chr.19: 55761687-55761687 on Build GRCh38
rs1960227 Chr.19: 55762138-55762138 on Build GRCh38
rs896571770 Chr.19: 55761802-55761802 on Build GRCh38
rs755373402 Chr.19: 55761930-55761930 on Build GRCh38
rs540829471 Chr.19: 55761906-55761906 on Build GRCh38
rs1056608651 Chr.19: 55762107-55762107 on Build GRCh38

View Full Product Details