Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57473562-57474061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF772
Entrez Gene ID
400720
Gene Name
zinc finger protein 772
Gene Aliases
-
UniGene
Hs.388810
Gene Chromosome Location
Chr.19: 57464198-57480950 on Build GRCh38
Transcripts
NM_001024596.2 NM_001144068.1 XM_005258943.4 XM_005258944.3 XM_011526980.2 XM_017026813.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397050 c.828C>T p.S276S Chr.19: 57473916-57473916 on Build GRCh38
7274751 c.1061A>G p.E354G Chr.19: 57473683-57473683 on Build GRCh38
4082080 c.857G>T p.R286I Chr.19: 57473887-57473887 on Build GRCh38
5461731 c.857G>A p.R286K Chr.19: 57473887-57473887 on Build GRCh38
5476129 c.1035T>C p.T345T Chr.19: 57473709-57473709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922282395 Chr.19: 57473677-57473677 on Build GRCh38
rs752515178 Chr.19: 57473658-57473658 on Build GRCh38
rs755008166 Chr.19: 57473841-57473841 on Build GRCh38
rs758733839 Chr.19: 57474017-57474017 on Build GRCh38
rs755409273 Chr.19: 57473725-57473725 on Build GRCh38
rs747487635 Chr.19: 57473883-57473883 on Build GRCh38
rs140411377 Chr.19: 57473856-57473856 on Build GRCh38
rs888709451 Chr.19: 57473859-57473859 on Build GRCh38
rs374680975 Chr.19: 57473736-57473736 on Build GRCh38
rs187566501 Chr.19: 57473831-57473831 on Build GRCh38
rs866774588 Chr.19: 57473785-57473785 on Build GRCh38
rs767521942 Chr.19: 57473914-57473914 on Build GRCh38
rs368900492 Chr.19: 57473746-57473746 on Build GRCh38
rs932994108 Chr.19: 57473589-57473589 on Build GRCh38
rs749976170 Chr.19: 57473821-57473821 on Build GRCh38
rs773280638 Chr.19: 57473751-57473751 on Build GRCh38
rs572622320 Chr.19: 57473890-57473890 on Build GRCh38
rs372201369 Chr.19: 57473783-57473783 on Build GRCh38
rs964233006 Chr.19: 57473922-57473922 on Build GRCh38
rs762683065 Chr.19: 57473661-57473661 on Build GRCh38
rs781098749 Chr.19: 57473855-57473855 on Build GRCh38
rs746170834 Chr.19: 57473894-57473894 on Build GRCh38
rs774013917 Chr.19: 57473797-57473797 on Build GRCh38
rs777572079 Chr.19: 57473918-57473918 on Build GRCh38
rs1018889032 Chr.19: 57473636-57473636 on Build GRCh38
rs1006259269 Chr.19: 57473902-57473902 on Build GRCh38
rs183964084 Chr.19: 57473641-57473641 on Build GRCh38
rs570623193 Chr.19: 57473686-57473686 on Build GRCh38
rs1011020782 Chr.19: 57473991-57473991 on Build GRCh38
rs1030103592 Chr.19: 57473998-57473998 on Build GRCh38
rs964311230 Chr.19: 57473639-57473639 on Build GRCh38
rs200221090 Chr.19: 57473629-57473629 on Build GRCh38
rs527708615 Chr.19: 57473601-57473601 on Build GRCh38
rs758953637 Chr.19: 57473865-57473865 on Build GRCh38
rs1018440587 Chr.19: 57474005-57474005 on Build GRCh38
rs557742897 Chr.19: 57474008-57474008 on Build GRCh38
rs757512427 Chr.19: 57473922-57473922 on Build GRCh38
rs747408352 Chr.19: 57473807-57473807 on Build GRCh38
rs753123599 Chr.19: 57474014-57474014 on Build GRCh38
rs755832086 Chr.19: 57473703-57473703 on Build GRCh38
rs145072663 Chr.19: 57474022-57474022 on Build GRCh38
rs1018522498 Chr.19: 57473904-57473904 on Build GRCh38
rs138230060 Chr.19: 57473908-57473908 on Build GRCh38
rs778217342 Chr.19: 57473881-57473881 on Build GRCh38
rs771177418 Chr.19: 57473775-57473775 on Build GRCh38
rs760948274 Chr.19: 57473653-57473653 on Build GRCh38
rs780579406 Chr.19: 57473605-57473605 on Build GRCh38
rs748391629 Chr.19: 57473860-57473860 on Build GRCh38
rs201303546 Chr.19: 57473799-57473799 on Build GRCh38
rs951056416 Chr.19: 57473680-57473680 on Build GRCh38
rs879100152 Chr.19: 57473909-57473909 on Build GRCh38
rs866252721 Chr.19: 57473738-57473738 on Build GRCh38
rs150415935 Chr.19: 57473714-57473714 on Build GRCh38
rs757135864 Chr.19: 57473687-57473687 on Build GRCh38
rs149967182 Chr.19: 57473941-57473941 on Build GRCh38
rs200670804 Chr.19: 57473820-57473820 on Build GRCh38
rs199665856 Chr.19: 57473830-57473830 on Build GRCh38
rs780811501 Chr.19: 57473691-57473691 on Build GRCh38
rs759735765 Chr.19: 57473781-57473781 on Build GRCh38
rs142150770 Chr.19: 57473600-57473600 on Build GRCh38
rs893883901 Chr.19: 57473949-57473949 on Build GRCh38
rs764735045 Chr.19: 57474009-57474009 on Build GRCh38
rs771050689 Chr.19: 57473886-57473886 on Build GRCh38
rs769318358 Chr.19: 57473613-57473613 on Build GRCh38
rs747029352 Chr.19: 57473768-57473768 on Build GRCh38
rs184861513 Chr.19: 57474029-57474029 on Build GRCh38
rs769842236 Chr.19: 57473900-57473900 on Build GRCh38
rs757759792 Chr.19: 57474030-57474030 on Build GRCh38
rs761466365 Chr.19: 57473912-57473912 on Build GRCh38
rs749597340 Chr.19: 57473607-57473607 on Build GRCh38
rs749894543 Chr.19: 57473693-57473693 on Build GRCh38
rs200429804 Chr.19: 57473654-57473654 on Build GRCh38
rs748962139 Chr.19: 57473959-57473959 on Build GRCh38
rs375469604 Chr.19: 57473915-57473915 on Build GRCh38
rs772236853 Chr.19: 57473633-57473633 on Build GRCh38
rs867112712 Chr.19: 57473620-57473620 on Build GRCh38
rs774874633 Chr.19: 57473628-57473628 on Build GRCh38
rs772450574 Chr.19: 57473747-57473747 on Build GRCh38
rs779812820 Chr.19: 57473710-57473710 on Build GRCh38
rs776882068 Chr.19: 57473777-57473777 on Build GRCh38
rs753391786 Chr.19: 57474003-57474003 on Build GRCh38
rs915769586 Chr.19: 57473795-57473795 on Build GRCh38
rs113246264 Chr.19: 57473788-57473788 on Build GRCh38
rs906496133 Chr.19: 57473873-57473873 on Build GRCh38
rs763771771 Chr.19: 57473668-57473668 on Build GRCh38
rs149042633 Chr.19: 57474027-57474027 on Build GRCh38
rs767832887 Chr.19: 57473959-57473959 on Build GRCh38
rs868720787 Chr.19: 57473872-57473872 on Build GRCh38

View Full Product Details