Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57701518-57702013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF154
Entrez Gene ID
7710
Gene Name
zinc finger protein 154
Gene Aliases
pHZ-92
UniGene
Hs.646378
Gene Chromosome Location
Chr.19: 57696275-57709211 on Build GRCh38
Transcripts
NM_001085384.2 NR_110974.1 NR_110975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4462171 c.1235C>T p.P412L Chr.19: 57701714-57701714 on Build GRCh38
7398779 c.1042C>T p.R348W Chr.19: 57701907-57701907 on Build GRCh38
7321710 c.1220A>G p.H407R Chr.19: 57701729-57701729 on Build GRCh38
1397161 c.1046G>A p.G349E Chr.19: 57701903-57701903 on Build GRCh38
4429425 c.1200C>A p.L400L Chr.19: 57701749-57701749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922817967 Chr.19: 57701765-57701765 on Build GRCh38
rs764298018 Chr.19: 57701728-57701728 on Build GRCh38
rs767824724 Chr.19: 57701603-57701603 on Build GRCh38
rs754249358 Chr.19: 57701890-57701890 on Build GRCh38
rs772585903 Chr.19: 57701755-57701755 on Build GRCh38
rs765017369 Chr.19: 57701614-57701614 on Build GRCh38
rs779170902 Chr.19: 57701832-57701832 on Build GRCh38
rs746120122 Chr.19: 57701885-57701885 on Build GRCh38
rs757721147 Chr.19: 57701968-57701968 on Build GRCh38
rs752065572 Chr.19: 57701963-57701963 on Build GRCh38
rs755762500 Chr.19: 57701644-57701644 on Build GRCh38
rs755678504 Chr.19: 57701810-57701810 on Build GRCh38
rs773305678 Chr.19: 57701680-57701680 on Build GRCh38
rs776822065 Chr.19: 57701788-57701788 on Build GRCh38
rs756405083 Chr.19: 57701807-57701807 on Build GRCh38
rs201756288 Chr.19: 57701710-57701710 on Build GRCh38
rs370603127 Chr.19: 57701895-57701895 on Build GRCh38
rs758385935 Chr.19: 57701631-57701631 on Build GRCh38
rs73066544 Chr.19: 57701877-57701877 on Build GRCh38
rs753353043 Chr.19: 57701909-57701909 on Build GRCh38
rs371360296 Chr.19: 57701953-57701953 on Build GRCh38
rs747082901 Chr.19: 57701776-57701776 on Build GRCh38
rs762593561 Chr.19: 57701705-57701705 on Build GRCh38
rs758870902 Chr.19: 57701852-57701852 on Build GRCh38
rs762677300 Chr.19: 57701683-57701683 on Build GRCh38
rs34746514 Chr.19: 57701798-57701798 on Build GRCh38
rs200693217 Chr.19: 57701695-57701695 on Build GRCh38
rs749733265 Chr.19: 57701740-57701740 on Build GRCh38
rs772052746 Chr.19: 57701678-57701678 on Build GRCh38
rs1006892734 Chr.19: 57701811-57701811 on Build GRCh38
rs200989135 Chr.19: 57701906-57701906 on Build GRCh38
rs771211786 Chr.19: 57701785-57701785 on Build GRCh38
rs200045380 Chr.19: 57701723-57701723 on Build GRCh38
rs753072103 Chr.19: 57701952-57701952 on Build GRCh38
rs773184069 Chr.19: 57701905-57701905 on Build GRCh38
rs775306046 Chr.19: 57701702-57701702 on Build GRCh38
rs769948576 Chr.19: 57701973-57701973 on Build GRCh38
rs762733856 Chr.19: 57701707-57701707 on Build GRCh38
rs948468517 Chr.19: 57701696-57701696 on Build GRCh38
rs376150960 Chr.19: 57701894-57701894 on Build GRCh38
rs780316805 Chr.19: 57701977-57701977 on Build GRCh38
rs201131086 Chr.19: 57701734-57701734 on Build GRCh38
rs955957441 Chr.19: 57701889-57701889 on Build GRCh38
rs550456452 Chr.19: 57701593-57701593 on Build GRCh38
rs766966512 Chr.19: 57701717-57701717 on Build GRCh38
rs745538842 Chr.19: 57701643-57701643 on Build GRCh38
rs766188861 Chr.19: 57701821-57701821 on Build GRCh38
rs1037231799 Chr.19: 57701752-57701752 on Build GRCh38
rs1024080116 Chr.19: 57701857-57701857 on Build GRCh38
rs140956119 Chr.19: 57701971-57701971 on Build GRCh38
rs776380083 Chr.19: 57701595-57701595 on Build GRCh38
rs142884205 Chr.19: 57701639-57701639 on Build GRCh38
rs911219231 Chr.19: 57701725-57701725 on Build GRCh38
rs774989842 Chr.19: 57701676-57701677 on Build GRCh38
rs564405661 Chr.19: 57701791-57701791 on Build GRCh38
rs780581615 Chr.19: 57701651-57701651 on Build GRCh38
rs967125727 Chr.19: 57701881-57701881 on Build GRCh38
rs562791985 Chr.19: 57701831-57701831 on Build GRCh38
rs1014007161 Chr.19: 57701540-57701540 on Build GRCh38
rs777943361 Chr.19: 57701959-57701959 on Build GRCh38
rs371886381 Chr.19: 57701828-57701828 on Build GRCh38
rs928749291 Chr.19: 57701719-57701719 on Build GRCh38
rs868541565 Chr.19: 57701733-57701733 on Build GRCh38
rs761583281 Chr.19: 57701604-57701604 on Build GRCh38
rs774971872 Chr.19: 57701673-57701673 on Build GRCh38
rs748313159 Chr.19: 57701674-57701674 on Build GRCh38
rs769357049 Chr.19: 57701665-57701665 on Build GRCh38
rs371361867 Chr.19: 57701728-57701728 on Build GRCh38
rs760908470 Chr.19: 57701613-57701613 on Build GRCh38
rs778340856 Chr.19: 57701869-57701869 on Build GRCh38
rs548319664 Chr.19: 57701878-57701878 on Build GRCh38
rs779259829 Chr.19: 57701970-57701970 on Build GRCh38
rs200841377 Chr.19: 57701833-57701833 on Build GRCh38
rs200741090 Chr.19: 57701687-57701687 on Build GRCh38
rs376109637 Chr.19: 57701794-57701794 on Build GRCh38
rs561745534 Chr.19: 57701884-57701884 on Build GRCh38
rs778222610 Chr.19: 57701721-57701721 on Build GRCh38
rs773216197 Chr.19: 57701771-57701771 on Build GRCh38
rs752429162 Chr.19: 57701619-57701619 on Build GRCh38
rs779274230 Chr.19: 57701748-57701748 on Build GRCh38
rs202179866 Chr.19: 57701907-57701907 on Build GRCh38
rs577728710 Chr.19: 57701795-57701795 on Build GRCh38
rs759313525 Chr.19: 57701914-57701914 on Build GRCh38
rs771966840 Chr.19: 57701904-57701904 on Build GRCh38
rs748637241 Chr.19: 57701754-57701754 on Build GRCh38
rs749659652 Chr.19: 57701655-57701655 on Build GRCh38
rs942821842 Chr.19: 57701735-57701735 on Build GRCh38
rs764915209 Chr.19: 57701927-57701927 on Build GRCh38
rs1041095502 Chr.19: 57701739-57701739 on Build GRCh38
rs774634462 Chr.19: 57701590-57701590 on Build GRCh38
rs755377909 Chr.19: 57701747-57701747 on Build GRCh38
rs750628807 Chr.19: 57701612-57701612 on Build GRCh38
rs780705185 Chr.19: 57701640-57701640 on Build GRCh38
rs201505711 Chr.19: 57701961-57701961 on Build GRCh38
rs757151640 Chr.19: 57701716-57701716 on Build GRCh38

View Full Product Details