Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58040365-58040868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN1
Entrez Gene ID
284312
Gene Name
zinc finger and SCAN domain containing 1
Gene Aliases
MZF-1, ZNF915
UniGene
Hs.643437
Gene Chromosome Location
Chr.19: 58033989-58058907 on Build GRCh38
Transcripts
NM_182572.3 XM_006723149.2 XM_017026609.1 XM_017026610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584316 c.420G>A p.E140E Chr.19: 58040499-58040499 on Build GRCh38
6152533 c.373C>A p.L125M Chr.19: 58040452-58040452 on Build GRCh38
3673041 c.448C>A p.Q150K Chr.19: 58040527-58040527 on Build GRCh38
6880098 c.424G>A p.E142K Chr.19: 58040503-58040503 on Build GRCh38
4745397 c.435G>A p.K145K Chr.19: 58040514-58040514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762110190 Chr.19: 58040541-58040541 on Build GRCh38
rs776175785 Chr.19: 58040528-58040528 on Build GRCh38
rs953167978 Chr.19: 58040662-58040662 on Build GRCh38
rs778846574 Chr.19: 58040420-58040420 on Build GRCh38
rs372180167 Chr.19: 58040406-58040406 on Build GRCh38
rs557023929 Chr.19: 58040839-58040839 on Build GRCh38
rs926143203 Chr.19: 58040690-58040690 on Build GRCh38
rs562232066 Chr.19: 58040700-58040700 on Build GRCh38
rs150688831 Chr.19: 58040810-58040810 on Build GRCh38
rs375907906 Chr.19: 58040552-58040552 on Build GRCh38
rs539459841 Chr.19: 58040786-58040786 on Build GRCh38
rs375992446 Chr.19: 58040443-58040443 on Build GRCh38
rs560288905 Chr.19: 58040557-58040557 on Build GRCh38
rs753956783 Chr.19: 58040558-58040558 on Build GRCh38
rs371329143 Chr.19: 58040575-58040575 on Build GRCh38
rs1038382213 Chr.19: 58040811-58040811 on Build GRCh38
rs774760226 Chr.19: 58040540-58040540 on Build GRCh38
rs1020615972 Chr.19: 58040391-58040391 on Build GRCh38
rs1055303435 Chr.19: 58040642-58040642 on Build GRCh38
rs1009207941 Chr.19: 58040842-58040842 on Build GRCh38
rs759138519 Chr.19: 58040459-58040459 on Build GRCh38
rs967176244 Chr.19: 58040686-58040686 on Build GRCh38
rs1019392503 Chr.19: 58040717-58040717 on Build GRCh38
rs916156834 Chr.19: 58040731-58040731 on Build GRCh38
rs918084813 Chr.19: 58040832-58040832 on Build GRCh38
rs768562283 Chr.19: 58040408-58040408 on Build GRCh38
rs994749363 Chr.19: 58040621-58040621 on Build GRCh38
rs745306671 Chr.19: 58040532-58040532 on Build GRCh38
rs769161736 Chr.19: 58040596-58040596 on Build GRCh38
rs748092795 Chr.19: 58040499-58040499 on Build GRCh38
rs139461841 Chr.19: 58040494-58040494 on Build GRCh38
rs770567249 Chr.19: 58040523-58040523 on Build GRCh38
rs147981030 Chr.19: 58040500-58040500 on Build GRCh38
rs1029356093 Chr.19: 58040835-58040835 on Build GRCh38
rs569643875 Chr.19: 58040831-58040831 on Build GRCh38
rs899004746 Chr.19: 58040829-58040829 on Build GRCh38
rs777593445 Chr.19: 58040560-58040560 on Build GRCh38
rs147151087 Chr.19: 58040493-58040493 on Build GRCh38
rs767685924 Chr.19: 58040487-58040487 on Build GRCh38
rs769440638 Chr.19: 58040536-58040536 on Build GRCh38
rs781332273 Chr.19: 58040402-58040402 on Build GRCh38
rs758331839 Chr.19: 58040559-58040559 on Build GRCh38
rs555114438 Chr.19: 58040476-58040476 on Build GRCh38
rs936266363 Chr.19: 58040527-58040527 on Build GRCh38
rs977154362 Chr.19: 58040687-58040687 on Build GRCh38
rs369956028 Chr.19: 58040561-58040561 on Build GRCh38
rs757474735 Chr.19: 58040483-58040483 on Build GRCh38
rs563228900 Chr.19: 58040663-58040663 on Build GRCh38
rs748860418 Chr.19: 58040589-58040589 on Build GRCh38
rs777591736 Chr.19: 58040502-58040502 on Build GRCh38
rs775194762 Chr.19: 58040537-58040537 on Build GRCh38
rs376155860 Chr.19: 58040401-58040401 on Build GRCh38
rs200020953 Chr.19: 58040444-58040444 on Build GRCh38
rs776182361 Chr.19: 58040449-58040449 on Build GRCh38
rs201982170 Chr.19: 58040468-58040468 on Build GRCh38
rs747900160 Chr.19: 58040430-58040430 on Build GRCh38
rs1032591253 Chr.19: 58040455-58040455 on Build GRCh38
rs887083260 Chr.19: 58040755-58040755 on Build GRCh38
rs371426023 Chr.19: 58040719-58040719 on Build GRCh38
rs1028909652 Chr.19: 58040654-58040654 on Build GRCh38
rs780929372 Chr.19: 58040567-58040567 on Build GRCh38
rs143428215 Chr.19: 58040469-58040469 on Build GRCh38
rs113289199 Chr.19: 58040669-58040669 on Build GRCh38
rs963170512 Chr.19: 58040608-58040608 on Build GRCh38
rs373374080 Chr.19: 58040568-58040568 on Build GRCh38
rs375604504 Chr.19: 58040597-58040597 on Build GRCh38
rs543941255 Chr.19: 58040809-58040809 on Build GRCh38
rs200244331 Chr.19: 58040569-58040569 on Build GRCh38
rs756131242 Chr.19: 58040555-58040555 on Build GRCh38
rs1026275686 Chr.19: 58040735-58040735 on Build GRCh38
rs45571742 Chr.19: 58040743-58040743 on Build GRCh38
rs376235386 Chr.19: 58040830-58040830 on Build GRCh38
rs570451291 Chr.19: 58040748-58040748 on Build GRCh38
rs116287017 Chr.19: 58040675-58040675 on Build GRCh38
rs765796829 Chr.19: 58040461-58040461 on Build GRCh38
rs1013330457 Chr.19: 58040655-58040655 on Build GRCh38
rs954031656 Chr.19: 58040692-58040692 on Build GRCh38
rs1042123980 Chr.19: 58040840-58040840 on Build GRCh38
rs142030527 Chr.19: 58040676-58040676 on Build GRCh38
rs540289774 Chr.19: 58040543-58040543 on Build GRCh38
rs527394909 Chr.19: 58040732-58040732 on Build GRCh38
rs902139735 Chr.19: 58040667-58040667 on Build GRCh38
rs755520798 Chr.19: 58040530-58040530 on Build GRCh38
rs894506145 Chr.19: 58040553-58040553 on Build GRCh38
rs751937817 Chr.19: 58040472-58040472 on Build GRCh38
rs534586174 Chr.19: 58040518-58040518 on Build GRCh38
rs776143491 Chr.19: 58040466-58040466 on Build GRCh38
rs764551513 Chr.19: 58040539-58040539 on Build GRCh38

View Full Product Details