Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 41538-42045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110C
Entrez Gene ID
642273
Gene Name
family with sequence similarity 110 member C
Gene Aliases
-
UniGene
Hs.8379
Gene Chromosome Location
Chr.2: 38814-47077 on Build GRCh38
Transcripts
NM_001077710.2 XM_011510372.2 XM_017004691.1

SNP Detail

SNP ID Chromosome Location
rs1010238300 Chr.2: 41719-41719 on Build GRCh38
rs780702359 Chr.2: 41934-41934 on Build GRCh38
rs528195106 Chr.2: 41642-41642 on Build GRCh38
rs914677478 Chr.2: 41707-41707 on Build GRCh38
rs898840754 Chr.2: 41606-41606 on Build GRCh38
rs953321840 Chr.2: 41787-41787 on Build GRCh38
rs756743255 Chr.2: 41932-41932 on Build GRCh38
rs918285422 Chr.2: 41781-41781 on Build GRCh38
rs764019243 Chr.2: 41662-41662 on Build GRCh38
rs868264304 Chr.2: 41718-41718 on Build GRCh38
rs530845814 Chr.2: 41850-41850 on Build GRCh38
rs201975695 Chr.2: 41938-41938 on Build GRCh38
rs186906574 Chr.2: 41868-41868 on Build GRCh38
rs540368251 Chr.2: 41767-41767 on Build GRCh38
rs778219814 Chr.2: 41675-41675 on Build GRCh38
rs748997045 Chr.2: 41595-41595 on Build GRCh38
rs377150701 Chr.2: 41615-41615 on Build GRCh38
rs755788735 Chr.2: 41583-41583 on Build GRCh38
rs777588440 Chr.2: 41585-41585 on Build GRCh38
rs980303607 Chr.2: 41721-41721 on Build GRCh38
rs551407872 Chr.2: 41768-41768 on Build GRCh38
rs773351807 Chr.2: 41623-41623 on Build GRCh38
rs374921114 Chr.2: 41851-41851 on Build GRCh38
rs1048242891 Chr.2: 41598-41598 on Build GRCh38
rs371974298 Chr.2: 41745-41745 on Build GRCh38
rs1031616388 Chr.2: 42003-42003 on Build GRCh38
rs968713278 Chr.2: 41731-41731 on Build GRCh38
rs528183041 Chr.2: 41896-41896 on Build GRCh38
rs769060954 Chr.2: 41614-41614 on Build GRCh38
rs868456528 Chr.2: 41627-41627 on Build GRCh38
rs77179864 Chr.2: 41647-41647 on Build GRCh38
rs138120439 Chr.2: 41720-41720 on Build GRCh38
rs1036081172 Chr.2: 41872-41872 on Build GRCh38
rs995288964 Chr.2: 41592-41592 on Build GRCh38
rs572998910 Chr.2: 41764-41764 on Build GRCh38
rs776805931 Chr.2: 41619-41619 on Build GRCh38
rs1002099676 Chr.2: 41955-41955 on Build GRCh38
rs778545375 Chr.2: 41609-41609 on Build GRCh38
rs546750039 Chr.2: 41981-41981 on Build GRCh38
rs901079096 Chr.2: 41972-41972 on Build GRCh38
rs570524274 Chr.2: 41620-41620 on Build GRCh38
rs931029440 Chr.2: 41783-41783 on Build GRCh38
rs940354272 Chr.2: 41580-41580 on Build GRCh38
rs557396675 Chr.2: 41708-41708 on Build GRCh38
rs151311604 Chr.2: 41613-41613 on Build GRCh38
rs1057337932 Chr.2: 41974-41974 on Build GRCh38
rs200542783 Chr.2: 41602-41602 on Build GRCh38
rs181892339 Chr.2: 41761-41761 on Build GRCh38
rs542031609 Chr.2: 41821-41821 on Build GRCh38
rs753528894 Chr.2: 41670-41670 on Build GRCh38
rs537583942 Chr.2: 41954-41954 on Build GRCh38
rs763259169 Chr.2: 41808-41808 on Build GRCh38
rs574575857 Chr.2: 41792-41792 on Build GRCh38
rs759320678 Chr.2: 41629-41629 on Build GRCh38
rs150484376 Chr.2: 41686-41686 on Build GRCh38
rs746302953 Chr.2: 41610-41610 on Build GRCh38
rs568599462 Chr.2: 41672-41672 on Build GRCh38
rs1036169497 Chr.2: 41659-41659 on Build GRCh38
rs563403590 Chr.2: 41822-41822 on Build GRCh38
rs920235190 Chr.2: 41904-41904 on Build GRCh38
rs930332208 Chr.2: 41930-41930 on Build GRCh38
rs545766786 Chr.2: 41867-41867 on Build GRCh38
rs373037350 Chr.2: 41612-41612 on Build GRCh38
rs764345865 Chr.2: 41658-41658 on Build GRCh38
rs769372062 Chr.2: 41680-41680 on Build GRCh38

View Full Product Details