Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27661042-27661554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT7L
Entrez Gene ID
9913
Gene Name
SPT7-like STAGA complex gamma subunit
Gene Aliases
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H
UniGene
Hs.6232
Gene Chromosome Location
Chr.2: 27642568-27663840 on Build GRCh38
Transcripts
NM_001282729.1 NM_001282730.1 NM_001282731.1 NM_001282732.1 NM_014860.2 XM_005264672.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6440294 c.278G>A p.G93D Chr.2: 27661125-27661125 on Build GRCh38
419205 c.38C>T p.S13L Chr.2: 27661365-27661365 on Build GRCh38
6440295 c.182G>A p.S61N Chr.2: 27661221-27661221 on Build GRCh38
4443405 c.25G>T p.E9* Chr.2: 27661378-27661378 on Build GRCh38
6452120 c.80G>A p.R27Q Chr.2: 27661323-27661323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773997268 Chr.2: 27661385-27661385 on Build GRCh38
rs917490393 Chr.2: 27661095-27661095 on Build GRCh38
rs776267517 Chr.2: 27661413-27661413 on Build GRCh38
rs764987395 Chr.2: 27661432-27661432 on Build GRCh38
rs1000299420 Chr.2: 27661289-27661289 on Build GRCh38
rs569641226 Chr.2: 27661450-27661450 on Build GRCh38
rs780476400 Chr.2: 27661139-27661139 on Build GRCh38
rs760706273 Chr.2: 27661410-27661410 on Build GRCh38
rs755452936 Chr.2: 27661316-27661316 on Build GRCh38
rs200392709 Chr.2: 27661072-27661072 on Build GRCh38
rs200959399 Chr.2: 27661323-27661323 on Build GRCh38
rs755809244 Chr.2: 27661107-27661107 on Build GRCh38
rs763626280 Chr.2: 27661411-27661411 on Build GRCh38
rs368383395 Chr.2: 27661369-27661369 on Build GRCh38
rs114487538 Chr.2: 27661182-27661182 on Build GRCh38
rs762663385 Chr.2: 27661361-27661363 on Build GRCh38
rs919943998 Chr.2: 27661371-27661371 on Build GRCh38
rs367572958 Chr.2: 27661173-27661173 on Build GRCh38
rs746953634 Chr.2: 27661144-27661145 on Build GRCh38
rs751395014 Chr.2: 27661084-27661084 on Build GRCh38
rs776912197 Chr.2: 27661278-27661281 on Build GRCh38
rs972250880 Chr.2: 27661457-27661457 on Build GRCh38
rs1020303374 Chr.2: 27661264-27661264 on Build GRCh38
rs746277653 Chr.2: 27661148-27661148 on Build GRCh38
rs774175291 Chr.2: 27661417-27661418 on Build GRCh38
rs374524858 Chr.2: 27661505-27661505 on Build GRCh38
rs201679796 Chr.2: 27661123-27661123 on Build GRCh38
rs372331209 Chr.2: 27661388-27661388 on Build GRCh38
rs550823686 Chr.2: 27661443-27661443 on Build GRCh38
rs777913728 Chr.2: 27661349-27661349 on Build GRCh38
rs759418062 Chr.2: 27661387-27661387 on Build GRCh38
rs767613710 Chr.2: 27661249-27661249 on Build GRCh38
rs202100505 Chr.2: 27661262-27661262 on Build GRCh38
rs779196980 Chr.2: 27661106-27661106 on Build GRCh38
rs775522415 Chr.2: 27661394-27661394 on Build GRCh38
rs536372540 Chr.2: 27661355-27661355 on Build GRCh38
rs1033069366 Chr.2: 27661328-27661328 on Build GRCh38
rs748274586 Chr.2: 27661180-27661180 on Build GRCh38
rs753378560 Chr.2: 27661272-27661272 on Build GRCh38
rs769815831 Chr.2: 27661337-27661337 on Build GRCh38
rs761355734 Chr.2: 27661421-27661421 on Build GRCh38
rs910281723 Chr.2: 27661527-27661527 on Build GRCh38
rs773381394 Chr.2: 27661183-27661183 on Build GRCh38
rs142583753 Chr.2: 27661437-27661437 on Build GRCh38
rs6756254 Chr.2: 27661091-27661091 on Build GRCh38
rs959152309 Chr.2: 27661422-27661422 on Build GRCh38
rs201942828 Chr.2: 27661109-27661109 on Build GRCh38
rs548044329 Chr.2: 27661263-27661263 on Build GRCh38
rs763258355 Chr.2: 27661202-27661202 on Build GRCh38
rs780273689 Chr.2: 27661307-27661307 on Build GRCh38
rs1038303297 Chr.2: 27661354-27661354 on Build GRCh38
rs762015093 Chr.2: 27661067-27661067 on Build GRCh38
rs558522538 Chr.2: 27661491-27661491 on Build GRCh38
rs774270293 Chr.2: 27661208-27661208 on Build GRCh38
rs781692578 Chr.2: 27661319-27661319 on Build GRCh38
rs759379847 Chr.2: 27661221-27661221 on Build GRCh38
rs191619402 Chr.2: 27661080-27661080 on Build GRCh38
rs536598831 Chr.2: 27661460-27661460 on Build GRCh38
rs531573550 Chr.2: 27661092-27661092 on Build GRCh38
rs753562086 Chr.2: 27661114-27661114 on Build GRCh38
rs745827063 Chr.2: 27661469-27661469 on Build GRCh38
rs971288974 Chr.2: 27661248-27661248 on Build GRCh38
rs776041964 Chr.2: 27661447-27661447 on Build GRCh38
rs769060906 Chr.2: 27661175-27661176 on Build GRCh38
rs112261868 Chr.2: 27661511-27661511 on Build GRCh38
rs552777370 Chr.2: 27661524-27661524 on Build GRCh38
rs774705095 Chr.2: 27661315-27661315 on Build GRCh38
rs752015900 Chr.2: 27661314-27661314 on Build GRCh38
rs770566368 Chr.2: 27661205-27661205 on Build GRCh38
rs771243712 Chr.2: 27661372-27661372 on Build GRCh38
rs756934640 Chr.2: 27661301-27661301 on Build GRCh38
rs777610958 Chr.2: 27661113-27661113 on Build GRCh38
rs913043421 Chr.2: 27661209-27661209 on Build GRCh38
rs577106284 Chr.2: 27661506-27661506 on Build GRCh38
rs751837051 Chr.2: 27661438-27661438 on Build GRCh38
rs903144517 Chr.2: 27661266-27661266 on Build GRCh38
rs370287900 Chr.2: 27661253-27661253 on Build GRCh38
rs752315236 Chr.2: 27661094-27661094 on Build GRCh38
rs184249965 Chr.2: 27661179-27661179 on Build GRCh38
rs529056582 Chr.2: 27661405-27661407 on Build GRCh38
rs747404460 Chr.2: 27661165-27661165 on Build GRCh38

View Full Product Details