Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29143652-29144189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP4
Entrez Gene ID
79745
Gene Name
CAP-Gly domain containing linker protein family member 4
Gene Aliases
RSNL2
UniGene
Hs.122927
Gene Chromosome Location
Chr.2: 29097676-29183813 on Build GRCh38
Transcripts
NM_001287527.1 NM_001287528.1 NM_024692.5 NR_109844.1 XM_005264562.2 XM_005264563.1 XM_006712104.2 XM_006712105.3 XM_011533109.1 XM_011533110.1 XM_017004958.1 XM_017004959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601255 c.785C>T p.S262L Chr.2: 29143845-29143845 on Build GRCh38
5823666 c.807T>A p.Y269* Chr.2: 29143867-29143867 on Build GRCh38
5713059 c.815delT p.V272fs*12 Chr.2: 29143875-29143875 on Build GRCh38
5354500 c.774A>C p.S258S Chr.2: 29143834-29143834 on Build GRCh38
3580971 c.682C>T p.P228S Chr.2: 29143742-29143742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776806345 Chr.2: 29143817-29143817 on Build GRCh38
rs755905179 Chr.2: 29143872-29143872 on Build GRCh38
rs146399446 Chr.2: 29143822-29143822 on Build GRCh38
rs3213947 Chr.2: 29143964-29143964 on Build GRCh38
rs759605334 Chr.2: 29143948-29143948 on Build GRCh38
rs770773865 Chr.2: 29143709-29143709 on Build GRCh38
rs138036770 Chr.2: 29143748-29143748 on Build GRCh38
rs138901368 Chr.2: 29143761-29143761 on Build GRCh38
rs908013011 Chr.2: 29144157-29144157 on Build GRCh38
rs959903700 Chr.2: 29144114-29144114 on Build GRCh38
rs767739877 Chr.2: 29143691-29143695 on Build GRCh38
rs377733033 Chr.2: 29144007-29144007 on Build GRCh38
rs1022796654 Chr.2: 29144148-29144148 on Build GRCh38
rs78064102 Chr.2: 29144014-29144014 on Build GRCh38
rs762163285 Chr.2: 29143742-29143742 on Build GRCh38
rs556428249 Chr.2: 29143798-29143798 on Build GRCh38
rs751811586 Chr.2: 29143853-29143853 on Build GRCh38
rs750023398 Chr.2: 29143986-29143986 on Build GRCh38
rs997137654 Chr.2: 29143758-29143758 on Build GRCh38
rs144223606 Chr.2: 29143897-29143897 on Build GRCh38
rs748505951 Chr.2: 29143919-29143919 on Build GRCh38
rs971706820 Chr.2: 29143773-29143773 on Build GRCh38
rs146544139 Chr.2: 29143915-29143915 on Build GRCh38
rs34100808 Chr.2: 29144013-29144014 on Build GRCh38
rs773730812 Chr.2: 29143923-29143923 on Build GRCh38
rs200713156 Chr.2: 29144009-29144009 on Build GRCh38
rs746624720 Chr.2: 29143995-29143995 on Build GRCh38
rs773742300 Chr.2: 29143823-29143823 on Build GRCh38
rs769144399 Chr.2: 29143808-29143808 on Build GRCh38
rs111656389 Chr.2: 29143832-29143832 on Build GRCh38
rs184820320 Chr.2: 29143704-29143704 on Build GRCh38
rs763411760 Chr.2: 29143927-29143927 on Build GRCh38
rs765375647 Chr.2: 29143747-29143747 on Build GRCh38
rs751758315 Chr.2: 29143765-29143765 on Build GRCh38
rs142175946 Chr.2: 29143952-29143952 on Build GRCh38
rs763069759 Chr.2: 29143826-29143826 on Build GRCh38
rs879195153 Chr.2: 29143711-29143711 on Build GRCh38
rs754791545 Chr.2: 29143770-29143770 on Build GRCh38
rs559668580 Chr.2: 29144095-29144095 on Build GRCh38
rs1034277136 Chr.2: 29144049-29144049 on Build GRCh38
rs58545152 Chr.2: 29143726-29143726 on Build GRCh38
rs745475089 Chr.2: 29143803-29143803 on Build GRCh38
rs771723506 Chr.2: 29143912-29143912 on Build GRCh38
rs770539151 Chr.2: 29143794-29143794 on Build GRCh38
rs1032765226 Chr.2: 29143749-29143749 on Build GRCh38
rs373136635 Chr.2: 29143858-29143858 on Build GRCh38
rs767765108 Chr.2: 29143867-29143867 on Build GRCh38
rs947379276 Chr.2: 29143880-29143880 on Build GRCh38
rs1028504947 Chr.2: 29143759-29143759 on Build GRCh38
rs763056332 Chr.2: 29144140-29144140 on Build GRCh38
rs772537855 Chr.2: 29143689-29143689 on Build GRCh38
rs748855460 Chr.2: 29143778-29143778 on Build GRCh38
rs368784525 Chr.2: 29144005-29144005 on Build GRCh38
rs762227443 Chr.2: 29143821-29143821 on Build GRCh38
rs958647847 Chr.2: 29144066-29144066 on Build GRCh38
rs771005284 Chr.2: 29143933-29143933 on Build GRCh38
rs932787121 Chr.2: 29143953-29143953 on Build GRCh38
rs763906063 Chr.2: 29143979-29143979 on Build GRCh38
rs777223042 Chr.2: 29143741-29143741 on Build GRCh38
rs375822749 Chr.2: 29143985-29143985 on Build GRCh38
rs968662574 Chr.2: 29144150-29144150 on Build GRCh38
rs113143402 Chr.2: 29144007-29144008 on Build GRCh38
rs34825217 Chr.2: 29144010-29144011 on Build GRCh38
rs3217572 Chr.2: 29144009-29144010 on Build GRCh38
rs572200409 Chr.2: 29143917-29143917 on Build GRCh38
rs752501561 Chr.2: 29143870-29143870 on Build GRCh38
rs113385772 Chr.2: 29144088-29144088 on Build GRCh38
rs771682545 Chr.2: 29143916-29143916 on Build GRCh38
rs374617989 Chr.2: 29144006-29144006 on Build GRCh38
rs775736015 Chr.2: 29143735-29143737 on Build GRCh38
rs551983685 Chr.2: 29144135-29144135 on Build GRCh38
rs115153920 Chr.2: 29143777-29143777 on Build GRCh38
rs145908487 Chr.2: 29143980-29143980 on Build GRCh38
rs888661823 Chr.2: 29143977-29143977 on Build GRCh38
rs779625281 Chr.2: 29143991-29143991 on Build GRCh38
rs778362993 Chr.2: 29143881-29143881 on Build GRCh38
rs146197855 Chr.2: 29143753-29143753 on Build GRCh38
rs990779389 Chr.2: 29144081-29144081 on Build GRCh38
rs981723995 Chr.2: 29144032-29144032 on Build GRCh38
rs201519559 Chr.2: 29143781-29143781 on Build GRCh38
rs778496677 Chr.2: 29143898-29143898 on Build GRCh38
rs900128948 Chr.2: 29143941-29143941 on Build GRCh38
rs1041257508 Chr.2: 29143750-29143750 on Build GRCh38
rs563242442 Chr.2: 29144002-29144002 on Build GRCh38
rs930718323 Chr.2: 29144090-29144090 on Build GRCh38
rs369326999 Chr.2: 29143833-29143833 on Build GRCh38
rs375223459 Chr.2: 29143762-29143762 on Build GRCh38
rs759545973 Chr.2: 29143685-29143685 on Build GRCh38
rs920673761 Chr.2: 29143835-29143835 on Build GRCh38
rs770172852 Chr.2: 29143922-29143922 on Build GRCh38
rs774112857 Chr.2: 29143723-29143723 on Build GRCh38
rs189297473 Chr.2: 29143987-29143987 on Build GRCh38
rs775704190 Chr.2: 29144055-29144055 on Build GRCh38
rs760405888 Chr.2: 29143975-29143975 on Build GRCh38
rs753775161 Chr.2: 29143893-29143893 on Build GRCh38
rs774471427 Chr.2: 29143942-29143942 on Build GRCh38
rs901321216 Chr.2: 29143751-29143751 on Build GRCh38
rs35024908 Chr.2: 29143780-29143780 on Build GRCh38

View Full Product Details