Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36509897-36510392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040567 c.1540T>A p.L514M Chr.2: 36510021-36510021 on Build GRCh38
4674695 c.1589G>A p.C530Y Chr.2: 36510070-36510070 on Build GRCh38
5034989 c.1652G>A p.G551E Chr.2: 36510133-36510133 on Build GRCh38
5010808 c.1527A>T p.K509N Chr.2: 36510008-36510008 on Build GRCh38
3962813 c.1509A>G p.E503E Chr.2: 36509990-36509990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995126353 Chr.2: 36510282-36510282 on Build GRCh38
rs758321326 Chr.2: 36510151-36510151 on Build GRCh38
rs967050453 Chr.2: 36509925-36509925 on Build GRCh38
rs772826709 Chr.2: 36510182-36510182 on Build GRCh38
rs528470253 Chr.2: 36510238-36510238 on Build GRCh38
rs767275131 Chr.2: 36509994-36509994 on Build GRCh38
rs958898640 Chr.2: 36510099-36510099 on Build GRCh38
rs1009254026 Chr.2: 36510293-36510293 on Build GRCh38
rs537181703 Chr.2: 36510355-36510355 on Build GRCh38
rs1024044444 Chr.2: 36510146-36510146 on Build GRCh38
rs756341111 Chr.2: 36509960-36509960 on Build GRCh38
rs769629338 Chr.2: 36510180-36510180 on Build GRCh38
rs746988885 Chr.2: 36510164-36510164 on Build GRCh38
rs535026338 Chr.2: 36509926-36509926 on Build GRCh38
rs752053361 Chr.2: 36509933-36509933 on Build GRCh38
rs896508470 Chr.2: 36509931-36509931 on Build GRCh38
rs573849913 Chr.2: 36510219-36510219 on Build GRCh38
rs760647739 Chr.2: 36510135-36510136 on Build GRCh38
rs200381845 Chr.2: 36510065-36510065 on Build GRCh38
rs576111073 Chr.2: 36510014-36510014 on Build GRCh38
rs776372225 Chr.2: 36510011-36510011 on Build GRCh38
rs368068258 Chr.2: 36509965-36509965 on Build GRCh38
rs761568969 Chr.2: 36510013-36510013 on Build GRCh38
rs750547617 Chr.2: 36510150-36510150 on Build GRCh38
rs77941106 Chr.2: 36510214-36510214 on Build GRCh38
rs764362372 Chr.2: 36510261-36510261 on Build GRCh38
rs932101088 Chr.2: 36509967-36509967 on Build GRCh38
rs755548689 Chr.2: 36509937-36509937 on Build GRCh38
rs778967172 Chr.2: 36509984-36509984 on Build GRCh38
rs767921061 Chr.2: 36509945-36509945 on Build GRCh38
rs201281263 Chr.2: 36510143-36510143 on Build GRCh38
rs768277117 Chr.2: 36510167-36510167 on Build GRCh38
rs1024491432 Chr.2: 36510315-36510315 on Build GRCh38
rs940690537 Chr.2: 36510272-36510272 on Build GRCh38
rs925841949 Chr.2: 36509929-36509929 on Build GRCh38
rs147714435 Chr.2: 36510079-36510079 on Build GRCh38
rs757408404 Chr.2: 36510275-36510275 on Build GRCh38
rs79366315 Chr.2: 36510068-36510068 on Build GRCh38
rs530947765 Chr.2: 36510217-36510217 on Build GRCh38
rs753078667 Chr.2: 36510054-36510054 on Build GRCh38
rs142291323 Chr.2: 36510152-36510152 on Build GRCh38
rs62136183 Chr.2: 36510304-36510304 on Build GRCh38
rs921121254 Chr.2: 36510245-36510245 on Build GRCh38
rs182909671 Chr.2: 36510302-36510302 on Build GRCh38
rs771958347 Chr.2: 36509988-36509988 on Build GRCh38
rs746819860 Chr.2: 36510003-36510003 on Build GRCh38
rs762414606 Chr.2: 36510188-36510188 on Build GRCh38
rs72866875 Chr.2: 36510235-36510235 on Build GRCh38
rs754004072 Chr.2: 36510078-36510078 on Build GRCh38
rs760107147 Chr.2: 36510020-36510020 on Build GRCh38
rs75011392 Chr.2: 36509955-36509955 on Build GRCh38
rs538830200 Chr.2: 36510274-36510274 on Build GRCh38
rs200292966 Chr.2: 36510036-36510036 on Build GRCh38
rs530926274 Chr.2: 36510004-36510004 on Build GRCh38
rs758876352 Chr.2: 36510362-36510362 on Build GRCh38
rs761008814 Chr.2: 36510240-36510240 on Build GRCh38
rs781137503 Chr.2: 36510170-36510170 on Build GRCh38
rs780784825 Chr.2: 36510019-36510019 on Build GRCh38
rs900694692 Chr.2: 36510267-36510268 on Build GRCh38
rs767760925 Chr.2: 36510309-36510310 on Build GRCh38
rs775244514 Chr.2: 36510132-36510133 on Build GRCh38
rs563602661 Chr.2: 36510216-36510216 on Build GRCh38
rs202157362 Chr.2: 36510176-36510176 on Build GRCh38
rs867664090 Chr.2: 36510095-36510095 on Build GRCh38
rs894664482 Chr.2: 36510288-36510288 on Build GRCh38
rs779872465 Chr.2: 36509995-36509995 on Build GRCh38
rs754093938 Chr.2: 36509964-36509964 on Build GRCh38
rs942176485 Chr.2: 36510260-36510260 on Build GRCh38
rs1056038132 Chr.2: 36510286-36510286 on Build GRCh38
rs750910151 Chr.2: 36510357-36510357 on Build GRCh38
rs186733127 Chr.2: 36510251-36510251 on Build GRCh38
rs201434329 Chr.2: 36510035-36510035 on Build GRCh38
rs777900599 Chr.2: 36509963-36509963 on Build GRCh38
rs12997487 Chr.2: 36509985-36509985 on Build GRCh38
rs553147837 Chr.2: 36509928-36509928 on Build GRCh38
rs899158204 Chr.2: 36510276-36510276 on Build GRCh38
rs967112512 Chr.2: 36510317-36510317 on Build GRCh38
rs1052297154 Chr.2: 36510262-36510262 on Build GRCh38
rs747649849 Chr.2: 36510278-36510278 on Build GRCh38
rs996991034 Chr.2: 36510267-36510267 on Build GRCh38
rs557537588 Chr.2: 36510010-36510010 on Build GRCh38
rs192038384 Chr.2: 36510265-36510265 on Build GRCh38
rs998483530 Chr.2: 36510329-36510329 on Build GRCh38

View Full Product Details