Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36696598-36697104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIT
Entrez Gene ID
5212
Gene Name
vitrin
Gene Aliases
-
UniGene
Hs.137415
Gene Chromosome Location
Chr.2: 36696685-36814794 on Build GRCh38
Transcripts
NM_001177969.1 NM_001177970.1 NM_001177971.1 NM_001177972.1 NM_053276.3 XM_011532905.1 XM_011532906.1 XM_017004325.1 XM_017004326.1

SNP Detail

SNP ID Chromosome Location
rs959808572 Chr.2: 36696817-36696817 on Build GRCh38
rs768755965 Chr.2: 36696837-36696837 on Build GRCh38
rs540350919 Chr.2: 36696657-36696657 on Build GRCh38
rs992304959 Chr.2: 36696823-36696825 on Build GRCh38
rs1052573282 Chr.2: 36697010-36697010 on Build GRCh38
rs7420213 Chr.2: 36696790-36696790 on Build GRCh38
rs906668867 Chr.2: 36696733-36696733 on Build GRCh38
rs1045834841 Chr.2: 36696980-36696980 on Build GRCh38
rs146195400 Chr.2: 36697073-36697074 on Build GRCh38
rs911964590 Chr.2: 36696809-36696809 on Build GRCh38
rs57176159 Chr.2: 36696655-36696655 on Build GRCh38
rs945492097 Chr.2: 36697005-36697008 on Build GRCh38
rs902915210 Chr.2: 36696998-36696998 on Build GRCh38
rs908479364 Chr.2: 36696675-36696675 on Build GRCh38
rs949758494 Chr.2: 36696942-36696942 on Build GRCh38
rs80048790 Chr.2: 36696985-36696985 on Build GRCh38
rs369229164 Chr.2: 36696755-36696757 on Build GRCh38
rs751245174 Chr.2: 36696733-36696736 on Build GRCh38
rs34118705 Chr.2: 36696801-36696802 on Build GRCh38
rs79351974 Chr.2: 36696757-36696757 on Build GRCh38
rs949708604 Chr.2: 36696643-36696643 on Build GRCh38
rs1046736931 Chr.2: 36696662-36696662 on Build GRCh38
rs941000181 Chr.2: 36696895-36696895 on Build GRCh38
rs1029372313 Chr.2: 36696771-36696771 on Build GRCh38
rs1055860345 Chr.2: 36696695-36696695 on Build GRCh38
rs958269980 Chr.2: 36696758-36696758 on Build GRCh38
rs991559552 Chr.2: 36696666-36696666 on Build GRCh38
rs1038279678 Chr.2: 36696692-36696692 on Build GRCh38
rs1011553714 Chr.2: 36697025-36697025 on Build GRCh38
rs958766420 Chr.2: 36696660-36696660 on Build GRCh38
rs932488839 Chr.2: 36696935-36696935 on Build GRCh38
rs796215191 Chr.2: 36696755-36696757 on Build GRCh38
rs982511382 Chr.2: 36696875-36696875 on Build GRCh38
rs6544026 Chr.2: 36696755-36696755 on Build GRCh38
rs987931828 Chr.2: 36696808-36696808 on Build GRCh38
rs1001557842 Chr.2: 36696791-36696791 on Build GRCh38
rs567080475 Chr.2: 36697073-36697073 on Build GRCh38
rs971869699 Chr.2: 36696870-36696870 on Build GRCh38
rs911704541 Chr.2: 36696636-36696636 on Build GRCh38
rs561517131 Chr.2: 36696686-36696686 on Build GRCh38
rs962125844 Chr.2: 36696850-36696850 on Build GRCh38
rs763238345 Chr.2: 36696730-36696730 on Build GRCh38
rs762366266 Chr.2: 36696735-36696736 on Build GRCh38
rs1024074932 Chr.2: 36696830-36696830 on Build GRCh38
rs940541049 Chr.2: 36696656-36696656 on Build GRCh38
rs894149003 Chr.2: 36696701-36696701 on Build GRCh38
rs747065888 Chr.2: 36696698-36696698 on Build GRCh38
rs112064376 Chr.2: 36696756-36696756 on Build GRCh38
rs953425796 Chr.2: 36696783-36696783 on Build GRCh38
rs375840274 Chr.2: 36697007-36697007 on Build GRCh38
rs1055718454 Chr.2: 36696902-36696902 on Build GRCh38
rs7563754 Chr.2: 36696897-36696897 on Build GRCh38
rs188658685 Chr.2: 36696762-36696762 on Build GRCh38
rs915538727 Chr.2: 36696941-36696941 on Build GRCh38
rs2160330 Chr.2: 36697034-36697034 on Build GRCh38
rs891178834 Chr.2: 36696740-36696740 on Build GRCh38
rs899700101 Chr.2: 36696722-36696722 on Build GRCh38
rs920495126 Chr.2: 36696674-36696674 on Build GRCh38
rs971113077 Chr.2: 36696841-36696841 on Build GRCh38
rs551541292 Chr.2: 36697012-36697012 on Build GRCh38
rs1014382428 Chr.2: 36696718-36696718 on Build GRCh38
rs76153919 Chr.2: 36696653-36696653 on Build GRCh38
rs562812758 Chr.2: 36696893-36696893 on Build GRCh38
rs67717359 Chr.2: 36696636-36696637 on Build GRCh38

View Full Product Details