Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69319888-69320418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT1
Entrez Gene ID
2673
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
UniGene
Hs.580300
Gene Chromosome Location
Chr.2: 69319769-69387254 on Build GRCh38
Transcripts
NM_001244710.1 NM_002056.3

SNP Detail

SNP ID Chromosome Location
rs997900583 Chr.2: 69320279-69320279 on Build GRCh38
rs1042208364 Chr.2: 69320197-69320197 on Build GRCh38
rs997558248 Chr.2: 69320036-69320036 on Build GRCh38
rs193289228 Chr.2: 69319998-69319998 on Build GRCh38
rs28613427 Chr.2: 69320067-69320067 on Build GRCh38
rs900606149 Chr.2: 69320278-69320278 on Build GRCh38
rs886056229 Chr.2: 69320063-69320063 on Build GRCh38
rs116374505 Chr.2: 69320290-69320290 on Build GRCh38
rs900495897 Chr.2: 69319921-69319921 on Build GRCh38
rs959280184 Chr.2: 69320152-69320152 on Build GRCh38
rs550960239 Chr.2: 69319988-69319988 on Build GRCh38
rs886656498 Chr.2: 69320312-69320312 on Build GRCh38
rs1044785 Chr.2: 69320392-69320392 on Build GRCh38
rs531253972 Chr.2: 69319973-69319973 on Build GRCh38
rs1039160457 Chr.2: 69319920-69319920 on Build GRCh38
rs553789635 Chr.2: 69319950-69319950 on Build GRCh38
rs945284205 Chr.2: 69320196-69320196 on Build GRCh38
rs950984771 Chr.2: 69320087-69320087 on Build GRCh38
rs143649075 Chr.2: 69320186-69320186 on Build GRCh38
rs1005093465 Chr.2: 69320328-69320328 on Build GRCh38
rs974227327 Chr.2: 69320120-69320120 on Build GRCh38
rs1039518444 Chr.2: 69320257-69320257 on Build GRCh38
rs573841882 Chr.2: 69320028-69320028 on Build GRCh38
rs7642 Chr.2: 69320104-69320104 on Build GRCh38
rs55888680 Chr.2: 69320095-69320095 on Build GRCh38
rs1016508768 Chr.2: 69320351-69320351 on Build GRCh38
rs184837165 Chr.2: 69320225-69320225 on Build GRCh38
rs991857353 Chr.2: 69320178-69320178 on Build GRCh38
rs886056230 Chr.2: 69320173-69320173 on Build GRCh38
rs926426868 Chr.2: 69320143-69320143 on Build GRCh38
rs749057215 Chr.2: 69320060-69320060 on Build GRCh38
rs1016394061 Chr.2: 69320103-69320103 on Build GRCh38
rs936554757 Chr.2: 69320244-69320244 on Build GRCh38
rs886056231 Chr.2: 69320288-69320288 on Build GRCh38
rs774631396 Chr.2: 69320325-69320325 on Build GRCh38
rs1030519030 Chr.2: 69320054-69320054 on Build GRCh38

View Full Product Details