Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73087958-73088455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP5
Entrez Gene ID
26056
Gene Name
RAB11 family interacting protein 5
Gene Aliases
GAF1, RIP11, pp75
UniGene
Hs.24557
Gene Chromosome Location
Chr.2: 73073382-73113018 on Build GRCh38
Transcripts
NM_015470.2 XM_005264251.3 XM_005264252.3 XM_005264253.4 XM_006711985.3 XM_011532753.2 XM_011532754.2 XM_011532755.2 XM_011532756.2 XM_017003788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2998118 c.1519A>T p.S507C Chr.2: 73088099-73088099 on Build GRCh38
5443792 c.1392C>T p.L464L Chr.2: 73088226-73088226 on Build GRCh38
2998121 c.1421G>A p.R474Q Chr.2: 73088197-73088197 on Build GRCh38
6893226 c.1257C>T p.H419H Chr.2: 73088361-73088361 on Build GRCh38
3972343 c.1550A>G p.D517G Chr.2: 73088068-73088068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771534256 Chr.2: 73088004-73088004 on Build GRCh38
rs780647922 Chr.2: 73088213-73088213 on Build GRCh38
rs902633860 Chr.2: 73088261-73088261 on Build GRCh38
rs142386441 Chr.2: 73088104-73088104 on Build GRCh38
rs531481854 Chr.2: 73088083-73088083 on Build GRCh38
rs780273439 Chr.2: 73088146-73088146 on Build GRCh38
rs760273170 Chr.2: 73088404-73088404 on Build GRCh38
rs372919327 Chr.2: 73088067-73088067 on Build GRCh38
rs145003108 Chr.2: 73088289-73088289 on Build GRCh38
rs781777356 Chr.2: 73088005-73088005 on Build GRCh38
rs572075758 Chr.2: 73088041-73088041 on Build GRCh38
rs200575843 Chr.2: 73088263-73088263 on Build GRCh38
rs746410144 Chr.2: 73088020-73088020 on Build GRCh38
rs763880279 Chr.2: 73088085-73088085 on Build GRCh38
rs760021988 Chr.2: 73088421-73088421 on Build GRCh38
rs148220987 Chr.2: 73088236-73088236 on Build GRCh38
rs748103648 Chr.2: 73088002-73088002 on Build GRCh38
rs145839919 Chr.2: 73088333-73088333 on Build GRCh38
rs369650590 Chr.2: 73088043-73088043 on Build GRCh38
rs760277863 Chr.2: 73088318-73088318 on Build GRCh38
rs59033238 Chr.2: 73088165-73088165 on Build GRCh38
rs963183027 Chr.2: 73088367-73088367 on Build GRCh38
rs772351421 Chr.2: 73088241-73088241 on Build GRCh38
rs768774709 Chr.2: 73088346-73088346 on Build GRCh38
rs925766122 Chr.2: 73088250-73088250 on Build GRCh38
rs752549381 Chr.2: 73088072-73088072 on Build GRCh38
rs766279734 Chr.2: 73088064-73088064 on Build GRCh38
rs761546586 Chr.2: 73088278-73088278 on Build GRCh38
rs373397434 Chr.2: 73088221-73088221 on Build GRCh38
rs773150895 Chr.2: 73088244-73088244 on Build GRCh38
rs1023911671 Chr.2: 73088341-73088341 on Build GRCh38
rs536535862 Chr.2: 73088339-73088339 on Build GRCh38
rs772098840 Chr.2: 73088168-73088168 on Build GRCh38
rs775520238 Chr.2: 73088257-73088257 on Build GRCh38
rs1001677387 Chr.2: 73088360-73088360 on Build GRCh38
rs767270562 Chr.2: 73088303-73088303 on Build GRCh38
rs779188928 Chr.2: 73088323-73088323 on Build GRCh38
rs764004895 Chr.2: 73088182-73088182 on Build GRCh38
rs969788701 Chr.2: 73088137-73088137 on Build GRCh38
rs916758480 Chr.2: 73088156-73088156 on Build GRCh38
rs749873719 Chr.2: 73088306-73088306 on Build GRCh38
rs150741859 Chr.2: 73088246-73088246 on Build GRCh38
rs1032590031 Chr.2: 73088118-73088118 on Build GRCh38
rs201625360 Chr.2: 73088176-73088176 on Build GRCh38
rs773343987 Chr.2: 73088175-73088175 on Build GRCh38
rs369090487 Chr.2: 73088350-73088350 on Build GRCh38
rs753357623 Chr.2: 73088411-73088411 on Build GRCh38
rs372194178 Chr.2: 73088028-73088028 on Build GRCh38
rs747218615 Chr.2: 73088336-73088336 on Build GRCh38
rs763017426 Chr.2: 73088258-73088258 on Build GRCh38
rs747179362 Chr.2: 73088245-73088245 on Build GRCh38
rs994434150 Chr.2: 73088403-73088403 on Build GRCh38
rs761712902 Chr.2: 73088365-73088365 on Build GRCh38
rs754300332 Chr.2: 73088321-73088321 on Build GRCh38
rs144951768 Chr.2: 73088342-73088342 on Build GRCh38
rs990948491 Chr.2: 73088150-73088150 on Build GRCh38
rs138258802 Chr.2: 73088179-73088179 on Build GRCh38
rs747892372 Chr.2: 73088401-73088401 on Build GRCh38
rs775116698 Chr.2: 73088177-73088177 on Build GRCh38
rs138135562 Chr.2: 73088407-73088407 on Build GRCh38
rs201414059 Chr.2: 73088042-73088042 on Build GRCh38
rs143156025 Chr.2: 73088183-73088183 on Build GRCh38
rs1052800374 Chr.2: 73088066-73088066 on Build GRCh38
rs140475519 Chr.2: 73088131-73088131 on Build GRCh38
rs1048086493 Chr.2: 73088190-73088190 on Build GRCh38
rs763916913 Chr.2: 73088259-73088259 on Build GRCh38
rs780787094 Chr.2: 73088114-73088114 on Build GRCh38
rs958242250 Chr.2: 73088310-73088310 on Build GRCh38
rs750011597 Chr.2: 73088193-73088193 on Build GRCh38
rs199945628 Chr.2: 73088237-73088237 on Build GRCh38
rs764872096 Chr.2: 73088071-73088071 on Build GRCh38
rs971289714 Chr.2: 73088347-73088347 on Build GRCh38
rs967097210 Chr.2: 73088100-73088100 on Build GRCh38
rs748301203 Chr.2: 73088238-73088238 on Build GRCh38
rs929680119 Chr.2: 73087988-73087988 on Build GRCh38
rs561703908 Chr.2: 73088338-73088338 on Build GRCh38
rs774600602 Chr.2: 73088164-73088164 on Build GRCh38
rs771070499 Chr.2: 73088268-73088270 on Build GRCh38
rs751092757 Chr.2: 73088092-73088092 on Build GRCh38
rs376039550 Chr.2: 73088154-73088154 on Build GRCh38
rs756977990 Chr.2: 73088192-73088192 on Build GRCh38
rs765846322 Chr.2: 73088409-73088409 on Build GRCh38
rs17857040 Chr.2: 73088110-73088110 on Build GRCh38
rs373507575 Chr.2: 73088065-73088065 on Build GRCh38
rs749724196 Chr.2: 73088148-73088148 on Build GRCh38
rs774406646 Chr.2: 73088060-73088060 on Build GRCh38
rs140983118 Chr.2: 73088328-73088328 on Build GRCh38
rs765798618 Chr.2: 73088430-73088430 on Build GRCh38
rs772868278 Chr.2: 73088382-73088382 on Build GRCh38
rs768832166 Chr.2: 73088159-73088159 on Build GRCh38
rs762617959 Chr.2: 73088348-73088348 on Build GRCh38
rs533920101 Chr.2: 73088208-73088209 on Build GRCh38
rs142576427 Chr.2: 73088198-73088198 on Build GRCh38
rs753117895 Chr.2: 73088325-73088325 on Build GRCh38
rs200150559 Chr.2: 73088140-73088140 on Build GRCh38
rs771598578 Chr.2: 73088370-73088370 on Build GRCh38

View Full Product Details