Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86807366-86807866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8A
Entrez Gene ID
925
Gene Name
CD8a molecule
Gene Aliases
CD8, Leu2, MAL, p32
UniGene
Hs.85258
Gene Chromosome Location
Chr.2: 86784605-86808396 on Build GRCh38
Transcripts
NM_001145873.1

SNP Detail

SNP ID Chromosome Location
rs1036513121 Chr.2: 86807538-86807538 on Build GRCh38
rs997658355 Chr.2: 86807551-86807551 on Build GRCh38
rs1029895526 Chr.2: 86807507-86807507 on Build GRCh38
rs772420403 Chr.2: 86807800-86807800 on Build GRCh38
rs532921302 Chr.2: 86807442-86807442 on Build GRCh38
rs748064543 Chr.2: 86807528-86807528 on Build GRCh38
rs984806420 Chr.2: 86807751-86807751 on Build GRCh38
rs1030765346 Chr.2: 86807698-86807698 on Build GRCh38
rs1006505090 Chr.2: 86807636-86807636 on Build GRCh38
rs74455479 Chr.2: 86807434-86807434 on Build GRCh38
rs59013526 Chr.2: 86807653-86807653 on Build GRCh38
rs923360684 Chr.2: 86807794-86807794 on Build GRCh38
rs960503151 Chr.2: 86807702-86807702 on Build GRCh38
rs1050588747 Chr.2: 86807846-86807846 on Build GRCh38
rs940810155 Chr.2: 86807537-86807537 on Build GRCh38
rs940771980 Chr.2: 86807769-86807769 on Build GRCh38
rs558609330 Chr.2: 86807668-86807668 on Build GRCh38
rs889373813 Chr.2: 86807630-86807630 on Build GRCh38
rs72936513 Chr.2: 86807432-86807432 on Build GRCh38
rs985452217 Chr.2: 86807562-86807562 on Build GRCh38
rs933458232 Chr.2: 86807837-86807837 on Build GRCh38
rs962749864 Chr.2: 86807633-86807633 on Build GRCh38
rs1042551318 Chr.2: 86807782-86807782 on Build GRCh38
rs553658359 Chr.2: 86807819-86807819 on Build GRCh38
rs868704065 Chr.2: 86807828-86807828 on Build GRCh38
rs887859435 Chr.2: 86807530-86807530 on Build GRCh38
rs947995304 Chr.2: 86807518-86807518 on Build GRCh38
rs1001193560 Chr.2: 86807404-86807404 on Build GRCh38
rs977950711 Chr.2: 86807443-86807443 on Build GRCh38
rs954215903 Chr.2: 86807542-86807542 on Build GRCh38
rs1029270585 Chr.2: 86807396-86807396 on Build GRCh38
rs377360561 Chr.2: 86807695-86807695 on Build GRCh38
rs55814938 Chr.2: 86807494-86807494 on Build GRCh38
rs577970475 Chr.2: 86807786-86807786 on Build GRCh38
rs916518149 Chr.2: 86807509-86807509 on Build GRCh38
rs1015819190 Chr.2: 86807516-86807516 on Build GRCh38
rs890041490 Chr.2: 86807814-86807819 on Build GRCh38
rs113290125 Chr.2: 86807659-86807659 on Build GRCh38
rs909227967 Chr.2: 86807759-86807759 on Build GRCh38
rs927223541 Chr.2: 86807720-86807720 on Build GRCh38
rs892894687 Chr.2: 86807757-86807757 on Build GRCh38
rs573092420 Chr.2: 86807402-86807402 on Build GRCh38
rs939238354 Chr.2: 86807483-86807483 on Build GRCh38
rs529189652 Chr.2: 86807425-86807425 on Build GRCh38
rs902037624 Chr.2: 86807540-86807540 on Build GRCh38
rs999711980 Chr.2: 86807692-86807692 on Build GRCh38
rs1029162670 Chr.2: 86807556-86807556 on Build GRCh38
rs916464734 Chr.2: 86807729-86807729 on Build GRCh38
rs923796242 Chr.2: 86807455-86807455 on Build GRCh38
rs918396901 Chr.2: 86807645-86807645 on Build GRCh38
rs1054164613 Chr.2: 86807406-86807406 on Build GRCh38
rs934586472 Chr.2: 86807737-86807738 on Build GRCh38
rs569622108 Chr.2: 86807548-86807548 on Build GRCh38
rs756565355 Chr.2: 86807393-86807393 on Build GRCh38
rs981791014 Chr.2: 86807665-86807665 on Build GRCh38
rs114045589 Chr.2: 86807722-86807722 on Build GRCh38
rs945770794 Chr.2: 86807775-86807775 on Build GRCh38
rs772900028 Chr.2: 86807746-86807746 on Build GRCh38
rs1022421088 Chr.2: 86807643-86807643 on Build GRCh38
rs781055588 Chr.2: 86807480-86807480 on Build GRCh38

View Full Product Details