Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102762471-102762980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM182
Entrez Gene ID
130827
Gene Name
transmembrane protein 182
Gene Aliases
-
UniGene
Hs.436203
Gene Chromosome Location
Chr.2: 102736906-102843838 on Build GRCh38
Transcripts
NM_001321343.1 NM_001321344.1 NM_001321345.1 NM_001321346.1 NM_144632.4 XM_006712287.1 XM_006712288.3 XM_011510632.2 XM_017003375.1 XM_017003376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982868 c.151C>T p.H51Y Chr.2: 102762605-102762605 on Build GRCh38
5859801 c.160G>A p.G54R Chr.2: 102762614-102762614 on Build GRCh38
1004722 c.142G>A p.V48I Chr.2: 102762596-102762596 on Build GRCh38
6489076 c.232A>G p.T78A Chr.2: 102762686-102762686 on Build GRCh38
7394190 c.200A>C p.E67A Chr.2: 102762654-102762654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562929340 Chr.2: 102762696-102762696 on Build GRCh38
rs17027923 Chr.2: 102762840-102762840 on Build GRCh38
rs570162702 Chr.2: 102762643-102762643 on Build GRCh38
rs759480151 Chr.2: 102762701-102762701 on Build GRCh38
rs760851927 Chr.2: 102762694-102762694 on Build GRCh38
rs777657337 Chr.2: 102762639-102762639 on Build GRCh38
rs1040651842 Chr.2: 102762655-102762655 on Build GRCh38
rs146395435 Chr.2: 102762754-102762754 on Build GRCh38
rs372309265 Chr.2: 102762704-102762704 on Build GRCh38
rs774764380 Chr.2: 102762552-102762552 on Build GRCh38
rs201874052 Chr.2: 102762583-102762583 on Build GRCh38
rs912647367 Chr.2: 102762804-102762804 on Build GRCh38
rs974273434 Chr.2: 102762756-102762756 on Build GRCh38
rs144693740 Chr.2: 102762635-102762635 on Build GRCh38
rs531010291 Chr.2: 102762664-102762664 on Build GRCh38
rs778026109 Chr.2: 102762938-102762938 on Build GRCh38
rs748717977 Chr.2: 102762669-102762669 on Build GRCh38
rs765365774 Chr.2: 102762706-102762706 on Build GRCh38
rs200283037 Chr.2: 102762599-102762599 on Build GRCh38
rs543124067 Chr.2: 102762868-102762868 on Build GRCh38
rs762025988 Chr.2: 102762553-102762553 on Build GRCh38
rs1054337572 Chr.2: 102762855-102762855 on Build GRCh38
rs939256465 Chr.2: 102762846-102762846 on Build GRCh38
rs944519191 Chr.2: 102762637-102762637 on Build GRCh38
rs756965995 Chr.2: 102762734-102762734 on Build GRCh38
rs766614241 Chr.2: 102762504-102762504 on Build GRCh38
rs948008266 Chr.2: 102762917-102762917 on Build GRCh38
rs367596735 Chr.2: 102762791-102762791 on Build GRCh38
rs773381180 Chr.2: 102762558-102762558 on Build GRCh38
rs931173661 Chr.2: 102762767-102762767 on Build GRCh38
rs754254177 Chr.2: 102762523-102762523 on Build GRCh38
rs80137523 Chr.2: 102762816-102762816 on Build GRCh38
rs1013003115 Chr.2: 102762713-102762713 on Build GRCh38
rs1026342812 Chr.2: 102762718-102762718 on Build GRCh38
rs552846347 Chr.2: 102762665-102762665 on Build GRCh38
rs763961790 Chr.2: 102762720-102762720 on Build GRCh38
rs139462348 Chr.2: 102762929-102762929 on Build GRCh38
rs754138139 Chr.2: 102762567-102762568 on Build GRCh38
rs762972048 Chr.2: 102762717-102762717 on Build GRCh38
rs746613840 Chr.2: 102762642-102762642 on Build GRCh38
rs183362403 Chr.2: 102762759-102762759 on Build GRCh38
rs370445639 Chr.2: 102762596-102762596 on Build GRCh38
rs191482296 Chr.2: 102762541-102762541 on Build GRCh38
rs536273324 Chr.2: 102762793-102762793 on Build GRCh38
rs985870367 Chr.2: 102762783-102762783 on Build GRCh38
rs546101131 Chr.2: 102762750-102762750 on Build GRCh38
rs752540475 Chr.2: 102762715-102762715 on Build GRCh38
rs895166905 Chr.2: 102762882-102762882 on Build GRCh38
rs367815136 Chr.2: 102762547-102762547 on Build GRCh38
rs540932130 Chr.2: 102762595-102762595 on Build GRCh38
rs975921056 Chr.2: 102762822-102762822 on Build GRCh38
rs201534618 Chr.2: 102762630-102762630 on Build GRCh38
rs148918467 Chr.2: 102762685-102762685 on Build GRCh38
rs751440468 Chr.2: 102762727-102762727 on Build GRCh38
rs954015285 Chr.2: 102762769-102762769 on Build GRCh38
rs532234987 Chr.2: 102762566-102762566 on Build GRCh38
rs745438301 Chr.2: 102762654-102762654 on Build GRCh38
rs772273346 Chr.2: 102762673-102762673 on Build GRCh38
rs780818921 Chr.2: 102762653-102762653 on Build GRCh38
rs368116221 Chr.2: 102762699-102762699 on Build GRCh38
rs964321852 Chr.2: 102762790-102762790 on Build GRCh38
rs377087257 Chr.2: 102762735-102762735 on Build GRCh38
rs1038892386 Chr.2: 102762921-102762921 on Build GRCh38
rs924414876 Chr.2: 102762825-102762825 on Build GRCh38
rs570621038 Chr.2: 102762786-102762787 on Build GRCh38
rs746048191 Chr.2: 102762688-102762689 on Build GRCh38
rs778010661 Chr.2: 102762778-102762778 on Build GRCh38
rs750104995 Chr.2: 102762738-102762738 on Build GRCh38
rs565523571 Chr.2: 102762770-102762770 on Build GRCh38
rs375329411 Chr.2: 102762851-102762851 on Build GRCh38
rs779250020 Chr.2: 102762601-102762602 on Build GRCh38
rs898564340 Chr.2: 102762923-102762923 on Build GRCh38
rs753172269 Chr.2: 102762571-102762571 on Build GRCh38
rs766420946 Chr.2: 102762570-102762570 on Build GRCh38
rs977948901 Chr.2: 102762830-102762830 on Build GRCh38

View Full Product Details