Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127700851-127701379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFT2D3
Entrez Gene ID
84826
Gene Name
SFT2 domain containing 3
Gene Aliases
-
UniGene
Hs.345849
Gene Chromosome Location
Chr.2: 127701023-127703833 on Build GRCh38
Transcripts
NM_032740.3

SNP Detail

SNP ID Chromosome Location
rs930921269 Chr.2: 127700910-127700910 on Build GRCh38
rs980060625 Chr.2: 127701204-127701204 on Build GRCh38
rs1029103113 Chr.2: 127701080-127701080 on Build GRCh38
rs767217238 Chr.2: 127701120-127701120 on Build GRCh38
rs1044413697 Chr.2: 127700900-127700900 on Build GRCh38
rs530942603 Chr.2: 127701203-127701203 on Build GRCh38
rs1040605614 Chr.2: 127701306-127701306 on Build GRCh38
rs760098968 Chr.2: 127701001-127701001 on Build GRCh38
rs536093657 Chr.2: 127701249-127701249 on Build GRCh38
rs1030919182 Chr.2: 127701121-127701121 on Build GRCh38
rs991985769 Chr.2: 127701138-127701138 on Build GRCh38
rs919901771 Chr.2: 127701353-127701353 on Build GRCh38
rs559742566 Chr.2: 127701101-127701101 on Build GRCh38
rs1049333658 Chr.2: 127700920-127700920 on Build GRCh38
rs1032368258 Chr.2: 127700877-127700877 on Build GRCh38
rs765791597 Chr.2: 127701193-127701193 on Build GRCh38
rs754875300 Chr.2: 127701313-127701313 on Build GRCh38
rs956709161 Chr.2: 127701137-127701137 on Build GRCh38
rs567160633 Chr.2: 127701199-127701199 on Build GRCh38
rs930681651 Chr.2: 127701215-127701215 on Build GRCh38
rs529579103 Chr.2: 127700928-127700928 on Build GRCh38
rs949197347 Chr.2: 127701170-127701170 on Build GRCh38
rs543367008 Chr.2: 127701082-127701082 on Build GRCh38
rs190919874 Chr.2: 127701325-127701325 on Build GRCh38
rs552859123 Chr.2: 127701310-127701310 on Build GRCh38
rs973155974 Chr.2: 127701343-127701343 on Build GRCh38
rs986889046 Chr.2: 127700968-127700968 on Build GRCh38
rs536924852 Chr.2: 127701150-127701150 on Build GRCh38
rs995841251 Chr.2: 127701066-127701066 on Build GRCh38
rs902246661 Chr.2: 127701195-127701195 on Build GRCh38
rs13389788 Chr.2: 127701244-127701244 on Build GRCh38
rs183168496 Chr.2: 127701202-127701202 on Build GRCh38
rs13392473 Chr.2: 127701243-127701243 on Build GRCh38
rs77162475 Chr.2: 127700902-127700902 on Build GRCh38
rs954247860 Chr.2: 127701309-127701309 on Build GRCh38
rs916376650 Chr.2: 127701166-127701166 on Build GRCh38
rs117566639 Chr.2: 127701161-127701161 on Build GRCh38
rs763183461 Chr.2: 127701100-127701100 on Build GRCh38
rs972742600 Chr.2: 127701032-127701032 on Build GRCh38
rs940047490 Chr.2: 127701024-127701031 on Build GRCh38
rs903476665 Chr.2: 127701319-127701319 on Build GRCh38
rs940832328 Chr.2: 127701277-127701277 on Build GRCh38
rs886585634 Chr.2: 127701000-127701000 on Build GRCh38
rs770324188 Chr.2: 127701018-127701018 on Build GRCh38
rs1028529641 Chr.2: 127701302-127701302 on Build GRCh38
rs927260598 Chr.2: 127701212-127701212 on Build GRCh38
rs190082856 Chr.2: 127701171-127701171 on Build GRCh38
rs777337053 Chr.2: 127700973-127700973 on Build GRCh38
rs201020714 Chr.2: 127701224-127701233 on Build GRCh38
rs901628067 Chr.2: 127701052-127701052 on Build GRCh38
rs542771729 Chr.2: 127700965-127700965 on Build GRCh38
rs187851210 Chr.2: 127701318-127701318 on Build GRCh38
rs749979970 Chr.2: 127701154-127701154 on Build GRCh38
rs1028414922 Chr.2: 127700923-127700923 on Build GRCh38

View Full Product Details