Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135840394-135840905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM6
Entrez Gene ID
4175
Gene Name
minichromosome maintenance complex component 6
Gene Aliases
MCG40308, Mis5, P105MCM
UniGene
Hs.444118
Gene Chromosome Location
Chr.2: 135839626-135876477 on Build GRCh38
Transcripts
NM_005915.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209049 c.2433G>T p.L811F Chr.2: 135840868-135840868 on Build GRCh38
6087502 c.2402G>A p.G801E Chr.2: 135840899-135840899 on Build GRCh38
441169 c.2465G>C p.*822S Chr.2: 135840836-135840836 on Build GRCh38
5937004 c.2426C>T p.P809L Chr.2: 135840875-135840875 on Build GRCh38
5732280 c.2411G>C p.S804T Chr.2: 135840890-135840890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187760399 Chr.2: 135840568-135840568 on Build GRCh38
rs2070069 Chr.2: 135840787-135840787 on Build GRCh38
rs772529167 Chr.2: 135840505-135840505 on Build GRCh38
rs1804609 Chr.2: 135840858-135840858 on Build GRCh38
rs894880032 Chr.2: 135840579-135840579 on Build GRCh38
rs142645038 Chr.2: 135840621-135840621 on Build GRCh38
rs943910412 Chr.2: 135840499-135840499 on Build GRCh38
rs936479247 Chr.2: 135840556-135840556 on Build GRCh38
rs201030245 Chr.2: 135840493-135840494 on Build GRCh38
rs113659730 Chr.2: 135840633-135840633 on Build GRCh38
rs904749516 Chr.2: 135840615-135840615 on Build GRCh38
rs530013823 Chr.2: 135840728-135840728 on Build GRCh38
rs953431380 Chr.2: 135840425-135840425 on Build GRCh38
rs200231978 Chr.2: 135840843-135840843 on Build GRCh38
rs1027749617 Chr.2: 135840815-135840815 on Build GRCh38
rs752428110 Chr.2: 135840876-135840876 on Build GRCh38
rs923733181 Chr.2: 135840544-135840544 on Build GRCh38
rs199970908 Chr.2: 135840785-135840785 on Build GRCh38
rs911715052 Chr.2: 135840437-135840437 on Build GRCh38
rs762431547 Chr.2: 135840844-135840844 on Build GRCh38
rs540325321 Chr.2: 135840583-135840583 on Build GRCh38
rs55660827 Chr.2: 135840873-135840873 on Build GRCh38
rs3087346 Chr.2: 135840514-135840514 on Build GRCh38
rs3087350 Chr.2: 135840746-135840746 on Build GRCh38
rs766711168 Chr.2: 135840569-135840570 on Build GRCh38
rs749609682 Chr.2: 135840797-135840798 on Build GRCh38
rs11656 Chr.2: 135840481-135840481 on Build GRCh38
rs760788163 Chr.2: 135840591-135840593 on Build GRCh38
rs1018915892 Chr.2: 135840760-135840760 on Build GRCh38
rs761190719 Chr.2: 135840791-135840791 on Build GRCh38
rs912481863 Chr.2: 135840491-135840491 on Build GRCh38
rs190104398 Chr.2: 135840775-135840775 on Build GRCh38
rs899375084 Chr.2: 135840663-135840663 on Build GRCh38
rs779003323 Chr.2: 135840605-135840605 on Build GRCh38
rs376372188 Chr.2: 135840610-135840610 on Build GRCh38
rs1027560039 Chr.2: 135840423-135840423 on Build GRCh38
rs771809754 Chr.2: 135840786-135840786 on Build GRCh38
rs184828444 Chr.2: 135840706-135840706 on Build GRCh38
rs1029521149 Chr.2: 135840720-135840720 on Build GRCh38
rs3087347 Chr.2: 135840430-135840430 on Build GRCh38
rs755487780 Chr.2: 135840602-135840602 on Build GRCh38
rs755836471 Chr.2: 135840878-135840878 on Build GRCh38
rs976796254 Chr.2: 135840492-135840492 on Build GRCh38
rs1014468684 Chr.2: 135840745-135840745 on Build GRCh38
rs747293176 Chr.2: 135840805-135840805 on Build GRCh38
rs750554384 Chr.2: 135840857-135840857 on Build GRCh38
rs1029074832 Chr.2: 135840440-135840440 on Build GRCh38
rs113071798 Chr.2: 135840710-135840710 on Build GRCh38
rs994997973 Chr.2: 135840667-135840667 on Build GRCh38
rs192736265 Chr.2: 135840703-135840703 on Build GRCh38
rs765368148 Chr.2: 135840850-135840850 on Build GRCh38

View Full Product Details